Dry Blend Buffer Packs
Thermo Scientific™ BupH™ Phosphate Buffered Saline Packs
Make 500mL of sodium phosphate buffer (PBS) for crosslinking and labeling reactions requiring amine-free conditions with these dry-blended powder pouches.
Thermo Scientific™ BupH™ Modified Dulbecco's PBS Packs
Make 500mL of Na- and K-phosphate buffered saline (D-PBS) for ELISA and Western blot diluents and wash buffers with these dry-blended powder pouches.
Thermo Scientific™ BupH™ MES Buffered Saline Packs
Make 500mL of optimized amine- and carboxyl-free buffer for carbodiimide (EDC) crosslinking and conjugation reactions with these dry-blended powder pouches.
Thermo Scientific™ BupH™ Tris Buffered Saline Packs
Make 500mL of Tris-NaCl buffer (TBS) for diluents and wash buffers in ELISA, Western blotting and other immunoassays with these dry-blended powder pouches.
Thermo Scientific™ BupH™ Tris-HEPES-SDS Running Buffer Packs
Make 500mL of Tris-NaCl buffer (TBS) for diluents and wash buffers in ELISA, Western blotting and other immunoassays with these dry-blended powder pouches.
Thermo Scientific™ BupH™ Borate Buffer Packs
Make 500mL of borate buffer for protein modification procedures requiring amine-free conditions and alkaline pH with these dry-blended powder pouches.
Thermo Scientific™ BupH™ Tris-Glycine-SDS Buffer Packs
Make 500mL of standard (pH 8.3) running buffer for SDS-PAGE (protein polyacrylamide gel electrophoresis) with these dry-blended powder pouches.
Thermo Scientific™ BupH™ Carbonate-Bicarbonate Buffer Packs
Make 500mL of alkaline (pH 9.4) Na-carbonate (carb-bicarb) buffer for protein and antibody coating procedures with these dry-blended powder pouches.
Thermo Scientific™ BupH™ Tris-Glycine Transfer Buffer Packs
Make 500mL of standard electrophoresis running buffer for wet or semi-dry Western blot transfer with these dry-blended powder pouches.
  spinner