Ether -Diethyl Ether
Diethyl ether, ACS reagent, anhydrous, ACROS Organics™
Chemical Name or Material: Diethyl ether CAS: 60-29-7 Purity Grade: ACS Reagent Grade Assay: ≥99.0% Molecular Formula: C4H10O Linear Formula: (CH3CH2)2O Formula Weight: 74.12 MDL Number: MFCD00011646 Synonym: Ether
Diethyl ether, +99%, extra pure, stabilized with BHT, ACROS Organics™ Available on GSA/VA Contract for Federal Government customers only.
Chemical Name or Material: Diethyl ether Name Note: Stabilized with BHT CAS: 60-29-7 Purity Grade: Extra Pure Assay: ≥99.0% Assay Percent Range: 99+% Molecular Formula: C4H10O Linear Formula: (CH3CH2)2O Formula Weight: 74.12 MDL Number: MFCD00011646 Synonym: Ether
Diethyl ether, >99%, inhibitor free, Alfa Aesar™
Chemical Name or Material: Diethyl ether CAS: 60-29-7 Purity Grade: Spectrophotometric grade Physical Form: Liquid Assay Percent Range: >99% Molecular Formula: C4H10O Formula Weight: 74.12 MDL Number: MFCD00011646 Synonym: Ether
Ethyl Ether Anhydrous (BHT Stabilized/Certified ACS), Fisher Chemical
Chemical Name or Material: Ethyl Ether Anhydrous Name Note: BHT Stabilized CAS: 60-29-7 Purity Grade: Certified ACS Reagent Grade Physical Form: Liquid Assay: ≥99% Assay Percent Range: ≥99 % Molecular Formula: C4H10O Linear Formula: C2H5OC2H5 Formula Weight: 74.12 MDL Number: MFCD00011646 Synonym: Diethyl Ether
Diethyl Ether, 99.5%, Extra Dry, Stabilized, AcroSeal™, Case of 4 x 25mL bottles, ACROS Organics™ Available on GSA/VA Contract for Federal Government customers only.
Chemical Name or Material: Diethyl ether CAS: 60-29-7 Purity Grade: Extra Dry Purity Grade Notes: Water <50ppm Assay: ≥99.4% (GC) Assay Percent Range: 99.5% Molecular Formula: C4H10O Linear Formula: (CH3CH2)2O Formula Weight: 74.12 MDL Number: MFCD00011646 Synonym: Ether
Ethyl Ether (Spectranalyzed™), Fisher Chemical Available on GSA/VA Contract for Federal Government customers only.
Chemical Name or Material: Ethyl Ether CAS: 60-29-7 CAS: 64-17-5 Purity Grade: Spectranalyzed™ Assay: ≥99% Assay Percent Range: ≥99 % Molecular Formula: C4H10O Linear Formula: C2H5OC2H5 Formula Weight: 74.12 MDL Number: MFCD00011646 Synonym: Diethyl Ether
Ethyl Ether (Pesticide), Fisher Chemical
Chemical Name or Material: Ethyl Ether CAS: 60-29-7 CAS: 64-17-5 Purity Grade: Pesticide Grade Assay: ≥99.5% Assay Percent Range: ≥99.5 % Molecular Formula: C4H10O Linear Formula: C2H5OC2H5 Formula Weight: 74.12 MDL Number: MFCD00011646 Synonym: ,
Diethyl ether, 99.5%, Extra Dry, stabilized, AcroSeal™, ACROS Organics™ Available on GSA/VA Contract for Federal Government customers only.
Chemical Name or Material: Diethyl ether CAS: 60-29-7 Purity Grade: Extra Dry Purity Grade Notes: Water <50ppm Assay: ≥99.4% (GC) Assay Percent Range: 99.5% Molecular Formula: C4H10O Linear Formula: (CH3CH2)2O Formula Weight: 74.12 MDL Number: MFCD00011646 Synonym: Ether
Ethyl Ether (Laboratory), Fat Extraction, Fisher Chemical
Chemical Name or Material: Ethyl Ether CAS: 60-29-7 Purity Grade: Laboratory Grade Purity Grade Notes: Suitable for fat extraction Molecular Formula: C4H10O Linear Formula: C2H5OC2H5 Formula Weight: 74.12 MDL Number: MFCD00011646 Synonym: Diethyl Ether
Diethyl ether, 99.5%, for analysis, stabilized with BHT, conforms to Eur.Ph., ACROS Organics™ Available on GSA/VA Contract for Federal Government customers only.
Chemical Name or Material: Diethyl ether Name Note: Stabilized with BHT CAS: 60-29-7 Purity Grade: For analysis Assay: ≥99.5% Assay Percent Range: 99.5% Molecular Formula: C4H10O Linear Formula: (CH3CH2)2O Formula Weight: 74.12 MDL Number: MFCD00011646 Synonym: Ether
Diethyl ether, 99+%, for spectroscopy, stabilized with ethanol, ACROS Organics™
Chemical Name or Material: Diethyl ether Name Note: Stabilized with ethanol, for spectroscopy CAS: 60-29-7 Purity Grade: For spectroscopy Assay: ≥99.0% (ex stabiliser) Assay Percent Range: 99+% Molecular Formula: C4H10O Linear Formula: (CH3CH2)2O Formula Weight: 74.12 MDL Number: MFCD00011646 Synonym: Ether
Diethyl ether, 99%, Alfa Aesar™
Chemical Name or Material: Diethyl ether CAS: 60-29-7 Physical Form: Liquid Assay Percent Range: 99% Molecular Formula: (C2H5)2O Formula Weight: 74.12 MDL Number: MFCD00011646
Ethyl Ether (Laboratory), Fisher Chemical
Chemical Name or Material: Ethyl Ether CAS: 60-29-7 Purity Grade: Laboratory Grade Molecular Formula: C4H10O Linear Formula: C2H5OC2H5 Formula Weight: 74.12 MDL Number: MFCD00011646 Synonym: Ethyl Oxide
Diethyl ether, 99.5%, Extra Dry over Molecular Sieve, Stabilized, AcroSeal™, ACROS Organics™ Available on GSA/VA Contract for Federal Government customers only.
Chemical Name or Material: Diethyl ether Name Note: Water <50 ppm, stabilized with BHT CAS: 60-29-7 Purity Grade: Extra Dry over Molecular Sieve Assay: ≥99.4% (GC) Assay Percent Range: 99.5% Molecular Formula: C4H10O Linear Formula: (CH3CH2)2O Formula Weight: 74.12 MDL Number: MFCD00011646 Synonym: Ether
Diethyl ether, 99+%, pure, stabilized with BHT, ACROS Organics™ Available on GSA/VA Contract for Federal Government customers only.
Chemical Name or Material: Diethyl ether CAS: 60-29-7 Purity Grade: Pure Assay: ≥99.0% (GC) Assay Percent Range: 99+% Molecular Formula: C4H10O Linear Formula: (CH3CH2)2O Formula Weight: 74.12 MDL Number: MFCD00011646 Synonym: Ether
Ethyl Ether, Anhydrous (Stabilized/HPLC), Fisher Chemical
Chemical Name or Material: Ethyl Ether Anhydrous Name Note: Stabilized CAS: 60-29-7 CAS: 64-17-5 Purity Grade: HPLC Grade Purity Grade Notes: Mobile phase for HPLC applications Physical Form: Liquid Assay: ≥99% Assay Percent Range: 99.0% min by GC Molecular Formula: C4H10O Linear Formula: C2H5OC2H5 Formula Weight: 74.12 MDL Number: MFCD00011646 Synonym: Diethyl Ether
Diethyl ether, for HPLC, stabilized, ACROS Organics™
Chemical Name or Material: Diethyl ether Name Note: Stabilized CAS: 60-29-7 Purity Grade: For HPLC Assay: ≥98% Molecular Formula: C4H10O Linear Formula: (CH3CH2)2O Formula Weight: 74.12 MDL Number: MFCD00011646 Synonym: Ether
Diethyl ether, anhydrous, ACS, 99% min, stab. with BHT
Chemical Name or Material: Diethyl Ether CAS: 60-29-7 Name Note: For ACS analysis Purity Grade: ACS Grade Molecular Formula: (C2H5)2O Linear Formula: (C2H5)2O Formula Weight: 74.12 MDL Number: MFCD00011646 UN Number: UN1155 Merck Index: 14,3806
Ethyl Ether, Anhydrous, ACS Grade
Chemical Name or Material: Ethyl Ether Anhydrous CAS: 60-29-7 Molecular Formula: (C2H5)2O
Ether ChromAR, Macron Fine Chemicals™
Chemical Name or Material: Ether CAS: 60-29-7 Molecular Formula: (C2H5)2O Formula Weight: 74.12
Ether USP, Macron Fine Chemicals™
Chemical Name or Material: Ether CAS: 60-29-7 Purity Grade: BP/EP/FCC/JP/NF/USP Molecular Formula: (C2H5)2O Formula Weight: 74.12
Ethyl Ether ACS AR, Macron Fine Chemicals™
Chemical Name or Material: Ethyl Ether CAS: 60-29-7 Purity Grade: ACS Reagent Grade Molecular Formula: (C2H5)2O Formula Weight: 74.12
Ether UltimAR, Macron Fine Chemicals™
Chemical Name or Material: Ether CAS: 60-29-7 Molecular Formula: (C2H5)2O Formula Weight: 74.12
Ether, ULTRA RESI-ANALYZED, J.T.Baker™
Chemical Name or Material: Ether CAS: 60-29-7 Purity Grade: ULTRA RESI-ANALYZED™ Assay Percent Range: 99.0 % min. Molecular Formula: (C2H5)2O Formula Weight: 74.12
Ether, Anhydrous, min. 99.0%, Baker Analyzed ACS Reagent, J.T.Baker™
CAS: 60-29-7 Purity Grade: Baker Analyzed™ ACS Reagent Physical Form: Liquid Assay Percent Range: min. 99.0%
Ethyl Ether, Anhydrous, Macron Fine Chemicals™
Chemical Name or Material: Ethyl Ether CAS: 60-29-7 Physical Form: Anhydrous Formula Weight: 74.12
Ethyl Ether, Anhydrous, 99.0% min., ACS, EMD Millipore Available on GSA/VA Contract for Federal Government customers only.
Chemical Name or Material: Ethyl Ether, Anhydrous Name Note: For ACS analysis CAS: 60-29-7 Physical Form: Liquid Assay Percent Range: 99.0% min. Molecular Formula: (CH3CH2)2O Formula Weight: 74.12 Synonym: Diethyl Ether
Ethyl Ether, Anhydrous (ACS), EMD Millipore Available on GSA/VA Contract for Federal Government customers only.
Chemical Name or Material: Ethyl Ether, Anhydrous (ACS) CAS: 60-29-7 Purity Grade: ACS Reagent Grade
Ethyl Ether, 98.0% min., ACS, EMD Millipore Available on GSA/VA Contract for Federal Government customers only.
Chemical Name or Material: Ethyl Ether Name Note: For ACS analysis CAS: 60-29-7 Purity Grade: ACS/NF/USP Physical Form: Liquid Assay Percent Range: 98.0%, min. Molecular Formula: (CH3CH2)2O Linear Formula: (CH3CH2)2O Formula Weight: 74.12 Synonym: Diethyl Ether
Ethyl Ether, Anhydrous, 99.8% min., DriSolv, EMD Millipore Available on GSA/VA Contract for Federal Government customers only.
Chemical Name or Material: Ethyl Ether, Anhydrous Name Note: DriSolv™ CAS: 60-29-7 Purity Grade: ACS/NF/USP Physical Form: Liquid Assay Percent Range: 99.8%, min. Molecular Formula: (CH3CH2)2O Linear Formula: (CH3CH2)2O Formula Weight: 74.12 Synonym: Diethyl Ether
  spinner