Fluoroboric Acid
Fluoroboric Acid, 50% (Laboratory), Fisher Chemical
Chemical Name or Material: Fluoroboric Acid Name Note: 50% CAS: 16872-11-0,7732-18-5, 10043-35-3 CAS: 7732-18-5 CAS: 10043-35-3 Purity Grade: Laboratory Grade Assay: 48 to 50% Assay Percent Range: 48 to 50% Molecular Formula: HBF4 Formula Weight: 87.81 MDL Number: MFCD00011345 Synonym: Hydrogen Tetrafluoroborate
  spinner