Fluosilicic Acid
Fluosilicic acid, 25 wt.% aqueous solution, ACROS Organics™ Available on GSA/VA Contract for Federal Government customers only.
Chemical Name or Material: Fluosilicic acid CAS: 16961-83-4 CAS: 7732-18-5 Assay: 19 to 20.6% (total fluorine) Molecular Formula: H2F6Si Linear Formula: H2SiF6 Formula Weight: 144.09 MDL Number: MFCD00036289 Synonym: Fluorosilicic acid
  spinner