Formamide
Formamide, 99+%, for spectroscopy, ACROS Organics™
Chemical Name or Material: Formamide CAS: 75-12-7 Purity Grade: For spectroscopy Assay: ≥99.0% Assay Percent Range: 99+% Molecular Formula: CH3NO Linear Formula: HCONH2 Formula Weight: 45.04 MDL Number: MFCD00007941
Formamide (Certified ACS), Fisher Chemical
Chemical Name or Material: Formamide CAS: 75-12-7 Purity Grade: Certified ACS Reagent Grade Assay: ≥99.5% Assay Percent Range: ≥99.5 % Molecular Formula: CH3NO Linear Formula: HCONH2 Formula Weight: 45.04 MDL Number: MFCD00007941 Synonym: Methanamide
Formamide, 99.5%, for analysis, ACROS Organics™ Available on GSA/VA Contract for Federal Government customers only.
Chemical Name or Material: Formamide CAS: 75-12-7 Purity Grade: For analysis Assay: ≥99.5% Assay Percent Range: 99.5% Molecular Formula: CH3NO Linear Formula: HCONH2 Formula Weight: 45.04 MDL Number: MFCD00007941
Formamide, 99%, for molecular biology, DNAse, RNAse and Protease free, ACROS Organics™ Available on GSA/VA Contract for Federal Government customers only.
Chemical Name or Material: Formamide Name Note: DNase, RNase and Protease Free CAS: 75-12-7 Purity Grade: For molecular biology Assay: ≥99% Assay Percent Range: 99% Molecular Formula: CH3NO Linear Formula: HCONH2 Formula Weight: 45.04 MDL Number: MFCD00007941
Formamide, 99.5+%, extra pure, ACROS Organics™
Chemical Name or Material: Formamide Name Note: 99.5+% CAS: 75-12-7 Purity Grade: Extra Pure Assay: ≥99.50% Assay Percent Range: 99.5+% Molecular Formula: CH3NO Linear Formula: HCONH2 Formula Weight: 45.04 MDL Number: MFCD00007941
Formamide, 99.6%, ACS reagent, ACROS Organics™ Available on GSA/VA Contract for Federal Government customers only.
Chemical Name or Material: Formamide CAS: 75-12-7 Purity Grade: ACS Reagent Grade Assay: ≥99.5% Assay Percent Range: 99.6% Molecular Formula: CH3NO Linear Formula: HCONH2 Formula Weight: 45.04 MDL Number: MFCD00007941
Formamide OR, J.T.Baker™
Chemical Name or Material: Formamide CAS: 75-12-7 Molecular Formula: HCONH2 Formula Weight: 45.04
Formamide (≥99.5%), Fisher BioReagents Available on GSA/VA Contract for Federal Government customers only.
Chemical Name or Material: Formamide CAS: 75-12-7 Purity Grade: Biotech Grade Physical Form: Liquid Assay: ≥99.5% Assay Percent Range: ≥99.5 % Molecular Formula: CH3NO Linear Formula: H2NCH:O Formula Weight: 45.04
Formamide (Molecular Biology), Fisher BioReagents Available on GSA/VA Contract for Federal Government customers only.
Chemical Name or Material: Formamide CAS: 75-12-7 Purity Grade: Molecular Biology Grade Physical Form: Liquid Assay: ≥99% Assay Percent Range: ≥99 % Molecular Formula: CH3NO Linear Formula: H2NCH:O Formula Weight: 45.04
Formamide, 99.5% min., ACS, EMD Millipore Available on GSA/VA Contract for Federal Government customers only.
Chemical Name or Material: Formamide Name Note: For ACS analysis CAS: 75-12-7 Purity Grade: ACS/EP/Reagent Grade Physical Form: Slightly Viscous Liquid Assay Percent Range: 99.5% min. Molecular Formula: CH3NO Linear Formula: HCONH2 Formula Weight: 45.4 Synonym: Formic acid amide
Formamide, Polarographic and Nonaqueous, Titration Grade, ACROS Organics™
Chemical Name or Material: Formamide CAS: 75-12-7 Purity Grade: Analytical Grade Molecular Formula: CH3NO Linear Formula: HCONH2 Formula Weight: 45.04 MDL Number: MFCD00007941
  spinner