Gallic Acid
Gallic acid monohydrate, ACROS Organics™
Chemical Name or Material: Gallic acid monohydrate Name Note: c.p. CAS: 5995-86-8 Assay: ≥99% (on dry substance) Molecular Formula: C7H6O5·H2O Linear Formula: (HO)3C6H2CO2H·H2O Formula Weight: 188.12 MDL Number: MFCD00149098 Synonym: 3, 4, 5-Trihydroxybenzoic acid monohydrate
Gallic Acid 98%, ACROS Organics™
Chemical Name or Material: Gallic acid Name Note: 98% CAS: 149-91-7 Assay: ≥97.5% Assay Percent Range: 98% Molecular Formula: C7H6O5 Formula Weight: 170.12 MDL Number: MFCD00002510 Synonym: 3, 4, 5-Trihydroxybenzoic acid
Gallic acid ethyleester, 99%, ACROS Organics™ Available on GSA/VA Contract for Federal Government customers only.
Chemical Name or Material: Gallic acid ethyleester CAS: 831-61-8 Assay: ≥98.5% Assay Percent Range: 99% Molecular Formula: C9H10O5 Formula Weight: 198.18 MDL Number: MFCD00016430
Gallic Acid Monohydrate (Powder/Certified) ACS, Fisher Chemical
Chemical Name or Material: Gallic Acid Monohydrate CAS: 5995-86-8 Purity Grade: Certified Grade Physical Form: Solid Molecular Formula: C6H2(OH)3COOH · H2O Linear Formula: C6H2(OH)3COOH·H2O Formula Weight: 188.14 MDL Number: MFCD00002510 Synonym: 3, 4, 5-Trihydroxybenzoic Acid
Gallic acid monohydrate, ACS reagent, ACROS Organics™
Chemical Name or Material: Gallic acid monohydrate CAS: 5995-86-8 Purity Grade: ACS Reagent Grade Molecular Formula: C7H6O5·H2O Formula Weight: 188.14 MDL Number: MFCD00149098 Synonym: 3, 4, 5-Trihydroxybenzoic acid monohydrate
Gallic Acid Solution, Saturated, Ricca Chemical
Chemical Name or Material: Gallic Acid Solution CAS: 5995-86-8 CAS: 7732-18-5 Physical Form: Liquid Assay Percent Range: Saturated
Gallic Acid, MP Biomedicals
Chemical Name or Material: Gallic Acid CAS: 149-91-7 Physical Form: Fine Powder Molecular Formula: C7H6O5 Formula Weight: 170.1 MDL Number: MFCD00002510 Synonym: 3, 4, 5-Trihydroxybenzoic acid
Gallic Acid, ACS, ≥98.0%; MP Biomedicals™
Chemical Name or Material: Gallic Acid CAS: 5995-86-8 Purity Grade: ACS Reagent Grade Assay Percent Range: ≥98% Molecular Formula: C7H6O5·H2O Formula Weight: 188.1 Synonym: 3, 4, 5-Trihydroxybenzoic acid
Gallic Acid Crystal, Macron Fine Chemicals™
Chemical Name or Material: Gallic Acid Name Note: Monohydrate CAS: 5995-86-8 Physical Form: Crystalline Molecular Formula: C6H2(OH)3COOH·H2O Formula Weight: 188.14
Gallic Acid Methyl Ester, MP Biomedicals™
Chemical Name or Material: Gallic Acid Methyl Ester CAS: 99-24-1 Molecular Formula: C8H8O5 Formula Weight: 184.1 Synonym: Methyl 3, 4, 5-trihydroxybenzoate
  spinner