GC MS and LC MS Standards
Bis(trimethylsilyl)trifluoroacetamide, BSTFA, EMD-Millipore Available on GSA/VA Contract for Federal Government customers only.
Chemical Name or Material: Bis(trimethylsilyl)trifluoroacetamide CAS: 25561-30-2 Molecular Formula: C8H18F3NOSi2 Formula Weight: 257.4 Synonym: 2,2,2-Trifluoro-N,O-bis(trimethylsilyl)acetamide
N-(Trimethylsilyl)imidazole, TMSI, EMD-Millipore
Chemical Name or Material: N-(Trimethylsilyl)imidazole CAS: 18156-74-6 Molecular Formula: C6H12N2Si Formula Weight: 140.25 Synonym: TSIM
n-Heptadecane, EMD-Millipore
Chemical Name or Material: n-Heptadecane CAS: 629-78-7 Assay Percent Range: ≥99.3% Molecular Formula: C17H36 Linear Formula: CH3(CH2)15CH3 Formula Weight: 240.47
1,1,1,3,3,3-Hexamethyldisilazane, HMDS, EMD-Millipore Available on GSA/VA Contract for Federal Government customers only.
Chemical Name or Material: 1,1,1,3,3,3-Hexamethyldisilazane CAS: 999-97-3 Molecular Formula: C6H19NSi2 Formula Weight: 161.39 Synonym: Bis(trimethylsilyl)amine, HMDS
n-Hexadecane, EMD-Millipore
Chemical Name or Material: n-Hexadecane CAS: 544-76-3 Assay Percent Range: ≥99.0% Molecular Formula: C16H34 Linear Formula: CH3(CH2)14CH3 Formula Weight: 226.44 Synonym: Cetane
n-Heptane, EMD-Millipore
Chemical Name or Material: n-Heptane CAS: 142-82-5 Assay Percent Range: ≥99.5% Molecular Formula: C7H16 Linear Formula: CH3(CH2)5CH3 Formula Weight: 100.2
n-Undecane, EMD-Millipore
Chemical Name or Material: n-Undecane CAS: 1120-21-4 Assay Percent Range: ≥99.5% Molecular Formula: C11H24 Linear Formula: CH3(CH2)9CH3 Formula Weight: 156.31
Methyl decanoate, EMD-Millipore
Chemical Name or Material: Methyl decanoate CAS: 110-42-9 Assay Percent Range: ≥99.5% Molecular Formula: C11H22O2 Formula Weight: 186.29 Synonym: Methyl caprate, Capric acid methyl ester, Decanoic acid methyl ester
n-Octadecane, EMD-Millipore
Chemical Name or Material: n-Octadecane CAS: 593-45-3 Assay Percent Range: ≥99.3% Molecular Formula: C18H38 Linear Formula: CH3(CH2)16CH3 Formula Weight: 254.49
D-Camphor, EMD-Millipore
Chemical Name or Material: D-Camphor CAS: 464-49-3 Molecular Formula: C10H16O Formula Weight: 152.23 Synonym: (+)-Camphor, (1R,4R)-Camphor, 1,7,7-Trimethylbicyclo[2.2.1]heptan-2-one, 2-Bornanone
Methyl laurate, EMD-Millipore
Chemical Name or Material: Methyl laurate CAS: 111-82-0 Assay Percent Range: ≥99.0% Molecular Formula: C13H26O2 Formula Weight: 214.34
n-Tridecane, EMD-Millipore
Chemical Name or Material: n-Tridecane CAS: 629-50-5 Assay Percent Range: ≥99.5% Molecular Formula: C13H28 Linear Formula: CH3(CH2)11CH3 Formula Weight: 184.36
m-Xylene, EMD-Millipore
Chemical Name or Material: m-Xylene CAS: 108-38-3 Assay Percent Range: ≥99.3% Molecular Formula: C8H10 Linear Formula: C6H4(CH3)2 Formula Weight: 106.16 Synonym: 1, 3-Dimethylbenzene
Methyl stearate, EMD-Millipore
Chemical Name or Material: Methyl stearate CAS: 112-61-8 Assay Percent Range: ≥99.0% Molecular Formula: C19H38O2 Formula Weight: 298.5 Synonym: Octadecanoic acid methyl ester
Dinonyl phthalate, EMD-Millipore
Chemical Name or Material: Dinonyl phthalate CAS: 84-76-4 Molecular Formula: C26H42O4 Formula Weight: 418.61 Synonym: DNP, Phthalic acid dinonyl ester
N,O-Bis(trimethylsilyl) acetamide, BSA, EMD-Millipore Available on GSA/VA Contract for Federal Government customers only.
Chemical Name or Material: N, O-Bis(trimethylsilyl) acetamide CAS: 10416-59-8 Molecular Formula: C8H21NOSi2 Linear Formula: CH3C[NSi(CH3)3]OSi(CH3)3 Formula Weight: 203.43 Synonym: BSA
n-Pentadecane, EEMD-Millipore
Chemical Name or Material: n-Pentadecane CAS: 629-62-9 Assay Percent Range: ≥99.5% Molecular Formula: C15H32 Linear Formula: CH3(CH2)13CH3 Formula Weight: 212.41
n-Dodecane, EMD-Millipore
Chemical Name or Material: n-Dodecane CAS: 112-40-3 Assay Percent Range: ≥99.0% Molecular Formula: C12H26 Linear Formula: CH3(CH2)10CH3 Formula Weight: 170.34
2,2,2-Trifluoro-N-methyl-N-(trimethylsilyl)acetamide, MSTFA, EMD-Millipore
Chemical Name or Material: 2, 2, 2-Trifluoro-N-methyl-N-(trimethylsilyl)acetamide CAS: 24589-78-4 Molecular Formula: C6H12F3NOSi Formula Weight: 199.25 Synonym: MSTFA
Ethyl methyl ketone, EMD-Millipore
Chemical Name or Material: Ethyl methyl ketone CAS: 78-93-3 Assay Percent Range: ≥99.5% Molecular Formula: C4H8O Linear Formula: C2H5COCH3 Formula Weight: 72.11 Synonym: 2-Butanone, Methyl ethyl ketone, MEK
n-Pentane, EMD-Millipore
Chemical Name or Material: n-Pentane CAS: 109-66-0 Assay Percent Range: ≥99.7% Molecular Formula: C5H12 Linear Formula: CH3(CH2)3CH3 Formula Weight: 72.15
o-Xylene, EMD-Millipore
Chemical Name or Material: o-Xylene CAS: 95-47-6 Assay Percent Range: ≥99.0% Molecular Formula: C8H10 Linear Formula: C6H4(CH3)2 Formula Weight: 106.17 Synonym: 1, 2-Dimethylbenzene
Activated Charcoal Sorbent, EMD-Millipore Available on GSA/VA Contract for Federal Government customers only.
Chemical Name or Material: Activated Charcoal Sorbent CAS: 7440-44-0 Molecular Formula: C Formula Weight: 12.01
Triton X-100, EMD-Millipore Available on GSA/VA Contract for Federal Government customers only.
Chemical Name or Material: Triton X-100 CAS: 9036-19-5 Molecular Formula: C14H21(C2H4O)nOH Linear Formula: C8H17C6H4(OCH2CH2)nOH Synonym: Octoxynol, (p-tert-Octylphenoxy)polyethoxyethanol, O-[4-(1,1,3,3-Tetramethylbutyl)phenoxy]polyethoxyethanol, Polyethyleneglycol-[4-(1,1,3,3-tetramethylbutyl)phenyl]-ether
Silicon oil 550, EMD-Millipore
Chemical Name or Material: Silicon oil 550
n-Decane, EMD-Millipore
Chemical Name or Material: n-Decane CAS: 124-18-5 Assay Percent Range: ≥99.5% Molecular Formula: C10H22 Linear Formula: CH3(CH2)8CH3 Formula Weight: 142.28
Dimethyl sulfoxide, EMD-Millipore
Chemical Name or Material: Dimethyl sulfoxide CAS: 67-68-5 Molecular Formula: C2H6OS Formula Weight: 78.1 Synonym: Methanesulfinylmethane, Methyl sulfoxide, Dimethyl(oxido)sulfur, DMSO
Ethylbenzene, EMD-Millipore
Chemical Name or Material: Ethylbenzene CAS: 100-41-4 Molecular Formula: C8H10 Formula Weight: 106.16
n-Tetradecane, EMD-Millipore
Chemical Name or Material: n-Tetradecane CAS: 629-59-4 Assay Percent Range: ≥99.0% Molecular Formula: C14H30 Formula Weight: 198.39
Cyclopentane, EMD-Millipore
Chemical Name or Material: Cyclopentane Assay Percent Range: ≥99.5% Molecular Formula: C5H10 Formula Weight: 70.13
  spinner