Gluconic Acid
Gluconic acid, sodium salt, 98%, ACROS Organics™ Available on GSA/VA Contract for Federal Government customers only.
Chemical Name or Material: Gluconic acid, sodium salt Assay: ≥97.5% Assay Percent Range: 98% Molecular Formula: C6H11NaO7 Linear Formula: HOCH2[CH(OH)]4CO2Na Formula Weight: 218.14 MDL Number: MFCD00064210 Synonym: Sodium gluconate
Gluconic Acid, 50 wt. % Solution in Water, ACROS Organics™ Available on GSA/VA Contract for Federal Government customers only.
Chemical Name or Material: Gluconic acid CAS: 526-95-4 CAS: 7732-18-5 Assay: 49 to 55% (Titrimetry other) Molecular Formula: C6H12O7 Formula Weight: 196.16 MDL Number: MFCD00004240
D-Gluconic acid, calcium salt, 99%, ACROS Organics™ Available on GSA/VA Contract for Federal Government customers only.
Chemical Name or Material: D-Gluconic acid, calcium salt Assay: 98.5 to 102.0% (as C12H22CaO14.H2O) (Titrimetry other) Assay Percent Range: 99% Molecular Formula: C12H22CaO14 Linear Formula: (HOCH2[CH(OH)]4CO2)2Ca Formula Weight: 430.38 MDL Number: MFCD00064209 Synonym: Calcium gluconate
D-Gluconic Acid Potassium Salt, ≥98%, MP Biomedicals™
Chemical Name or Material: D-Gluconic Acid Potassium Salt CAS: 299-27-4 Physical Form: Crystalline Powder Assay Percent Range: ≥98% Molecular Formula: C6H11O7K Formula Weight: 234.3 MDL Number: MFCD00064211 Synonym: Gluconsan
Gluconic Acid, 50% Soln, MP Biomedicals
Chemical Name or Material: Gluconic Acid CAS: 526-95-4 Formula Weight: 196.16
D-Gluconic acid, zinc salt hydrate, 97+%, ACROS Organics™
Chemical Name or Material: D-Gluconic acid, zinc salt hydrate Assay: ≥97.0% (on dry substance) Assay Percent Range: 97+% Molecular Formula: C12H22O14Zn·xH2O Linear Formula: [CH2(OH)(CH(OH))4COO]2Zn·xH2O MDL Number: MFCD04037230 Synonym: Zinc gluconate hydrate
  spinner