Glutamic Acid -Glu and Derivatives
L(+)-Glutamic acid hydrochloride, 99%, ACROS Organics™
Chemical Name or Material: L(+)-Glutamic acid hydrochloride Name Note: 99% CAS: 138-15-8 Assay: 98.5 to 101.5% Assay Percent Range: 99% Molecular Formula: C5H9NO4·HCl Linear Formula: HO2CCH2CH2CH(NH2)CO2H·HCl Formula Weight: 183.59 MDL Number: MFCD00012619
L(+)-Glutamic acid, 99%, ACROS Organics™
Chemical Name or Material: L(+)-Glutamic acid Assay: 98.5 to 100.5% (on dried substance) Assay Percent Range: 99% Molecular Formula: C5H9NO4 Linear Formula: HO2CCH2CH2CH(NH2)CO2H Formula Weight: 147.13
L-(+)-Glutamic Acid (Powder/Certified ACS), Fisher Chemical Available on GSA/VA Contract for Federal Government customers only.
Chemical Name or Material: L-(+)-Glutamic Acid CAS: 56-86-0 Purity Grade: Certified Grade Physical Form: Solid Assay: ≥99% Assay Percent Range: ≥99 % Molecular Formula: C5H9NO4 Linear Formula: HOOCCHNH2CH2CH2COOH Formula Weight: 147.13 MDL Number: MFCD00002634 Synonym: Glutaminol
L-Glutamic acid, 99+%
Chemical Name or Material: L-Glutamic Acid CAS: 56-86-0 Molecular Formula: C5H9NO4 Formula Weight: 147.13 MDL Number: MFCD00002634 Beilstein: 1723801 Merck Index: 14,4469
N-Acetylglycyl-D-glutamic acid, Tocris
Chemical Name or Material: N-Acetylglycyl-D-glutamic acid Assay Percent Range: >98% Molecular Formula: C9H14N2O6 Formula Weight: 246.22
(±)-threo-3-Methylglutamic acid, Tocris
Chemical Name or Material: (±)-threo-3-Methylglutamic acid Assay Percent Range: >98% Molecular Formula: C6H11NO4 Formula Weight: 161.16
SYM 2081, Tocris
Chemical Name or Material: SYM 2081 Assay Percent Range: >97% Molecular Formula: C6H11NO4 Formula Weight: 161.16 Synonym: (2S, 4R)-4-Methylglutamic acid
L-Glutamic acid, Tocris
Chemical Name or Material: L-Glutamic acid CAS: 56-86-0 Assay Percent Range: >98% Molecular Formula: C5H9NO4 Formula Weight: 147.13 Synonym: (S)-1-Aminopropane-1, 3-dicarboxylic acid
(2R,3S)-Chlorpheg, Tocris
Chemical Name or Material: (2R, 3S)-Chlorpheg CAS: 140924-23-8 Assay Percent Range: >98% Molecular Formula: C11H12ClNO4 Formula Weight: 257.67 Synonym: (2R, 3S)-β-p-Chlorophenylglutamic acid
D-Glutamic acid, Tocris
Chemical Name or Material: D-Glutamic acid CAS: 6893-26-1 Assay Percent Range: >98% Molecular Formula: C5H9NO4 Formula Weight: 147.13 Synonym: (R)-1-Aminopropane-1, 3-dicarboxylic acid
ZJ 43, Tocris
Chemical Name or Material: ZJ 43 CAS: 723331-20-2 Assay Percent Range: >98% Molecular Formula: C12H20N2O7 Formula Weight: 304.3 Synonym: N-[[[(1S)-1-Carboxy-3-methylbutyl]amino]carbonyl]-L-glutamic acid
EGLU, Tocris
Chemical Name or Material: EGLU Assay Percent Range: >95% Molecular Formula: C7H13NO4 Formula Weight: 175.18 Synonym: (2S)-α-Ethylglutamic acid
Methotrexate, Tocris
Chemical Name or Material: Methotrexate CAS: 59-05-2 Assay Percent Range: >99% Molecular Formula: C20H22N8O5 Formula Weight: 454.44 Synonym: N-[4-[[(2, 4-Diamino-6-pteridinyl)methyl]methylamino]benzoyl]-L-glutamic acid
Spaglumic acid, Tocris
Chemical Name or Material: Spaglumic acid CAS: 3106-85-2 Assay Percent Range: >96% Molecular Formula: C11H16N2O8 Formula Weight: 304.26 Synonym: N-Acetyl-L-aspartyl-L-glutamic acid
N-Acetyl-ASP-GLU, MP Biomedicals™
Chemical Name or Material: N-Acetyl-ASP-GLU CAS: 3106-85-2 Physical Form: Powder Molecular Formula: C11H16N2O8 Formula Weight: 304.3 MDL Number: MFCD00076652 Synonym: N-Ac-D-E
L-Glutamic Acid, 99+%, Monosodium Salt, MP Biomedicals
Chemical Name or Material: L-Glutamic Acid, 99+% Name Note: Monosodium Salt CAS: 142-47-2 Purity Grade: Cell Culture Reagent Grade Molecular Formula: C5H8NO4Na Formula Weight: 169.1 Synonym: Monosodium glutamate
L-Glutamic Acid, ≥99%, MP Biomedicals™
Chemical Name or Material: L-Glutamic Acid CAS: 56-86-0 Physical Form: Powder Assay Percent Range: ≥99% Molecular Formula: C5H9NO4 Formula Weight: 147.1 MDL Number: MFCD00002634 Synonym: (S)-(+)-glutamic acid
L-Glutamic Acid, free acid, ≥99%, MP Biomedicals™
Chemical Name or Material: L-Glutamic acid Monosodium Salt CAS: 142-47-2 Physical Form: Powder Assay Percent Range: ≥99% Molecular Formula: C5H8NO4Na · H2O Formula Weight: 187.13 MDL Number: MFCD00013074 Synonym: MSG
L-Glutamic Acid (White Crystals or Crystalline Powder), Fisher BioReagents
Chemical Name or Material: L-Glutamic Acid CAS: 56-86-0 Physical Form: Solid Assay: 99.0 to 100.5% Assay Percent Range: 99.0 to 100.5 % Molecular Formula: C5H9NO4 Formula Weight: 147.13
L-Glutamic Acid, Sodium Salt OR, Macron Fine Chemicals™
Chemical Name or Material: L-Glutamic Acid, Sodium Salt Name Note: Monohydrate CAS: 6106-04-3 Molecular Formula: HOCOCH2CH2CH(NH2)COONa·H2O Formula Weight: 187.13
  spinner