Gold -Au
Gold foil, >99.9975% (metals basis), Alpha Aesar, Premion™
Chemical Name or Material: Gold CAS: 7440-57-5 Physical Form: Foil, 0.1mm (0.004 in.) thick Assay Percent Range: >99.9975% (Metals basis) MDL Number: MFCD00003436
Gold Wire, 0.5mm (0.02 in.) dia., 99.95% (Metals basis), Alfa Aesar™
Chemical Name or Material: Gold CAS: 7440-57-5 Physical Form: Wire, 0.5mm (0.02 in.) dia. Assay Percent Range: 99.95% (Metals basis) MDL Number: MFCD00003436
Gold Gauze, 52 mesh woven from 0.102mm (0.004 in.) dia. Wire, 99.99% (Metals basis), Alfa Aesar™
Chemical Name or Material: Gold CAS: 7440-57-5 Physical Form: Gauze Assay Percent Range: 99.99% (Metals basis) MDL Number: MFCD00003436
Gold Rod, 3mm (0.1 in.) dia., Premion™, 99.9985% (Metals basis), Alfa Aesar™
Chemical Name or Material: Gold CAS: 7440-57-5 Physical Form: Rod, 3mm (0.1 in.) dia. Assay Percent Range: 99.9985% (Metals basis) MDL Number: MFCD00003436
Gold Powder, spherical, APS 1.5-3.0 micron, 99.96+% (Metal Basis), Alfa Aesar™
Chemical Name or Material: Gold CAS: 7440-57-5 Physical Form: Spherical Powder Assay Percent Range: 99.96+%, (metal basis) MDL Number: MFCD00003436
Gold, 99.99%, (trace metal basis), shot, ACROS Organics™ Available on GSA/VA Contract for Federal Government customers only.
Chemical Name or Material: Gold CAS: 7440-57-5 Purity Grade: For trace metal basis analysis Assay Percent Range: 99.99% Molecular Formula: Au Linear Formula: Au Formula Weight: 196.97
Gold foil, 99.95% (metal basis), Alfa Aesar™
Chemical Name or Material: Gold CAS: 7440-57-5 Physical Form: Wire, 0.5mm (0.02 in.) dia. Assay Percent Range: 99.999% (Metals basis) MDL Number: MFCD00003436
Gold wire 0.5mm dia, 99.999% (metal basis), Premion™, Alfa Aesar™
Chemical Name or Material: Gold CAS: 7440-57-5 MDL Number: MFCD00003436
Gold Splatters, 99.9% (Metals basis), Alfa Aesar™
Chemical Name or Material: Gold CAS: 7440-57-5 Physical Form: Splatters Assay Percent Range: 99.9% (Metals basis) MDL Number: MFCD00003436
Gold Wire, 1.0mm (0.04 in.) dia., Premion™, 99.999% (Metals basis), Alfa Aesar™
Chemical Name or Material: Gold CAS: 7440-57-5 Physical Form: Wire, 1.0mm (0.04 in.) dia. Assay Percent Range: 99.999% (Metals basis) MDL Number: MFCD00003436
Gold slug, Premion™, 99.999% (metals basis), Alfa Aesar™
Chemical Name or Material: Gold CAS: 7440-57-5 Physical Form: Slug, dia. X L: 6.35 X 6.35mm (0.25 X 0.25 in.) Assay Percent Range: 99.999% (Metals basis) MDL Number: MFCD00003436
Gold Wire, 1.0mm (0.04 in.) dia., 99.9% (Metals basis), Alfa Aesar™
Chemical Name or Material: Gold CAS: 7440-57-5 Physical Form: Wire, 1.0mm (0.04 in.) dia. Assay Percent Range: 99.9% (Metals basis) MDL Number: MFCD00003436
Gold Shots, semi-spherical, 6.35mm (0.25in.) and down, Premion™, 99.999% (Metals basis), Alfa Aesar™
Chemical Name or Material: Gold CAS: 7440-57-5 Physical Form: Shots, Semi-Spherical, 6.35mm (0.25 in.) and down Assay Percent Range: 99.999% (Metals basis) MDL Number: MFCD00003436
Gold Wire, 0.25mm (0.01 in.) dia., Premion™, 99.999% (Metals basis), Alfa Aesar™
Chemical Name or Material: Gold CAS: 7440-57-5 Physical Form: Wire, 0.25mm (0.01 in.) dia. Assay Percent Range: 99.999% (Metals basis) MDL Number: MFCD00003436
Gold powder, 99.>96%% (metals basis), Alfa Aesar™
Chemical Name or Material: Gold CAS: 7440-57-5 Physical Form: Spherical Powder Assay Percent Range: >99.96% (Metals basis) MDL Number: MFCD00003436
Gold Foil, 0.127mm (0.005 in.) thick, Premion™, 99.99% (Metals basis), Alfa Aesar™
Chemical Name or Material: Gold CAS: 7440-57-5 Physical Form: Foil, 0.127mm (0.005 in.) thick Assay Percent Range: 99.99% (Metals basis) MDL Number: MFCD00003436
Gold Wire, 0.25mm (0.01 in.) dia., 99.95% (Metals basis), Alfa Aesar™
Chemical Name or Material: Gold CAS: 7440-57-5 Physical Form: Wire, 0.25mm (0.01 in.) dia. Assay Percent Range: 99.95% (Metals basis) MDL Number: MFCD00003436
Gold Shots, Semi-spherical, 6.35mm (0.25 in.) & down, 99.95% (Metals basis), Alfa Aesar™
Chemical Name or Material: Gold CAS: 7440-57-5 Physical Form: Shots, Semi-Spherical, 6.35mm (0.25 in.) and down Assay Percent Range: 99.95% (Metals basis) MDL Number: MFCD00003436
Gold Wire, 0.05mm (0.002 in.) dia., Premion™, 99.995% (Metals basis), Alfa Aesar™
Chemical Name or Material: Gold CAS: 7440-57-5 Physical Form: Wire, 0.05mm (0.002 in.) dia. Assay Percent Range: 99.995% (Metals basis) MDL Number: MFCD00003436
Gold Slug, Dia. X L: 3.175 x 6.35mm (0.125 x 0.25 in.), Premion™, 99.99% (Metals basis), Alfa Aesar™
Chemical Name or Material: Gold CAS: 7440-57-5 Physical Form: Slug, dia. X L: 3.175 X 6.35mm (0.125 X 0.25 in.) Assay Percent Range: 99.99% (Metals basis) MDL Number: MFCD00003436
Gold Gauze, 100 mesh woven from 0.064mm (0.0025 in.) dia. Wire, 99.99% (Metals basis), Alfa Aesar™
Chemical Name or Material: Gold CAS: 7440-57-5 Physical Form: Gauze Assay Percent Range: 99.99% (Metals basis) MDL Number: MFCD00003436
Gold Wire, 0.025mm (0.001 in.) dia., 99.95% (Metals basis), Alfa Aesar™
Chemical Name or Material: Gold CAS: 7440-57-5 Physical Form: Wire, 0.025mm (0.001 in.) dia. Assay Percent Range: 99.95% (Metals basis) MDL Number: MFCD00003436
Gold Foil, 0.025mm (0.001 in.) thick, Premion™, 99.985% (Metals basis), Alfa Aesar™
Chemical Name or Material: Gold CAS: 7440-57-5 Physical Form: Foil, 0.025mm (0.001 in.) thick Assay Percent Range: 99.985% (Metals basis) MDL Number: MFCD00003436
Gold Wire, 0.1mm (0.004 in.) dia., 99.95% (Metals basis), Alfa Aesar™
Chemical Name or Material: Gold CAS: 7440-57-5 Physical Form: Wire, 0.1mm (0.004 in.) dia. Assay Percent Range: 99.95% (Metals basis) MDL Number: MFCD00003436
Gold Wire, 0.2mm (0.008 in.) dia., 99.9% (Metals basis), Alfa Aesar™
Chemical Name or Material: Gold CAS: 7440-57-5 Physical Form: Wire, 0.2mm (0.008 in.) dia. Assay Percent Range: 99.9% (Metals basis) MDL Number: MFCD00003436
Gold Powder, spherical, APS 0.8-1.5 micron, 99.96+% (Metal Basis), Alfa Aesar™
Chemical Name or Material: Gold CAS: 7440-57-5 Physical Form: Powder, Spherical, APS 1.5 To 3.0μ M Assay Percent Range: 99.96+%, (metal basis) MDL Number: MFCD00003436
Gold wire, 99.9985% (metals basis), Alfa Aesar™, Premion™
Chemical Name or Material: Gold CAS: 7440-57-5 Physical Form: Wire, 0.5mm (0.02 in.) dia. Assay Percent Range: 99.9985% (Metals basis) MDL Number: MFCD00003436
Gold wire, 0.25mm (0.01 in.) dia., 99.9% (metals basis)
Chemical Name or Material: Gold wire CAS: 7440-57-5 Name Note: 0.25mm (0.01 in.) dia. MDL Number: MFCD00003436
Gold Gauze, 82 mesh woven from 0.06mm (0.0025 in) dia wire, 99.9% (Metals basis), Alfa Aesar™
Chemical Name or Material: Gold CAS: 7440-57-5 Physical Form: Gauze Assay Percent Range: 99.9% (Metals basis) MDL Number: MFCD00003436
Gold Foil, 0.05mm (0.002 in.) thick, 99.95% (Metals basis), Alfa Aesar™
Chemical Name or Material: Gold CAS: 7440-57-5 Physical Form: Foil, 0.05mm (0.002 in.) thick Assay Percent Range: 99.95% (Metals basis) MDL Number: MFCD00003436
  spinner