Hexane -n-
n-Hexane, 99+%, for analysis, ACROS Organics™ Available on GSA/VA Contract for Federal Government customers only.
Chemical Name or Material: n-Hexane CAS: 110-54-3 CAS Min %: 99.0 CAS Max %: 100.0 Purity Grade: For analysis Assay: ≥99.0% (GC) Assay Percent Range: 99+% Molecular Formula: C6H14 Linear Formula: CH3(CH2)4CH3 Formula Weight: 86.18 MDL Number: MFCD00009520 Synonym: Hex
n-Hexane, 96+%, Extra Dry, AcroSeal™, ACROS Organics™
Chemical Name or Material: n-Hexane CAS: 110-54-3 CAS Min %: 96.0 CAS Max %: 100.0 Purity Grade: Extra Dry Assay: ≥96.0% (GC) Assay Percent Range: 96+% Molecular Formula: C6H14 Linear Formula: CH3(CH2)4CH3 Formula Weight: 86.18 MDL Number: MFCD00009520 Synonym: Hex
n-Hexane, for analysis, contains appr. 95% n-Hexane, ACROS Organics™
Chemical Name or Material: n-Hexane CAS: 110-54-3 Purity Grade: For analysis Assay: ≥95.0% Molecular Formula: C6H14 Linear Formula: CH3(CH2)4CH3 Formula Weight: 86.18 MDL Number: MFCD00009520 Synonym: Hex
n-Hexane, Anhydrous, Alfa Aesar
Chemical Name or Material: n-Hexane CAS: 110-54-3 Physical Form: Liquid Molecular Formula: C6H14 Formula Weight: 86.18 MDL Number: MFCD00009520
n-Hexane, 97%, Extra Dry over Molecular Sieve, AcroSeal™, ACROS Organics™ Available on GSA/VA Contract for Federal Government customers only.
Chemical Name or Material: n-Hexane Name Note: Over Molecular Sieves CAS: 110-54-3 CAS Min %: 96.0 CAS Max %: 100.0 Purity Grade: Extra Dry over Molecular Sieve Assay: ≥96.0% (GC) Assay Percent Range: 97% Molecular Formula: C6H14 Linear Formula: CH3(CH2)4CH3 Formula Weight: 86.18 MDL Number: MFCD00009520 Synonym: Hex
n-Hexane, 95+%, extra pure, ACROS Organics™ Available on GSA/VA Contract for Federal Government customers only.
Chemical Name or Material: n-Hexane Name Note: 95% CAS: 110-54-3 CAS Min %: 95.0 CAS Max %: 100.0 Purity Grade: Extra Pure Assay: ≥95.0% Assay Percent Range: 95+% Molecular Formula: C6H14 Linear Formula: CH3(CH2)4CH3 Formula Weight: 86.18 MDL Number: MFCD00009520 Synonym: Hex
Hexanes, mixed isomers, (60+% n-hexane)
Chemical Name or Material: Hexanes, mixed isomers CAS: 92112-69-1 Name Note: Mixed isomers Molecular Formula: C6H14 Formula Weight: 86.18 MDL Number: MFCD02179311 UN Number: UN1208
Hexanes (95% n-hexane), ULTRA RESI-ANALYZED, J.T.Baker™
Chemical Name or Material: Hexanes (95% n-hexane) CAS: 110-54-3 Purity Grade: 99.5 % min. Assay Percent Range: 95% n-hexane Molecular Formula: CH3(CH2)4CH3 Formula Weight: 86.18g/mol
n-Hexane, For ACS analysis, MP Biomedicals™
Chemical Name or Material: n-Hexane Name Note: For ACS analysis CAS: 110-54-3 Physical Form: Liquid Assay Percent Range: 99.9% Molecular Formula: C6H14 Formula Weight: 86.18 Synonym: Esani
n-Hexane, 99% , 99.0% min., EMD Millipore Available on GSA/VA Contract for Federal Government customers only.
Chemical Name or Material: n-Hexane CAS: 110-54-3 Physical Form: Liquid Assay Percent Range: 99.0%, min. Molecular Formula: CH3(CH2)4CH3 Linear Formula: CH3(CH2)4CH3 Formula Weight: 86.18
n-Hexane, >95% min., Ultra Scientific
Chemical Name or Material: n-Hexane CAS: 000110-54-3 CAS Max %: 100.0 Assay Percent Range: >95% min.
  spinner