Hydrobromic Acid
Hydrobromic Acid (OPTIMA™), Fisher Chemical Available on GSA/VA Contract for Federal Government customers only.
Chemical Name or Material: Hydrobromic Acid Name Note: 44–49% CAS: 10035-10-6 (Hydrogen Bromide) CAS: 7732-18-5 (Water) Purity Grade: OPTIMA™ Molecular Formula: HBr Formula Weight: 80.91 MDL Number: MFCD00011323 Synonym: Hydrogen Bromide
Hydrobromic Acid, ACS, 47.0-49%, Alfa Aesar
Chemical Name or Material: Hydrobromic acid CAS: 10035-10-6 CAS: 7732-18-5 Purity Grade: ACS Grade Physical Form: Liquid Assay Percent Range: 47.0-49.0% Molecular Formula: HBr Formula Weight: 80.92 MDL Number: MFCD00011323 Synonym: Hydrogen bromide
Hydrobromic acid, for analysis, ca. 48% solution in water, ACROS Organics™
Chemical Name or Material: Hydrobromic acid CAS: 10035-10-6 CAS: 7732-18-5 Purity Grade: For analysis Assay: 47 to 49% (Argentometry) Molecular Formula: HBr Linear Formula: HBr Formula Weight: 80.9
Hydrobromic Acid, ca 48 wt. % Solution in Water (Tech.), ACROS Organics™ Available on GSA/VA Contract for Federal Government customers only.
Chemical Name or Material: Hydrobromic acid Name Note: tech. CAS: 10035-10-6 CAS: 7732-18-5 Purity Grade: Pure Assay: ≥47% Molecular Formula: HBr Linear Formula: HBr Formula Weight: 80.9 MDL Number: MFCD00011323
Hydrobromic Acid, Reagent ACS, ACROS Organics™
Chemical Name or Material: Hydrobromic acid CAS: 10035-10-6 CAS: 7732-18-5 Purity Grade: ACS Reagent Grade Assay: 47.0 to 49.0% Molecular Formula: HBr Linear Formula: HBr Formula Weight: 80.9 MDL Number: MFCD00011323
Hydrobromic Acid, 47-49% (Certified ACS), Fisher Chemical Available on GSA/VA Contract for Federal Government customers only.
Chemical Name or Material: Hydrobromic Acid, 47-49% CAS: 10035-10-6 CAS: 7732-18-5 Purity Grade: Certified ACS Grade Assay: 47.0 to 49.0% Assay Percent Range: 47.0 to 49.0 % Molecular Formula: HBr Formula Weight: 80.91 MDL Number: MFCD00011323
Hydrobromic acid, Alfa Aesar™
Chemical Name or Material: Hydrobromic acid CAS: 10035-10-6 Physical Form: Liquid Molecular Formula: HBr Formula Weight: 80.92 MDL Number: MFCD00011323 Synonym: Hydrogen bromide
Hydrobromic acid, 99.9999% (metals basis), 48% w/w aq. soln., Alfa Aesar™
Chemical Name or Material: Hydrobromic acid CAS: 10035-10-6 CAS: 7732-18-5 Physical Form: Liquid Assay Percent Range: 99.9999% (metals basis) Molecular Formula: HBr Formula Weight: 80.92 MDL Number: MFCD00011323 Synonym: Hydrogen bromide
  spinner