Hydrochloric Acid
Hydrochloric Acid, Certified ACS Plus, Fisher Chemical Available on GSA/VA Contract for Federal Government customers only.
Assay: 36.5 to 38.0% (w/w) Molecular Formula: HCl · H2O
Hydrogen chloride, 5 to 6N solution in 2-propanol, AcroSeal™, ACROS Organics™ Available on GSA/VA Contract for Federal Government customers only.
Chemical Name or Material: Hydrogen chloride Name Note: 5 to 6N solution in 2-propanol CAS: 7647-01-0 CAS: 67-63-0 Molecular Formula: ClH Linear Formula: HCl Formula Weight: 36.45 MDL Number: MFCD00011324
Hydrochloric acid, ACS reagent, ca. 37% solution in water, ACROS Organics™ Available on GSA/VA Contract for Federal Government customers only.
Chemical Name or Material: Hydrochloric acid Name Note: ca. 37% Solution in Water CAS: 7647-01-0 CAS: 7732-18-5 Purity Grade: ACS Reagent Grade Assay: 36.5 to 38.0% Molecular Formula: ClH Linear Formula: HCl Formula Weight: 36.45 MDL Number: MFCD00011324 Synonym: Muriatic acid
Hydrogen chloride, 3M solution in CPME, AcroSeal™, ACROS Organics™
Chemical Name or Material: Hydrogen chloride CAS: 7647-01-0 CAS: 5614-37-9 Molecular Formula: ClH Linear Formula: HCl Formula Weight: 36.45
Hydrochloric Acid Solution, 0.01N (N/100) (Certified), Fisher Chemical
Chemical Name or Material: Hydrochloric Acid Solution, 0.01N (N/100) CAS: 7647-01-0 CAS: 7732-18-5 Purity Grade: Certified Grade Molecular Formula: HCl MDL Number: MFCD00011324
Hydrochloric acid, for analysis, 25% solution in water, ACROS Organics™ Available on GSA/VA Contract for Federal Government customers only.
Chemical Name or Material: Hydrochloric acid CAS: 7647-01-0 CAS: 7732-18-5 Purity Grade: For analysis Assay: ≥25.0% Molecular Formula: ClH Linear Formula: HCl Formula Weight: 36.45 MDL Number: MFCD00011324 Synonym: Muriatic acid, 25%
Hydrochloric Acid Solution, 0.5N (N/2) (Certified), Fisher Chemical Available on GSA/VA Contract for Federal Government customers only.
Chemical Name or Material: Hydrochloric Acid Solution, 0.5N (N/2) CAS: 7647-01-0 CAS: 7732-18-5 Purity Grade: Certified Grade Molecular Formula: HCl MDL Number: MFCD00011324 Synonym: Muriatic Acid
Hydrogen Chloride , 0.5M solution in methanol, ACROS Organics™ Available on GSA/VA Contract for Federal Government customers only.
Chemical Name or Material: Hydrogen chloride Name Note: 0.5M solution in methanol CAS: 7647-01-0 CAS: 67-56-1 Molecular Formula: ClH Linear Formula: HCl Formula Weight: 36.46 MDL Number: MFCD00011324
Hydrogen chloride, pure, 1N solution in diethyl ether, AcroSeal™, ACROS Organics™
Chemical Name or Material: Hydrogen chloride Name Note: 1N Solution in Diethyl Ether CAS: 7647-01-0 CAS: 60-29-7 Purity Grade: Pure Molecular Formula: ClH Linear Formula: HCl Formula Weight: 36.45 MDL Number: MFCD00011324
Hydrochloric acid, 10% v/v aq. soln., Alfa Aesar™
Chemical Name or Material: Hydrochloric acid CAS: 7647-01-0 CAS: 7732-18-5 Physical Form: Liquid Molecular Formula: HCl Formula Weight: 36.46 MDL Number: MFCD00011324
Hydrochloric Acid, 25% v/v (1+3), Fisher Chemical
Chemical Name or Material: Hydrochloric Acid, 25% (v/v) (1+3) CAS: 7732-18-5 CAS: 7647-01-0 Molecular Formula: HCl·H2O MDL Number: MFCD00011324
Hydrochloric acid, 99.999% (metals basis), 36.5% min., Alfa Aesar™
Chemical Name or Material: Hydrochloric acid CAS: 7647-01-0 Physical Form: Liquid Assay Percent Range: 99.999% (metals basis) Molecular Formula: HCl Formula Weight: 36.46 MDL Number: MFCD00011324
Hydrochloric acid, 0.1 N standard solution, ACROS Organics™ Available on GSA/VA Contract for Federal Government customers only.
Chemical Name or Material: Hydrochloric acid Name Note: 0.1 N Standard Solution CAS: 7647-01-0 CAS: 7732-18-5 Molecular Formula: ClH Linear Formula: HCl Formula Weight: 36.45 MDL Number: MFCD00011324
Hydrochloric Acid Solution, 6N (Certified), Fisher Chemical Available on GSA/VA Contract for Federal Government customers only.
Chemical Name or Material: Hydrochloric Acid Solution, 6N CAS: 7732-18-5 CAS: 7647-01-0 Purity Grade: Certified Grade Molecular Formula: HCl·H2O MDL Number: MFCD00011324 Synonym: Muriatic Acid
Hydrochloric acid, 4N solution in 1,4-Dioxane/water, ACROS Organics™ Available on GSA/VA Contract for Federal Government customers only.
Chemical Name or Material: Hydrochloric acid Name Note: 4N Solution in 1, 4-dioxane/water CAS: 7647-01-0 CAS: 7732-18-5 CAS: 123-91-1 Molecular Formula: ClH Linear Formula: HCl Formula Weight: 36.45 MDL Number: MFCD00011324
Hydrochloric acid, 0.5N Standardized Solution, Alfa Aesar™
Chemical Name or Material: Hydrochloric acid CAS: 7647-01-0 CAS: 7732-18-5 Physical Form: Liquid Molecular Formula: HCl Formula Weight: 36.46 MDL Number: MFCD00011324
Hydrochloric Acid Solutions, Normal Products for Science Education
Chemical Name or Material: Hydrochloric Acid Solutions CAS: 7647-01-0 CAS: 7732-18-5 Physical Form: Liquid Assay Percent Range: 3.65% v/v Linear Formula: HCl Formula Weight: 36.46 Synonym: Hydrogen Chloride Solution
Hydrochloric Acid (1+3), Fisher Chemical
Chemical Name or Material: Hydrochloric Acid (1+3) CAS: 7732-18-5 CAS Min %: 76.0 CAS Max %: 79.0 CAS: 7647-01-0 CAS Min %: 22.32 CAS Max %: 23.24
Hydrochloric Acid (Technical), Fisher Chemical Available on GSA/VA Contract for Federal Government customers only.
Chemical Name or Material: Hydrochloric Acid CAS: 7647-01-0, 7732-18-5 CAS: 7732-18-5 Purity Grade: Technical Grade Molecular Formula: HCl · H2O Formula Weight: 36.46 MDL Number: MFCD00011324 Synonym: Muriatic Acid
Hydrochloric acid, pure, fuming, 37% solution in water, ACROS Organics™ Available on GSA/VA Contract for Federal Government customers only.
Chemical Name or Material: Hydrochloric acid CAS: 7647-01-0 CAS: 7732-18-5 Purity Grade: Pure Assay: 35.0 to 39.0% Molecular Formula: ClH Linear Formula: HCl Formula Weight: 36.45 MDL Number: MFCD00011324 Synonym: Muriatic acid
Hydrochloric acid, pure, 25% solution in water, ACROS Organics™
Chemical Name or Material: Hydrochloric acid CAS: 7647-01-0 CAS: 7732-18-5 Purity Grade: Pure Assay: ≥25.0% Molecular Formula: ClH Linear Formula: HCl Formula Weight: 36.45 MDL Number: MFCD00011324 Synonym: Muriatic acid, 25%
Hydrochloric acid, Environmental Grade, 34-37.5%, Alfa Aesar™
Chemical Name or Material: Hydrochloric acid Name Note: Single sub-boiling quartz distillation CAS: 7647-01-0 Purity Grade: Environmental Grade Physical Form: Liquid Assay Percent Range: 34-37.5% Molecular Formula: HCl Formula Weight: 36.46 MDL Number: MFCD00011324
Hydrochloric Acid Solution (0.2N (N/5)/Certified), Fisher Chemical
Chemical Name or Material: Hydrochloric Acid Solution, 0.2N (N/5) CAS: 7647-01-0 CAS: 7732-18-5 Purity Grade: Certified Grade Molecular Formula: HCl MDL Number: MFCD00011324
Hydrochloric Acid Solution, 0.1N (N/10) (Certified), Fisher Chemical Available on GSA/VA Contract for Federal Government customers only.
Chemical Name or Material: Hydrochloric Acid Solution, 0.1N (N/10) CAS: 7647-01-0 CAS: 7732-18-5 Purity Grade: Certified Grade Molecular Formula: HCl MDL Number: MFCD00011324
Hydrochloric Acid, ACS, 12M Products for Science Education
Chemical Name or Material: Hydrochloric Acid CAS: 7647-01-0 CAS: 7732-18-5 Purity Grade: ACS Grade Physical Form: Liquid Assay Percent Range: 36.5% v/v Linear Formula: HCl Formula Weight: 36.46 Synonym: Hydrogen Chloride
Hydrochloric Acid Solutions (Molar) Products for Science Education
Chemical Name or Material: Hydrochloric Acid Solution CAS: 7647-01-0 CAS: 7732-18-5 Physical Form: Liquid Synonym: Hydrogen Chloride Solution
hydrogen chloride, 1M solution in acetic acid, AcroSeal™, ACROS Organics™ Available on GSA/VA Contract for Federal Government customers only.
Chemical Name or Material: Hydrogen chloride Name Note: 1M solution in acetic acid CAS: 7647-01-0 CAS: 64-19-7 Molecular Formula: ClH Linear Formula: HCl Formula Weight: 36.45 MDL Number: MFCD00011324
Hydrochloric acid, 37%, extra pure
Chemical Name or Material: Hydrochloric Acid CAS: 7647-01-0, 7732-18-5 CAS: 7732-18-5 Purity Grade: Extra Pure Molecular Formula: ClH Formula Weight: 36.45 Synonym: Muriatic Acid
Hydrochloric Acid Solution, 1N (Certified), Fisher Chemical Available on GSA/VA Contract for Federal Government customers only.
Chemical Name or Material: Hydrochloric Acid Solution, 1N CAS: 7647-01-0 CAS: 7732-18-5 Purity Grade: Certified Grade Molecular Formula: HCl MDL Number: MFCD00011324 Synonym: Muriatic Acid
Hydrochloric acid, for analysis, fuming, 37% solution in water, ACROS Organics™ Available on GSA/VA Contract for Federal Government customers only.
Chemical Name or Material: Hydrochloric acid CAS: 7647-01-0 CAS: 7732-18-5 Purity Grade: For analysis Assay: 35.0 to 39.0% Molecular Formula: ClH Linear Formula: HCl Formula Weight: 36.45 MDL Number: MFCD00011324 Synonym: Muriatic acid
  spinner