Indium -In
Indium(III) acetate, 99.99% (metals basis), Alfa Aesar™
Chemical Name or Material: Indium(III) acetate CAS: 25114-58-3 Physical Form: Powder Assay Percent Range: 99.99% (metals basis) Molecular Formula: In(OOCCH3)3 Formula Weight: 291.96 MDL Number: MFCD00054347
Indium Arsenide, 99.9999% (metals basis), Alfa Aesar™
Chemical Name or Material: Indium arsenide Name Note: Hardly attacked by mineral acids CAS: 1303-11-3 Physical Form: 1.5 To 9.5mm Polycrystalline Pieces Assay Percent Range: 99.9999% (Metal basis) Molecular Formula: InAs Formula Weight: 189.74 MDL Number: MFCD00016144
Indium wire, >99.998% (metals basis), Alfa Aesar™, Puratronic™
Chemical Name or Material: Indium CAS: 7440-74-6 Physical Form: Wire, 1.0mm (0.04 in.) dia. Assay Percent Range: >99.998% (Metals basis) MDL Number: MFCD00134048
Indium Shots, 1-3mm (0.04-0.12 in.), Puratronic™, 99.999% (Metals basis), Alfa Aesar™
Chemical Name or Material: Indium CAS: 7440-74-6 Physical Form: Shots, 1-3mm (0.04-0.12 in.) Assay Percent Range: 99.999% (Metals basis) MDL Number: MFCD00134048
Indium Powder, -100 mesh, 99.9% (Metals basis), Alfa Aesar™
Chemical Name or Material: Indium CAS: 7440-74-6 Physical Form: Powder Assay Percent Range: 99.9% (Metals basis) Molecular Formula: In Formula Weight: 114.82 MDL Number: MFCD00134048
Indium(III) oxide, 99.99% (metals basis), Alfa Aesar™
Chemical Name or Material: Indium(III) oxide CAS: 1312-43-2 Physical Form: Powder/Granules Assay Percent Range: 99.994% (Metal basis) Molecular Formula: In2O3 Formula Weight: 277.64 MDL Number: MFCD00011060
Indium Wire, 0.5mm (0.02 in.) dia., Puratronic™, >99.998% (Metals basis), Alfa Aesar™
Chemical Name or Material: Indium CAS: 7440-74-6 Physical Form: Wire, 0.5mm (0.02 in.) dia. Assay Percent Range: >99.998% (Metals basis) MDL Number: MFCD00134048
Indium wire, 99.99% (metals basis), Alfa Aesar™, Puratronic™
Chemical Name or Material: Indium CAS: 7440-74-6 Physical Form: Wire, 1.0mm (0.04 in.) dia. Assay Percent Range: 99.999% (Metals basis) MDL Number: MFCD00134048
Alfa Aesar™ Indium foil, 0.25mm thick, 99.99% (metals basis), Alfa Aesar™
Chemical Name or Material: Indium CAS: 7440-74-6 MDL Number: MFCD00134048
Indium(III) acetate, 99.99%, (trace metal basis), anhydrous, ACROS Organics™
Chemical Name or Material: Indium(III) acetate CAS: 25114-58-3 Purity Grade: For trace metal basis analysis Assay: 35.3 to 43.3% (In) Assay Percent Range: 99.99% Molecular Formula: C6H9InO6 Formula Weight: 291.95
Indium spheres, 1mm (0.04 in.) dia., 99.99% (metals basis), Alfa Aesar™
Chemical Name or Material: Indium CAS: 7440-74-6 Physical Form: Spheres, 1mm (0.04 in.) dia. Assay Percent Range: 99.99% (Metals basis) MDL Number: MFCD00134048
Indium Wire, 0.25mm (0.01 in.) dia., Puratronic™, >99.998% (Metals basis), Alfa Aesar™
Chemical Name or Material: Indium CAS: 7440-74-6 Physical Form: Wire, 0.25mm (0.01 in.) dia. Assay Percent Range: >99.998% (Metals basis) MDL Number: MFCD00134048
Indium tin oxide, 99.99% (metals basis), Alfa Aesar™
Chemical Name or Material: Indium tin oxide CAS: 50926-11-9 Physical Form: Powder Assay Percent Range: 99.99% (Metal basis) Molecular Formula: In2O3:SnO2; 90:10 wt.% MDL Number: MFCD00171662
Indium(III) chloride, White to off-white crystalline, 99.99% (Metals basis), Alfa Aesar™
Chemical Name or Material: Indium(III) chloride CAS: 10025-82-8 Physical Form: Crystals Assay Percent Range: 99.99% (Metals basis) Molecular Formula: InCl3 Formula Weight: 221.18 MDL Number: MFCD00011058
Indium Shots, 4-8mm (0.2-0.3 in.), Puratronic™, 99.9999% (Metals basis), Alfa Aesar™
Chemical Name or Material: Indium CAS: 7440-74-6 Physical Form: Shots, 4-8mm (0.2-0.3 in.) Assay Percent Range: 99.9999% (Metals basis) MDL Number: MFCD00134048
Indium(III) chloride, White to off-white powder, 99.999% (Metals basis), Alfa Aesar™
Chemical Name or Material: Indium(III) chloride CAS: 10025-82-8 Physical Form: Powder Assay Percent Range: 99.999% (Metals basis) Molecular Formula: InCl3 Formula Weight: 221.18 MDL Number: MFCD00011058
Indium wire, 99.99% (metals basis), Alfa Aesar™
Chemical Name or Material: Indium wire CAS: 7440-74-6 Physical Form: ∽12.9g/m Assay Percent Range: 99.99% (metals basis) MDL Number: MFCD00134048
Indium(III) oxide, 99.9% (metals basis), Alfa Aesar™
Chemical Name or Material: Indium(III) oxide CAS: 1312-43-2 Physical Form: Powder Assay Percent Range: 99.994% (Metal basis) Molecular Formula: In2O3 Formula Weight: 277.64 MDL Number: MFCD00011060
Indium(III) nitrate hydrate, 99.99% (metals basis), Alfa Aesar™
Chemical Name or Material: Indium(III) nitrate hydrate CAS: 207398-97-8 Physical Form: Lump/Powder Assay Percent Range: 99.99% (Metal basis) Molecular Formula: InN3O9·xH2O Formula Weight: 300.83 MDL Number: MFCD00149742
Indium(III) oxide, Yellow powder, 99.994% (Metals basis), Alfa Aesar™
Chemical Name or Material: Indium(III) oxide CAS: 1312-43-2 Physical Form: Powder Assay Percent Range: 99.994% (Metals basis) Molecular Formula: In2O3 Formula Weight: 277.64 MDL Number: MFCD00011060
Indium(III) oxide, 99.9997%, (trace metal basis), ACROS Organics™ Available on GSA/VA Contract for Federal Government customers only.
Chemical Name or Material: Indium(III) oxide CAS: 1312-43-2 Purity Grade: For trace metal basis analysis Assay: 81.0 to 84.3% (In) Assay Percent Range: 100.00% Molecular Formula: In2O3 Linear Formula: In2O3 Formula Weight: 277.63
Indium(III) bromide 99.99%, ACROS Organics™ Available on GSA/VA Contract for Federal Government customers only.
Chemical Name or Material: Indium(III) bromide CAS: 13465-09-3 Purity Grade: For trace metal basis analysis Assay Percent Range: 99.99% Molecular Formula: Br3In Linear Formula: InBr3 Formula Weight: 354.52 MDL Number: MFCD00011057
Indium(III) chloride, 99.995%, (trace metal basis), ACROS Organics™ Available on GSA/VA Contract for Federal Government customers only.
Chemical Name or Material: Indium(III) chloride CAS: 10025-82-8 Purity Grade: Trace Metal Basis Assay Percent Range: 99.995% Molecular Formula: Cl3In Linear Formula: InCl3 Formula Weight: 221.18
Indium(III) oxide, Yellow pieces, 99.995% (Metals basis), Alfa Aesar™
Chemical Name or Material: Indium(III) oxide CAS: 1312-43-2 Physical Form: Pieces Assay Percent Range: 99.995% (Metals basis) Molecular Formula: In2O3 Formula Weight: 277.64 MDL Number: MFCD00011060
Indium foil, 99.99% (metals basis), Alfa Aesar™
Chemical Name or Material: Indium CAS: 7440-74-6 Physical Form: Foil, 0.127mm (0.005 in.) thick Assay Percent Range: 99.99% (Metals basis) MDL Number: MFCD00134048
Indium(III) iodide, Red to yellow crystalline, 99.994% (Metals basis), Alfa Aesar™
Chemical Name or Material: Indium(III) iodide CAS: 13510-35-5 Physical Form: Crystalline Assay Percent Range: 99.994% (Metals basis) Molecular Formula: I3In Formula Weight: 495.53 MDL Number: MFCD00016151
Indium, 99.999%, Powder, ACROS Organics™
Chemical Name or Material: Indium CAS: 7440-74-6 Purity Grade: Trace Metal Basis Assay Percent Range: 100.00% Molecular Formula: In Linear Formula: In Formula Weight: 114.82
Indium, 99.99% (trace metal basis), foil, 0.127mm thick, ACROS Organics™ Available on GSA/VA Contract for Federal Government customers only.
Chemical Name or Material: Indium CAS: 7440-74-6 Purity Grade: Trace Metal Basis Physical Form: Foil Assay Percent Range: 99.99% Molecular Formula: In Linear Formula: In Formula Weight: 114.82 MDL Number: MFCD00134048
Indium(III) iodide, 99.998%, (trace metal basis), ACROS Organics™
Chemical Name or Material: Indium(III) iodide CAS: 13510-35-5 Purity Grade: For trace metal basis analysis Assay: 22.7 to 23.5% (In) Assay Percent Range: 100.00% Molecular Formula: InI3 Linear Formula: InI3 Formula Weight: 495.53 MDL Number: MFCD00016151
Indium powder, 99.99% (metals basis), Alfa Aesar™, Puratronic™
Chemical Name or Material: Indium CAS: 7440-74-6 Physical Form: Powder Assay Percent Range: 99.999% (Metals basis) Molecular Formula: In Formula Weight: 114.82 MDL Number: MFCD00134048
  spinner