Isobutyric Acid
Isobutyric Acid, 99+%, ACROS Organics™
Chemical Name or Material: Isobutyric acid Name Note: 99+% CAS: 79-31-2 Assay: ≥99.0% Assay Percent Range: 99+% Molecular Formula: C4H8O2 Linear Formula: (CH3)2CHCO2H Formula Weight: 88.11 MDL Number: MFCD00002658 Synonym: 2-Methylpropionic acid
  spinner