Isoleucine -Ile and Derivatives
L-Isoleucine, 99%, ACROS Organics™
Chemical Name or Material: L-Isoleucine Assay: 98.5 to 101.5% Assay Percent Range: 99% Molecular Formula: C6H13NO2 Linear Formula: C2H5CH(CH3)CH(NH2)CO2H Formula Weight: 131.17 MDL Number: MFCD00064222 Synonym: (2S, 3S)-2-Amino-3-methylpentanoic acid
L-allo-Isoleucine, 99%, 99% ee
Chemical Name or Material: L-allo-Isoleucine CAS: 1509-34-8 Name Note: 99% Molecular Formula: C6H13NO2 Linear Formula: CH3CH2CH(CH3)CH(NH2)COOH Formula Weight: 131.17
FMOC-L-Isoleucine, 99%, ACROS Organics™
Chemical Name or Material: FMOC-L-Isoleucine Name Note: 99% CAS: 71989-23-6 Assay: ≥98.5% Assay Percent Range: 99% Molecular Formula: C21H23NO4 Linear Formula: FMOC-NHCH(COOH)CH(CH3)C2H5 Formula Weight: 353.42 MDL Number: MFCD00037125 Synonym: N-(9-Fluorenylmethoxycarbonyl)-L-isoleucine
L-Isoleucine, 99%
Chemical Name or Material: L-Isoleucine CAS: 73-32-5 Molecular Formula: C6H13NO2 Formula Weight: 131.18 MDL Number: MFCD00064222 Beilstein: 1721792 Merck Index: 14,5179
L-Isoleucine (White Crystals or Crystalline Powder), Fisher BioReagents™ Available on GSA/VA Contract for Federal Government customers only.
Chemical Name or Material: L-Isoleucine CAS: 73-32-5 Physical Form: Powder/Solid Assay: 98.5 to 101.0% Assay Percent Range: 98.5 to 101.0 % Molecular Formula: C6H13NO2 Formula Weight: 131.17
L-Isoleucine, 99%, MP Biomedicals
Chemical Name or Material: L-Isoleucine, 99% CAS: 73-32-5 Purity Grade: Cell Culture Reagent Grade Physical Form: Crystalline Molecular Formula: C6H13NO2 Formula Weight: 131.2
L-allo-Isoleucine, MP Biomedicals™
Chemical Name or Material: L-allo-Isoleucine Name Note: For TLC anaylsis CAS: 1509-34-8 Physical Form: Powder Molecular Formula: C6H13NO2 Formula Weight: 131.17 MDL Number: MFCD00066446 Synonym: H-allo-lle-OH, (2S, 3R)-2-Amino-3-methylpentanoic acid
DL-Isoleucine, ≥99%, MP Biomedicals™
Chemical Name or Material: DL-Isoleucine CAS: 443-79-8 Physical Form: Powder Assay Percent Range: ≥99% Molecular Formula: C6H13NO2 Formula Weight: 131.17 MDL Number: MFCD00004268 Synonym: 2-Amino-3-Methylpentanoic Acid
D-Isoleucine 98%, ACROS Organics™
Chemical Name or Material: D-Isoleucine Name Note: 98% CAS: 319-78-8 Assay: ≥97.5% Assay Percent Range: 98% Molecular Formula: C6H13NO2 Linear Formula: CH3CH2CH(CH3)CH(NH2)COOH Formula Weight: 131.17 MDL Number: MFCD00064221 Synonym: (2R, 3R)-2-Amino-3-methylpentanoic acid
  spinner