Isooctane
2,2,4-Trimethylpentane, 99+%, ACROS Organics™ Available on GSA/VA Contract for Federal Government customers only.
Chemical Name or Material: 2, 2, 4-Trimethylpentane Name Note: For analysis ACS CAS: 540-84-1 CAS Min %: 99.0 CAS Max %: 100.0 Purity Grade: ACS Reagent Grade Assay: ≥99.0% Assay Percent Range: 99+% Molecular Formula: C8H18 Linear Formula: (CH3)2CHCH2C(CH3)3 Formula Weight: 114.23 MDL Number: MFCD00008943 Synonym: Isooctane
2,2,4-Trimethylpentane, Reagent ACS, Spectro Grade, +99%, ACROS Organics™ Available on GSA/VA Contract for Federal Government customers only.
Chemical Name or Material: 2, 2, 4-Trimethylpentane CAS: 540-84-1 CAS Min %: 99.0 CAS Max %: 100.0 Purity Grade: For spectroscopy ACS Assay: ≥99.0% Assay Percent Range: 99+% Molecular Formula: C8H18 Linear Formula: (CH3)2CHCH2C(CH3)3 Formula Weight: 114.23 MDL Number: MFCD00008943 Synonym: Isooctane
2,2,4-Trimethylpentane, 99.5%, for residue analysis, ECD tested for pesticide analysis, ACROS Organics™ Available on GSA/VA Contract for Federal Government customers only.
Chemical Name or Material: 2, 2, 4-Trimethylpentane Name Note: ECD Tested, for Pesticide Analysis CAS: 540-84-1 CAS Min %: 99.4 CAS Max %: 100.0 Purity Grade: Residue Analysis Grade Assay: ≥99.5% Assay Percent Range: 99.50% Molecular Formula: C8H18 Linear Formula: (CH3)CHCHC(CH3)3 Formula Weight: 114.23 MDL Number: MFCD00008943 Synonym: Isooctane
Isooctane (HPLC), Fisher Chemical Available on GSA/VA Contract for Federal Government customers only.
Chemical Name or Material: Isooctane CAS: 540-84-1 Purity Grade: HPLC Grade Purity Grade Notes: Also meets ACS specifications Assay: ≥99% Assay Percent Range: ≥99 % Molecular Formula: C8H18 Formula Weight: 114.23 MDL Number: MFCD00008943 Synonym: 2, 2, 4-Trimethylpentane
2,2,4-Trimethylpentane, Pesticide Residueanalysis Grade, ACROS Organics™
Chemical Name or Material: 2, 2, 4-Trimethylpentane CAS: 540-84-1 Purity Grade: Residue Analysis Grade Molecular Formula: C8H18 Linear Formula: (CH3)2CHCH2C(CH3)3 Formula Weight: 114.23 MDL Number: MFCD00008943 Synonym: Isooctane
2,2,4-Trimethylpentane, 99.5%, for HPLC, ACROS Organics™
Chemical Name or Material: 2, 2, 4-Trimethylpentane CAS: 540-84-1 CAS Min %: 99.4 CAS Max %: 100.0 Purity Grade: HPLC Grade Assay: ≥99.4% (GC) Assay Percent Range: 99.5% Molecular Formula: C8H18 Linear Formula: (CH3)2CHCH2C(CH3)3 Formula Weight: 114.23 MDL Number: MFCD00008943 Synonym: Isooctane
2,2,4-Trimethylpentane, 99.5%, for analysis, ACROS Organics™
Chemical Name or Material: 2, 2, 4-Trimethylpentane CAS: 540-84-1 CAS Min %: 99.4 CAS Max %: 100.0 Purity Grade: For analysis Assay: ≥99.5% Assay Percent Range: 99.50% Molecular Formula: C8H18 Linear Formula: (CH3)2CHCH2C(CH3)3 Formula Weight: 114.23 MDL Number: MFCD00008943 Synonym: Isooctane
2,2,4-Trimethylpentane, 99.5%, Extra Dry, AcroSeal™, ACROS Organics™
Chemical Name or Material: 2, 2, 4-Trimethylpentane CAS: 540-84-1 CAS Min %: 99.4 CAS Max %: 100.0 Purity Grade: Extra Dry Assay: ≥99.4% (GC) Assay Percent Range: 99.50% Molecular Formula: C8H18 Linear Formula: (CH3)CHCHC(CH3)3 Formula Weight: 114.23 MDL Number: MFCD00008943 Synonym: Isooctane
Isooctane (Pesticide), Fisher Chemical
Chemical Name or Material: Isooctane CAS: 540-84-1 Purity Grade: Pesticide Grade Molecular Formula: C8H18 Linear Formula: (CH3)3CCH2CH(CH3)2 Formula Weight: 114.23 MDL Number: MFCD00008943 Synonym: 2, 2, 4-Trimethylpentane
2,2,4-Trimethylpentane, HPLC Grade 99.0%, ACROS Organics™
Chemical Name or Material: 2, 2, 4-Trimethylpentane Name Note: 99.0% CAS: 540-84-1 Purity Grade: HPLC Grade Assay: ≥99.0% Assay Percent Range: 99% Molecular Formula: C8H18 Linear Formula: (CH3)2CHCH2C(CH3)3 Formula Weight: 114.23 MDL Number: MFCD00008943 Synonym: Isooctane
2,2,4-Trimethylpentane, 99.5%, Extra Dry over Molecular Sieve, AcroSeal™, ACROS Organics™ Available on GSA/VA Contract for Federal Government customers only.
Chemical Name or Material: 2, 2, 4-Trimethylpentane Name Note: Over Molecular Sieves CAS: 540-84-1 CAS Min %: 99.4 CAS Max %: 100.0 Purity Grade: Extra Dry over Molecular Sieve Assay: ≥99.4% (GC) Assay Percent Range: 99.5% Molecular Formula: C8H18 Linear Formula: (CH3)2CHCH2C(CH3)3 Formula Weight: 114.23 MDL Number: MFCD00008943 Synonym: Isooctane
2,2,4-Trimethylpentane, 99%, for spectroscopy, ACROS Organics™
Chemical Name or Material: 2, 2, 4-Trimethylpentane Name Note: 99% CAS: 540-84-1 CAS Min %: 98.5 CAS Max %: 100.0 Purity Grade: For spectroscopy Assay: ≥98.5% Assay Percent Range: 99% Molecular Formula: C8H18 Linear Formula: (CH3)2CHCH2C(CH3)3 Formula Weight: 114.23 MDL Number: MFCD00008943 Synonym: Isooctane
Isooctane (Optima™), Fisher Chemical
Chemical Name or Material: Isooctane CAS: 540-84-1 Purity Grade: OPTIMA™ Purity Grade Notes: High purity mobile phase for HPLC and/or extraction solvent for GC applications Assay: ≥99% Assay Percent Range: 99% min by GC Molecular Formula: C8H18 Linear Formula: (CH3)3CCH2CH(CH3)2 Formula Weight: 114.23 MDL Number: MFCD00008943 Synonym: 2, 2, 4-Trimethylpentane
Isooctane (Certified ACS), Fisher Chemical Available on GSA/VA Contract for Federal Government customers only.
Chemical Name or Material: Isooctane CAS: 540-84-1 Purity Grade: Certified ACS Reagent Grade Assay: ≥99% Assay Percent Range: ≥99 % Molecular Formula: C8H18 Linear Formula: (CH3)3CCH2CH(CH3)2 Formula Weight: 114.23 MDL Number: MFCD00008943 Synonym: 2, 2, 4-Trimethylpentane
2,2,4-Trimethylpentane, 99+%, pure, ACROS Organics™ Available on GSA/VA Contract for Federal Government customers only.
Chemical Name or Material: 2, 2, 4-Trimethylpentane CAS: 540-84-1 CAS Min %: 99.0 CAS Max %: 100.0 Purity Grade: Pure Assay: ≥99.0% Assay Percent Range: 99+% Molecular Formula: C8H18 Linear Formula: (CH3)2CHCH2C(CH3)3 Formula Weight: 114.23 MDL Number: MFCD00008943 Synonym: Isooctane
2,2,4-Trimethylpentane ChromAR, Macron Fine Chemicals™
Chemical Name or Material: 2,2,4-Trimethylpentane CAS: 540-84-1 Molecular Formula: (CH3)3CCH2CH(CH3)2 Formula Weight: 114.23
2,2,4-Trimethylpentane UltimAR, Macron Fine Chemicals™
Chemical Name or Material: 2,2,4-Trimethylpentane CAS: 540-84-1 Molecular Formula: (CH3)3CCH2CH(CH3)2 Formula Weight: 114.23
2,2,4-Trimethylpentane, EMD Millipore Available on GSA/VA Contract for Federal Government customers only.
Chemical Name or Material: 2, 2, 4-Trimethylpentane Name Note: SeccoSolv™, Dried CAS: 540-84-1 Molecular Formula: C8H18 Linear Formula: CH3C(CH3)2CH2CH(CH3)CH3 Formula Weight: 114.23 Synonym: 2, 2, 4-Trimethylpentane
2,2,4-Trimethylpentane, 9.0% min., ACS, EMD Millipore Available on GSA/VA Contract for Federal Government customers only.
Chemical Name or Material: 2, 2, 4-Trimethylpentane Name Note: For ACS analysis CAS: 540-84-1 Purity Grade: ACS/NF/USP Physical Form: Liquid Assay Percent Range: 99.0% min. Molecular Formula: (CH3)3CCH2CH(CH3)2 Linear Formula: (CH3)3CCH2CH(CH3)2 Formula Weight: 114.23 Synonym: Isooctane
2,2,4-Trimethylpentane, 99+%, pure, ACROS Organics™
Chemical Name or Material: 2, 2, 4-Trimethylpentane Name Note: 99+% CAS: 540-84-1 CAS Min %: 99.0 CAS Max %: 100.0 Purity Grade: Pure Assay: ≥99.0% Assay Percent Range: 99+% Molecular Formula: C8H18 Linear Formula: (CH3)2CHCH2C(CH3)3 Formula Weight: 114.23 MDL Number: MFCD00008943 Synonym: Isooctane
  spinner