Isotopically Labeled Compounds
Chloroform-d, for NMR, 99.6+% atom D, ACROS Organics™
Chemical Name or Material: Chloroform-d Name Note: 99.6+ atom % D CAS: 865-49-6 Purity Grade: For NMR Molecular Formula: CCl3D Linear Formula: CDCl3 Formula Weight: 120.39 MDL Number: MFCD00000827
Methyl sulfoxide-d6, for NMR, +99% atom D, ACROS Organics™
Chemical Name or Material: Methyl sulfoxide-d6 CAS: 2206-27-1 Purity Grade: For NMR Molecular Formula: C2D6OS Formula Weight: 84.18 MDL Number: MFCD00002090 Synonym: Dimethyl-d6 sulfoxide
Chloroform-d, for NMR, 99.8% atom D, ACROS Organics™ Available on GSA/VA Contract for Federal Government customers only.
Chemical Name or Material: Chloroform-d Name Note: 99.8 atom % D CAS: 865-49-6 Purity Grade: For NMR Molecular Formula: CCl3D Linear Formula: CDCl3 Formula Weight: 120.39 MDL Number: MFCD00000827
Ethylbenzene-d10, for NMR, packaged in ampoules, 99+% atom D, ACROS Organics™
Chemical Name or Material: Ethylbenzene-d10 Name Note: 99+ atom% D CAS: 25837-05-2 Purity Grade: For NMR Molecular Formula: C8D10 Linear Formula: C6D5C2D5 Formula Weight: 116.25 MDL Number: MFCD00044343
Methyl sulfoxide-d6, for NMR, 99.8 atom % D, ACROS Organics™ Available on GSA/VA Contract for Federal Government customers only.
Chemical Name or Material: Methyl sulfoxide-d6 Name Note: 99.8 atom % D CAS: 2206-27-1 Purity Grade: For NMR Molecular Formula: C2D6OS Linear Formula: (CD3)2SO Formula Weight: 84.18 MDL Number: MFCD00002090 Synonym: Dimethyl-d6 sulfoxide
Nitrobenzene-d5, for NMR, 99% atom D, ACROS Organics™
Chemical Name or Material: Nitrobenzene-d5 Name Note: 99 atom % D CAS: 4165-60-0 Purity Grade: For NMR Molecular Formula: C6D5NO2 Linear Formula: C6D5NO2 Formula Weight: 128.15 MDL Number: MFCD00044415
Acetone-d6, for NMR, +99% atom D, ACROS Organics™ Available on GSA/VA Contract for Federal Government customers only.
Chemical Name or Material: Acetone-d6 Name Note: 99+ atom % D CAS: 666-52-4 Purity Grade: For NMR Molecular Formula: C3D6O Linear Formula: CD3COCD3 Formula Weight: 64.13 MDL Number: MFCD00044635
Chloroform-d, for NMR, 99.8+% D, ACROS Organics™, stab. with silver foil, 0.03 v/v% TMS, ACROS Organics™ Available on GSA/VA Contract for Federal Government customers only.
Chemical Name or Material: Chloroform-d Name Note: 99.8+ atom% D CAS: 865-49-6 Purity Grade: For NMR Molecular Formula: CCl3D Linear Formula: CDCl3 Formula Weight: 120.39 MDL Number: MFCD00000827
n-Hexane-d14, for NMR, packaged in ampoules, 99% atom D, ACROS Organics™
Chemical Name or Material: n-Hexane-d14 Name Note: 99 atom% D, for NMR CAS: 21666-38-6 Purity Grade: For NMR Molecular Formula: C6D14 Linear Formula: D3C(CD2)4CD3 Formula Weight: 100.28 MDL Number: MFCD00037562
Methanol-d4, for NMR, with 0.03% TMS, in 0.75mL ampoules, 99.8% atom D, ACROS Organics™
Chemical Name or Material: Methanol-d4 Name Note: 99.8 atom% D CAS: 811-98-3 Purity Grade: For NMR Molecular Formula: CD4O Linear Formula: CD3OD Formula Weight: 36.06 MDL Number: MFCD00044637
Acetonitrile-d3, for NMR, packaged in 0.75mL ampoules, 99.6% atom D, ACROS Organics™
Chemical Name or Material: Acetonitrile-d3 Name Note: 99.6 atom % D CAS: 2206-26-0 Purity Grade: For NMR Molecular Formula: C2D3N Linear Formula: CD3CN Formula Weight: 44.07 MDL Number: MFCD00001881
Acetone-d6, for NMR, 99.5 atom % D, ACROS Organics™
Chemical Name or Material: Acetone-d6 Name Note: 99.5 atom % D CAS: 666-52-4 Purity Grade: For NMR Molecular Formula: C3D6O Linear Formula: CD3COCD3 Formula Weight: 64.13 MDL Number: MFCD00044635
Methanol-d4, for NMR, 99.8 atom% D, AcroSeal™, ACROS Organics™
Chemical Name or Material: Methanol-d4 Name Note: For NMR, 99.8 atom% D CAS: 811-98-3 Purity Grade: For NMR Molecular Formula: CD4O Linear Formula: CD3OD Formula Weight: 36.06 MDL Number: MFCD00044637 Synonym: Methyl-d3 alcohol-d
Deuterium oxide, for NMR, 100.0% atom D, ACROS Organics™ Available on GSA/VA Contract for Federal Government customers only.
Chemical Name or Material: Deuterium oxide Name Note: 100.0 atom % D CAS: 7789-20-0 Purity Grade: For NMR Molecular Formula: D2O Linear Formula: D2O Formula Weight: 20.02 MDL Number: MFCD00044636 Synonym: Water, heavy
Methanol-d4, for NMR, 99.5% atom D, ACROS Organics™ Available on GSA/VA Contract for Federal Government customers only.
Chemical Name or Material: Methanol-d4 Name Note: 99.5 atom % D CAS: 811-98-3 Purity Grade: For NMR Molecular Formula: CD4O Linear Formula: CD3OD Formula Weight: 36.06 MDL Number: MFCD00044637
Pyridine-d5, for NMR, packaged in 0.75mL ampoules, 99.8 atom% D, ACROS Organics™
Chemical Name or Material: Pyridine-d5 Name Note: 99.8 atom% D CAS: 7291-22-7 Purity Grade: For NMR Molecular Formula: C5D5N Formula Weight: 84.14 MDL Number: MFCD00044639
Chloroform-d, for NMR, 100% atom D, ACROS Organics™, packaged in 7.5mL ampoules, ACROS Organics™
Chemical Name or Material: Chloroform-d Name Note: 100 atom % D CAS: 865-49-6 Purity Grade: For NMR Molecular Formula: CCl3D Linear Formula: CDCl3 Formula Weight: 120.39 MDL Number: MFCD00000827
Nitromethane-d3, for NMR, 99 atom % D, ACROS Organics™
Chemical Name or Material: Nitromethane-d3 Name Note: 99 atom % D CAS: 13031-32-8 Purity Grade: For NMR Molecular Formula: CD3NO2 Linear Formula: CD3NO2 Formula Weight: 64.06 MDL Number: MFCD00044214
Pyridine-d5, for NMR, with 0.03% TMS, 100.0 atom% D, ACROS Organics™
Chemical Name or Material: Pyridine-d5 Name Note: 100.0 atom% D CAS: 7291-22-7 CAS: 75-76-3 Purity Grade: For NMR Molecular Formula: C5D5N Formula Weight: 84.14 MDL Number: MFCD00044639
Sodium deuteroxide, for NMR, 40 wt.% solution in D2O, 99+% atom D, ACROS Organics™ Available on GSA/VA Contract for Federal Government customers only.
Chemical Name or Material: Sodium deuteroxide Name Note: 99+ atom % D CAS: 14014-06-3 CAS: 7789-20-0 Purity Grade: For NMR Molecular Formula: DNaO Linear Formula: NaOD Formula Weight: 41 MDL Number: MFCD00037669
Chloroform-d, for NMR, 99.8+% atom D, contains 0.03 v/v% TMS, ACROS Organics™ Available on GSA/VA Contract for Federal Government customers only.
Chemical Name or Material: Chloroform-d Name Note: 99.8+ atom % D CAS: 865-49-6 Purity Grade: For NMR Molecular Formula: CCl3D Linear Formula: CDCl3 Formula Weight: 120.39 MDL Number: MFCD00000827
Acetone-d6, for NMR, contains 1 v/v% TMS, 99.5% atom D, ACROS Organics™ Available on GSA/VA Contract for Federal Government customers only.
Chemical Name or Material: Acetone-d6 Name Note: 99.5 atom % D CAS: 666-52-4 CAS Min %: 99.4 CAS Max %: 100.0 Purity Grade: For NMR Molecular Formula: C3D6O Linear Formula: CD3COCD3 Formula Weight: 64.13 MDL Number: MFCD00044635
Methanol-d4, for NMR, contains 0.03 v/v% TMS, 99.8%% atom D, AcroSeal™, ACROS Organics™
Chemical Name or Material: Methanol-d4 Name Note: For NMR, contains 0.03 v/v% TMS, 99.8% atom% D CAS: 811-98-3 Purity Grade: For NMR Molecular Formula: CD4O Linear Formula: CD3OD Formula Weight: 36.06 MDL Number: MFCD00044637 Synonym: Methyl-d3 alcohol-d
1,1,2,2-Tetrachloroethane-d2, for NMR, 99.5+ atom% D, ACROS Organics™ Available on GSA/VA Contract for Federal Government customers only.
Chemical Name or Material: 1,1,2,2-Tetrachloroethane-d2 Name Note: 99.5+ atom% D, for NMR CAS: 33685-54-0 Purity Grade: For NMR Molecular Formula: C2Cl4D2 Linear Formula: (Cl2CD)2 Formula Weight: 169.86 MDL Number: MFCD00037672
Deuterium oxide, for NMR, packaged in 0.75mL ampoules, 99.9% atom D, ACROS Organics™
Chemical Name or Material: Deuterium oxide Name Note: 99.9 atom % D CAS: 7789-20-0 Purity Grade: For NMR Molecular Formula: D2O Linear Formula: D2O Formula Weight: 20.02 MDL Number: MFCD00044636 Synonym: Water, heavy
Toluene-d8, for NMR, +99% atom D, ACROS Organics™ Available on GSA/VA Contract for Federal Government customers only.
Chemical Name or Material: Toluene-d8 Name Note: 99+ atom % D CAS: 2037-26-5 Purity Grade: For NMR Molecular Formula: C7D8 Linear Formula: C6D5CD3 Formula Weight: 100.21 MDL Number: MFCD00044638 Synonym: Methylbenzene-d8
n-Hexane-d14, for NMR, +99.5% atom D, ACROS Organics™
Chemical Name or Material: n-Hexane-d14 Name Note: 99.5+ atom % D CAS: 21666-38-6 Purity Grade: For NMR Molecular Formula: C6D14 Linear Formula: D3C(CD2)4CD3 Formula Weight: 100.28 MDL Number: MFCD00037562
Pyridine-d5, for NMR, contains 1 v/v% TMS, 99 atom % D, ACROS Organics™
Chemical Name or Material: Pyridine-d5 Name Note: 99 atom % D CAS: 7291-22-7 CAS: 75-76-3 Purity Grade: For NMR Molecular Formula: C5D5N Formula Weight: 84.14 MDL Number: MFCD00044639
Methyl sulfoxide-d6, for NMR, 99.9% atom D, ACROS Organics™ Available on GSA/VA Contract for Federal Government customers only.
Chemical Name or Material: Methyl sulfoxide-d6 Name Note: 99.9 atom % D CAS: 2206-27-1 Purity Grade: For NMR Molecular Formula: C2D6OS Linear Formula: (CD3)2SO Formula Weight: 84.18 MDL Number: MFCD00002090 Synonym: Dimethyl-d6 sulfoxide
Iodomethane-d3, for NMR, 99+ atom % D, ACROS Organics™ Available on GSA/VA Contract for Federal Government customers only.
Chemical Name or Material: Iodomethane-d3 Name Note: 99+ atom % D CAS: 865-50-9 Purity Grade: For NMR Molecular Formula: CD3I Linear Formula: CD3I Formula Weight: 144.96 MDL Number: MFCD00001074
  spinner