Karl Fischer -KF Reagents
Karl Fischer Aqualine™ Complete 5, for Karl Fischer Titration By Volumetry, Fisher Chemical
CAS: 111-90-0 CAS: 288-32-4 CAS: 7446-09-5 CAS: 7553-09-5 ChemAlert Storage Symbol: Gray Chemical Name or Material: Aqualine™ Complete 5 Cleanup Symbol: Solvent Color: Brown Physical Form: Liquid Equivalence: 5.2mg of water/mL min. Health Hazard 1: Signal Word:DANGER! Health Hazard 2: DANGER! Health Hazard 3: Emergency Overview
Causes burns by all exposure routes. Harmful by inhalation. Possible risk of harm to the unborn child.
NFPA
Health: Name Note: Karl Fischer Reagent Purity or Free of Statement: Pyridine free
Aqualine™ Water Standard in Methanol 5.0, Karl Fischer, Fisher Chemical
Chemical Name or Material: Aqualine™ Water in Methanol Standard 5.1 Name Note: Karl Fischer Reagent CAS: 67-56-1 MDL Number: MFCD00081734
Karl Fischer Reagent (Stabilized), Fisher Chemical
Chemical Name or Material: Karl Fischer Reagent CAS: 109-86-4 CAS: 110-86-1 CAS: 7446-09-5 CAS: 7553-56-2 Physical Form: Liquid MDL Number: MFCD00081734
Karl Fischer Aqualine™ Solvent, for Karl Fischer Titration By Volumetry, Fisher Chemical
CAS: 67-56-1 CAS: 288-32-4 CAS: 7446-09-5 ChemAlert Storage Symbol: Red Chemical Name or Material: Aqualine™ Solvent Cleanup Symbol: Solvent Color: Clear colorless Physical Form: Liquid DOT Information: DOT Class 3, : Flammable Liquid Health Hazard 1: GHS Signal Word: Danger MDL Number: 81734 Name Note: Karl Fischer Reagent Packaging: Amber glass bottle Purity or Free of Statement: Pyridine free UN Number: 1992 Use Test: Pass Test Vapor Density: > 1.0
Karl Fischer Aqualine™ Solvent CM, for Karl Fischer Titration By Volumetry, Fisher Chemical
CAS: 67-56-1 CAS: 67-66-3 CAS: 288-32-4 CAS: 7446-09-5 ChemAlert Storage Symbol: Red Chemical Name or Material: Aqualine™ Solvent CM Cleanup Symbol: Solvent Physical Form: Liquid DOT Information: DOT Class 3, : Flammable Liquid MDL Number: 81734 Name Note: Karl Fischer Reagent Packaging: Amber glass bottle Purity or Free of Statement: Pyridine free Use Test: Pass Test
Karl Fischer Aqualine™ Complete 1, for Karl Fischer Titration By Volumetry, Fisher Chemical
CAS: 111-90-0 CAS: 288-32-4 CAS: 7446-09-5 CAS: 7553-56-2 ChemAlert Storage Symbol: Gray Chemical Name or Material: Aqualine™ Complete 1 Cleanup Symbol: Solvent Color: Clear colorless Physical Form: Liquid Equivalence: 1.2mg of water/mL min. Health Hazard 1: Signal Word:DANGER! Health Hazard 2: DANGER! Health Hazard 3: Emergency Overview
Irritating to skin. Irritating to eyes. Possible risk of harm to the unborn child.
NFPA
Health: 1 Molecular Formula: Solution Name Note: Karl Fischer Reagent Purity or Free of Statement: Pyridine free
Karl Fischer Aqualine™ Electrolyte A, for Karl Fischer Titration By Coulometry, Fisher Chemical
Chemical Name or Material: Aqualine™ Electrolyte A Name Note: Karl Fischer Reagent CAS: 67-56-1 CAS: 67-66-3 CAS: 108-75-8 CAS: 111-42-2 CAS: 7446-09-5 Physical Form: Liquid MDL Number: 81734
Karl Fischer reagent diluent, ACROS Organics™
Chemical Name or Material: Karl Fischer™ reagent diluent CAS: 109-86-4 MDL Number: MFCD00081734
Karl Fischer reagent, stabilized, ACROS Organics™
Chemical Name or Material: Karl Fischer reagent CAS: 7553-56-2 MDL Number: MFCD00081734
Coulometric Conditioner Solution, Fisher Chemical
Chemical Name or Material: Coulometric Conditioner Solution CAS: 67-56-1 Molecular Formula: CH4O MDL Number: MFCD00211835
Karl Fischer Aqualine™ Water Standard 10mg, for Karl Fischer Titration, Fisher Chemical
Chemical Name or Material: Aqualine™ Standard 10.1 Name Note: Karl Fischer Reagent CAS: 71-36-3 MDL Number: 240423
Karl Fischer Reagent Diluent, Fisher Chemical
Chemical Name or Material: Karl Fischer Reagent Diluent CAS: 109-86-4 CAS: 110-86-1 MDL Number: MFCD00081734
Karl Fischer Aqualine™ Sodium Tartrate-2H2O, for Karl Fischer Titration, Fisher Chemical
Chemical Name or Material: Aqualine™ Sodium Tartrate Dihydrate Water Standard Name Note: Karl Fischer Reagent CAS: 6106-24-7 Physical Form: Powder/Solid MDL Number: 3101471
Karl Fischer Aqualine™ Titrant 5, for Karl Fischer Titration By Volumetry, Fisher Chemical
CAS: 67-56-1 ChemAlert Storage Symbol: Red Chemical Name or Material: Aqualine™ Titrant 5 Cleanup Symbol: Solvent Boiling Point: 64°C Color: Clear colorless Physical Form: Liquid DOT Information: DOT Class 3, : Flammable Liquid Equivalence: 5.0mg of water/mL min. Health Hazard 2: DANGER! Health Hazard 3: Emergency Overview
Poison, may be fatal or cause blindness if swallowed. Vapor harmful. Toxic by inhalation, in contact with skin and if swallowed. Irritating to eyes and skin. May cause central nervous system effects. Aspiration hazard if swallowed - can enter lungs and cause damage. Danger of very serious irreversible effects. WARNING! This product contains a chemical known in the State of California to cause birth defects or. Wear personal protective equipment. Keep away from open flames, hot surfaces and sources of ignition. Use only under a chemical fume hood. Wash off immediately with plenty of water for at least 15 minutes. Wash off immediately with plenty of water for at least 15 minutes. Obtain medical attention.
NFPA
Health:2
Flammability:3
Instability:0 MDL Number: 132730 Name Note: Karl Fischer Reagent Packaging: Amber glass bottle Purity or Free of Statement: Pyridine free UN Number: 1992 Vapor Density: > 1.0
Water Standard in Methanol (1mL = 1mg Water/Certified), Fisher Chemical
Chemical Name or Material: Water Standard in Methanol CAS: 67-56-1 Purity Grade: Certified Grade Molecular Formula: CH4O MDL Number: MFCD00011332
Karl Fischer Aqualine™ Water Standard 1.0mg, for Karl Fischer Titration, Fisher Chemical
Chemical Name or Material: Aqualine™ Standard 1.1 Name Note: Karl Fischer Reagent CAS: 100-66-3 CAS: 105-13-5 MDL Number: 240423
Aqualine™ Water Standard 5.0, Karl Fischer, Fisher Chemical
Chemical Name or Material: Aqualine™ Standard 5.1 Name Note: Karl Fischer Reagent CAS: 71-36-3 CAS: 1330-20-7 MDL Number: MFCD00077264
Karl Fischer Aqualine™ Electrolyte CG, for Karl Fischer Titration By Coulometry, Fisher Chemical
Chemical Name or Material: Aqualine™ Electrolyte CG Name Note: Karl Fischer Reagent CAS: 67-56-1 CAS: 108-75-8 CAS: 7446-09-5 CAS: 7553-56-2 Physical Form: Liquid MDL Number: 3097388
Karl Fischer Aqualine™ Electrolyte AG, for Karl Fischer Titration By Coulometry, Fisher Chemical
Chemical Name or Material: Aqualine™ Electrolyte AG Name Note: Karl Fischer Reagent CAS: 67-56-1 CAS: 108-75-8 CAS: 111-42-2 CAS: 7446-09-5 CAS: 7553-56-2 Physical Form: Liquid MDL Number: 3097387
CombiMethanol, 99.8% min, AQUASTAR™, EMD Millipore Available on GSA/VA Contract for Federal Government customers only.
Chemical Name or Material: CombiMethanol CAS: 67-56-1 Physical Form: Liquid Assay Percent Range: 99.8% min. Molecular Formula: CH3OH Formula Weight: 32.04
AquaStar Solvent MC, EMD Millipore Available on GSA/VA Contract for Federal Government customers only.
Chemical Name or Material: AquaStar™ Solvent MC Name Note: Reagent for Volumetric Karl Fischer Titrations CAS: 67-56-1 (Methanol) CAS: 67-66-3 (Chloroform)
AquaStar Coulomat A (Original Formulation), EMD Millipore Available on GSA/VA Contract for Federal Government customers only.
Chemical Name or Material: AQUASTAR Coulomat A Name Note: Original Formulation CAS: 7553-56-2 Physical Form: Liquid Synonym: Coulometric Anode Solution
CombiSolvent Oils, AQUASTAR™, EMD Millipore Available on GSA/VA Contract for Federal Government customers only.
Chemical Name or Material: CombiSolvent Oils Name Note: AQUASTAR™, Karl Fischer Solvent for Mineral Oils Physical Form: Liquid
Solvent MC, AQUASTAR™, EMD Millipore Available on GSA/VA Contract for Federal Government customers only.
Chemical Name or Material: Solvent MC Physical Form: Liquid
AQUASTAR™ Titrant 5, EMD Millipore™
Chemical Name or Material: AQUASTAR Titrant 5 CAS: 67-56-1,7553-56-2 CAS Min %: 50.0 CAS: 7553-56-2 CAS Min %: 10.0 Physical Form: Liquid
AquaStar Coulomat A (Safer Formulation), EMD Millipore Available on GSA/VA Contract for Federal Government customers only.
Chemical Name or Material: AQUASTAR™ Coulomat A Name Note: Safer Formulation CAS: 7553-56-2
Coulomat AK Anode Solution, AQUASTAR™, EMD Chemicals
Physical Form: Liquid Synonym: Coulometric Anode Solution
  spinner