Lactic Acid
Lactic acid, ACS, 85.0-90.0% aq. soln.
Chemical Name or Material: Lactic acid CAS: 50-21-5 Name Note: For analysis ACS Purity Grade: ACS Grade Molecular Formula: C3H6O3 Formula Weight: 90.08 MDL Number: MFCD00004520 Beilstein: 1209341 Merck Index: 14,5336 Synonym: 2-Hydroxypropionic acid
Lactic Acid (USP/FCC), Fisher Chemical
Chemical Name or Material: Lactic Acid CAS: 50-21-5, 97-73-4 CAS: 97-73-4 Purity Grade: FCC/USP Assay: 95.0 to 105.0% (FCC) Molecular Formula: C3H6O3 Linear Formula: CH3CH(OH)CO2H Formula Weight: 90.08 MDL Number: MFCD00004520 Synonym: 2-Hydroxypropanoic Acid
Lactic acid, lithium salt, 99%, ACROS Organics™
Chemical Name or Material: Lactic acid, lithium salt CAS: 867-55-0 Purity Grade: Extra Pure Assay: ≥98.5% Assay Percent Range: 99% Molecular Formula: C3H5LiO3 Linear Formula: CH3CH(OH)CO2Li Formula Weight: 96.01 MDL Number: MFCD00065399 Synonym: Lactic acid lithium salt
DL-Lactic acid, 85+%, ACS reagent, ACROS Organics™ Available on GSA/VA Contract for Federal Government customers only.
Chemical Name or Material: DL-Lactic acid CAS: 50-21-5 CAS: 7732-18-5 Purity Grade: ACS Reagent Grade Assay: 85.0 to 90.0% Assay Percent Range: 85+% Molecular Formula: C3H6O3 Linear Formula: CH3CH(OH)CO2H Formula Weight: 90.08 MDL Number: MFCD00004520 Synonym: DL-2-Hydroxypropanoic acid
L(+)-Lactic acid, 20 wt.% solution in water, ACROS Organics™
Chemical Name or Material: L(+)-Lactic acid CAS: 79-33-4 CAS: 7732-18-5 Assay: ≥18% Molecular Formula: C3H6O3 Formula Weight: 90.08 MDL Number: MFCD00064266 Synonym: (S)-(+)-2-Hydroxypropanoic acid
DL-Lactic acid, 85%, ACROS Organics™ Available on GSA/VA Contract for Federal Government customers only.
Chemical Name or Material: DL-Lactic acid CAS: 50-21-5 CAS: 7732-18-5 Assay: ≥85% Assay Percent Range: 85% Molecular Formula: C3H6O3 Linear Formula: CH3CH(OH)CO2H Formula Weight: 90.08 MDL Number: MFCD00004520 Synonym: DL-2-Hydroxypropanoic acid
L(+)-Lactic acid, 90% solution in water, ACROS Organics™
Chemical Name or Material: L(+)-Lactic acid Assay: ≥88% Molecular Formula: C3H6O3 Linear Formula: CH3CH(OH)CO2H Formula Weight: 90.08 MDL Number: MFCD00064266 Synonym: (S)-(+)-2-Hydroxypropanoic acid
Lactic Acid (85% w/w/Certified ACS), Fisher Chemical
Chemical Name or Material: Lactic Acid Name Note: 85% (w/w) CAS: 50-21-5, 97-73-4 CAS: 97-73-4 Purity Grade: Certified ACS Grade Assay: 85.0 to 90.0% Assay Percent Range: 85.0 to 90.0% Molecular Formula: C3H6O3 Linear Formula: CH3CH(OH)COOH MDL Number: MFCD00004520 Synonym: 2-Hydroxypropanoic Acid
Lactic Acid Products for Science Education
Chemical Name or Material: Lactic Acid, 85% CAS: 50-21-5 CAS: 7732-18-5 Assay Percent Range: >85% w/v Linear Formula: CH3CHOHCOOH Formula Weight: 90.08 Synonym: 2-Hydroxypropanoic Acid
Lactic Acid, 0.100 Normal, Ricca Chemical
Chemical Name or Material: Lactic Acid CAS: 7732-18-5 CAS Min %: 97.0 CAS Max %: 100.0 CAS: 50-21-5 CAS Min %: 0.99 CAS Max %: 1.04 Physical Form: Liquid
Lactic Acid, 10% (v/v) Aqueous Solution, Ricca Chemical
Chemical Name or Material: Lactic Acid CAS: 7732-18-5 CAS Min %: 89.0 CAS Max %: 91.0 CAS: 50-21-5 CAS Min %: 9.0 CAS Max %: 11.0
Lactic Acid, 1.00N, Ricca Chemical
Chemical Name or Material: Lactic Acid
L-(+)-Lactic Acid (Free Acid), Fisher BioReagents
Chemical Name or Material: L-(+)-Lactic Acid Name Note: Free acid CAS: 79-33-4 Physical Form: Powder/Solid Assay Percent Range: ≥98 % Molecular Formula: C3H6O3 Formula Weight: 90.08 Synonym: (2-Hydroxpropionic acid) Free acid
Lactic Acid, 1.00 Normal, Ricca Chemical
Chemical Name or Material: Lactic Acid CAS: 7732-18-5 CAS Min %: 89.0 CAS Max %: 93.0 CAS: 50-21-5 CAS Min %: 9.12 CAS Max %: 9.49 Physical Form: Liquid
Lactic Acid, 20% (w/v), Ricca Chemical
Chemical Name or Material: Lactic Acid CAS: 7732-18-5 CAS Min %: 80.0 CAS Max %: 83.0 CAS: 50-21-5 CAS Min %: 18.2 CAS Max %: 18.94 Physical Form: Liquid
Lactic Acid, 85%, ACS Reagent Grade, Ricca Chemical
CAS: 7732-18-5 Purity Grade: ACS Reagent Grade Physical Form: Solution Assay: 85% Assay Percent Range: 85%
L-Lactic Acid, ≥95%, MP Biomedicals™
Chemical Name or Material: L-Lactic Acid Assay Percent Range: ≥95% Molecular Formula: C6H10O6Ca.5H2O Formula Weight: 308.3 Synonym: Calcium lactate pentahydrate
L-(+)-Lactic acid lithium salt, 99%, MP Biomedicals™
Chemical Name or Material: L-(+)-Lactic acid lithium salt CAS: 867-55-0 Physical Form: Powder Molecular Formula: C3H5O3Li Formula Weight: 96.1 Synonym: Lithium L-lactate
DL-Lactic Acid Lithium Salt, MP Biomedicals™
Chemical Name or Material: DL-Lactic Acid Lithium Salt CAS: 867-55-0 Physical Form: Powder Molecular Formula: C3H5O3Li Formula Weight: 96.01 MDL Number: MFCD00065399 Synonym: Hydroxypropionic acid lithium salt
L-(+)-Lactic acid sodium salt, 40% solution, MP Biomedicals™
Chemical Name or Material: L-(+)-Lactic acid sodium salt CAS: 867-56-1 Physical Form: Crystalline Powder To Liquid Molecular Formula: C3H5O3Na Formula Weight: 112.1 Synonym: (S)-2-Hydroxypropanoic acid monosodium salt
L-Lactic Acid (Free Acid), MP Biomedicals™
Chemical Name or Material: L-Lactic Acid Name Note: Free Acid CAS: 79-33-4 Physical Form: Crystalline Powder Molecular Formula: C3H6O3 Formula Weight: 90.08 MDL Number: MFCD00064266 Synonym: (S)-2-Hydroxypropanoic Acid
L-(+)-Lactic acid, >98%, MP Biomedicals™
Chemical Name or Material: L-(+)-Lactic acid CAS: 79-33-4 Physical Form: Crystalline Powder To Liquid Molecular Formula: C3H6O3 Formula Weight: 90.08 Synonym: (S)-2-Hydroxypropanoic acid
Lactic Acid, 85.0-90.0%, ACS, EMD Millipore
Chemical Name or Material: Lactic Acid CAS: 50-21-5 Name Note: For ACS analysis Purity Grade: ACS Grade Molecular Formula: CH3CHOHCOOH Linear Formula: CH3CHOHCOOH in water Formula Weight: 9.8
  spinner