Lead -Pb
Lead(II) sulfate 99%, ACROS Organics™ Available on GSA/VA Contract for Federal Government customers only.
Chemical Name or Material: Lead(II) sulfate Assay Percent Range: 99% Molecular Formula: O4PbS Formula Weight: 303.25 MDL Number: MFCD00011166
Lead(II) chloride, 99%, ACROS Organics™ Available on GSA/VA Contract for Federal Government customers only.
Chemical Name or Material: Lead(II) chloride Assay: ≥98.5% Assay Percent Range: 99% Molecular Formula: Cl2Pb Linear Formula: PbCl2 Formula Weight: 278.11 MDL Number: MFCD00011157
Lead(II) nitrate, 99.999%, (trace metal basis), ACROS Organics™ Available on GSA/VA Contract for Federal Government customers only.
Chemical Name or Material: Lead(II) nitrate CAS: 10099-74-8 Purity Grade: Trace Metal Basis Assay Percent Range: 99.999% Molecular Formula: N2O6Pb Linear Formula: Pb(NO3)2 Formula Weight: 331.2 MDL Number: MFCD00011153
Lead(II) acetate trihydrate, 99.999%, (trace metal basis), extra pure, ACROS Organics™ Available on GSA/VA Contract for Federal Government customers only.
Chemical Name or Material: Lead(II) acetate trihydrate Name Note: 99.999% CAS: 6080-56-4 Purity Grade: Trace Metal Basis Assay Percent Range: 99.999% Molecular Formula: C4H6O4Pb·3H2O Linear Formula: (CH3CO2)2Pb·3H2O Formula Weight: 379.33 MDL Number: MFCD00150023 Synonym: Acetic acid, lead salt trihydrate
Lead(II) iodide, ultra dry, Yellow powder, 99.999% (Metals basis), Alfa Aesar™
Chemical Name or Material: Lead(II) Iodide, Ultra Dry CAS: 10101-63-0 Purity Grade: Ultra dry Physical Form: Powder Assay Percent Range: 99.999% (Metals Basis) Molecular Formula: I2Pb Formula Weight: 461 MDL Number: MFCD00011163
Lead(II) sulfate, Reagent Grade, Alfa Aesar™
Chemical Name or Material: Lead(II) sulfate CAS: 7446-14-2 Purity Grade: Reagent Grade Physical Form: Powder/Lump Molecular Formula: PbSO4 Formula Weight: 303.25 MDL Number: MFCD00011166
Lead, +99%, granular, ACROS Organics™
Chemical Name or Material: Lead Name Note: p.a. CAS: 7439-92-1 Assay: ≥99.0% Assay Percent Range: 99+% Molecular Formula: Pb Linear Formula: Pb Formula Weight: 207.19 MDL Number: MFCD00134050
Lead Wire, 1.0mm (0.04 in.) dia., Puratronic™, 99.998% (Metals basis), Alfa Aesar™
Chemical Name or Material: Lead CAS: 7439-92-1 Physical Form: Wire, 1.0mm (0.04 in.) dia. Assay Percent Range: 99.998% (Metals basis) MDL Number: MFCD00134050
Lead(II) carbonate, White powder, Puratronic™, 99.999% (Metals basis), Alfa Aesar™
Chemical Name or Material: Lead(II) carbonate CAS: 598-63-0 Physical Form: Powder Assay Percent Range: 99.999% (Metals basis) Molecular Formula: PbCO3 Formula Weight: 267.2 MDL Number: MFCD00064631
Lead(II) sulfide, 99.9% (metals basis), Alfa Aesar™
Chemical Name or Material: Lead(II) sulfide CAS: 1314-87-0 Physical Form: -200 Mesh Powder Assay Percent Range: 99.9% (metals basis) Molecular Formula: PbS Formula Weight: 239.25 MDL Number: MFCD00016280
Lead(II) carbonate, ACS reagent, ACROS Organics™
Chemical Name or Material: Lead(II) carbonate CAS: 598-63-0 Purity Grade: ACS Reagent Grade Molecular Formula: CO3Pb Linear Formula: PbCO3 Formula Weight: 267.2
Lead(II) bromide, White powder, 99.999% (Metals basis), Alfa Aesar™
Chemical Name or Material: Lead(II) bromide CAS: 10031-22-8 Physical Form: Powder Assay Percent Range: 99.999% (Metals basis) Molecular Formula: Br2Pb Formula Weight: 367.01 MDL Number: MFCD00011156
Lead (Shot, Size No. 8), Fisher Chemical Available on GSA/VA Contract for Federal Government customers only.
Chemical Name or Material: Lead CAS: 7439-92-1 CAS: 7440-36-0 CAS: 7440-38-2 Physical Form: Solid Molecular Formula: Pb Formula Weight: 207.2 MDL Number: MFCD00134050
Lead Granules, +30 mesh, 99.99% (Metals basis), Alfa Aesar™
Chemical Name or Material: Lead CAS: 7439-92-1 Physical Form: Granular Assay Percent Range: 99.99% (Metals basis) MDL Number: MFCD00134050
Lead(II) tetrafluoroborate, Colorless liquid, Alfa Aesar™
Chemical Name or Material: Lead(II) tetrafluoroborate CAS: 13814-96-5 Physical Form: Liquid Molecular Formula: B2F8Pb Formula Weight: 380.81 MDL Number: MFCD00016270
Lead(IV) oxide, 97+%, extra pure, ACROS Organics™
Chemical Name or Material: Lead(IV) oxide Name Note: p.a. CAS: 1309-60-0 Purity Grade: Extra Pure Assay: ≥97.0% Assay Percent Range: 97+% Molecular Formula: O2Pb Linear Formula: PbO2 Formula Weight: 239.19 MDL Number: MFCD00011165 Synonym: Lead dioxide
Lead(II) telluride, Gray powder, 99.99% (Metals basis), Alfa Aesar™
Chemical Name or Material: Lead(II) telluride CAS: 1314-91-6 Physical Form: Powder Assay Percent Range: 99.99% (Metals basis) Molecular Formula: PbTe Formula Weight: 334.79 MDL Number: MFCD00016274
Lead Slug, Dia. X L: 6.35 x 6.35mm (0.25 x 0.25in.), Puratronic™, 99.999% (Metals basis), Alfa Aesar™
Chemical Name or Material: Lead CAS: 7439-92-1 Physical Form: Slug, dia. X L: 6.35 X 6.35mm (0.25 X 0.25 in.) Assay Percent Range: 99.999% (Metals basis) MDL Number: MFCD00134050
Lead(IV) oxide, Brown to black powder, Puratronic™, 99.995% (Metals basis), Alfa Aesar™
Chemical Name or Material: Lead(IV) oxide CAS: 1309-60-0 Physical Form: Powder Assay Percent Range: 99.995% (Metals basis) Molecular Formula: PbO2 Formula Weight: 239.19 MDL Number: MFCD00011165 Synonym: Lead dioxide
Lead(II) oxide, Yellow powder, 99.9%, Alfa Aesar™
Chemical Name or Material: Lead(II) oxide CAS: 1317-36-8 Physical Form: Powder Assay Percent Range: 99.9% Molecular Formula: PbO Formula Weight: 223.19 MDL Number: MFCD00011164
Lead(II) carbonate, Alfa Aesar™
Chemical Name or Material: Lead(II) carbonate Name Note: For analysis ACS CAS: 598-63-0 Purity Grade: ACS Grade Physical Form: Powder Molecular Formula: PbCO3 Formula Weight: 267.2 MDL Number: MFCD00064631
Lead powder, 99% (metals basis), Alfa Aesar™
Chemical Name or Material: Lead CAS: 7439-92-1 Physical Form: Powder Assay Percent Range: 99% (Metals basis) MDL Number: MFCD00134050
Lead Granules, 99.999% (Metals basis), Alfa Aesar™
Chemical Name or Material: Lead CAS: 7439-92-1 Physical Form: Granular Assay Percent Range: 99.999% (Metals basis) MDL Number: MFCD00134050
Lead(II) chloride, White Crystalline Powder, 99%, Alfa Aesar™
Chemical Name or Material: Lead(II) chloride CAS: 7758-95-4 Purity Grade: Reagent Grade Physical Form: Crystalline Powder Assay Percent Range: 99% Molecular Formula: PbCl2 Formula Weight: 278.1 MDL Number: MFCD00011157
Lead(II) perchlorate trihydrate, 99%, ACROS Organics™ Available on GSA/VA Contract for Federal Government customers only.
Chemical Name or Material: Lead(II) perchlorate trihydrate Name Note: 99% CAS: 13453-62-8 Assay: ≥98.5% Assay Percent Range: 99% Molecular Formula: Cl2O8Pb·3H2O Linear Formula: Pb(ClO4)2·3H2O Formula Weight: 460.14 MDL Number: MFCD00149817
Lead Wire, 0.5mm (0.02 in.) dia., 99.998% (metals basis), Alfa Aesar™, Puratronic™
Chemical Name or Material: Lead CAS: 7439-92-1 Physical Form: Wire, 0.5mm (0.02 in.) dia. Assay Percent Range: 99.998% (Metals basis) MDL Number: MFCD00134050
Lead Chloride Products for Science Education
Chemical Name or Material: Lead Chloride CAS: 7758-95-4 Purity Grade: Reagent Grade Assay Percent Range: >99% w/v Linear Formula: PbCl2 Formula Weight: 278.11 Synonym: Lead Dichloride
Lead(II) selenide, 3-6mm Fused gray lump, >99.999% (Metals basis), Alfa Aesar™
Chemical Name or Material: Lead(II) selenide CAS: 12069-00-0 Physical Form: 3 to 6mm Fused Gray Lump Assay Percent Range: >99.999% (Metals basis) Molecular Formula: PbSe Formula Weight: 286.15 MDL Number: MFCD00016273
Lead shot, 3mm (0.1 in.), 99.999% (metals basis), Alfa Aesar™
Chemical Name or Material: Lead CAS: 7439-92-1 Physical Form: Shot, 3mm (0.1 in.) Assay Percent Range: 99.999% (Metals basis) Molecular Formula: Pb MDL Number: MFCD00134050
Lead(II) acetate trihydrate, 99+%, ACS reagent, ACROS Organics™
Chemical Name or Material: Lead(II) acetate trihydrate CAS: 6080-56-4 Purity Grade: ACS Reagent Grade Assay: 99.0 to 103.0% Assay Percent Range: 99+% Molecular Formula: C4H6O4Pb·3H2O Linear Formula: (CH3CO2)2Pb·3H2O Formula Weight: 379.33 MDL Number: MFCD00150023 Synonym: Acetic acid, lead salt trihydrate
  spinner