Life Science Buffers
MOPS sodium salt, 98%, ACROS Organics™
Chemical Name or Material: MOPS sodium salt Name Note: 98% CAS: 71119-22-7 CAS Min %: 97.5 CAS Max %: 100.0 Assay: ≥97.5% Assay Percent Range: 98% Molecular Formula: C7H14NNaO4S Formula Weight: 231.25 MDL Number: MFCD00064350 Synonym: 3-(4-Morpholino)propanesulfonic acid sodium salt
TAPS, 99+%, for biochemistry, ACROS Organics™ Available on GSA/VA Contract for Federal Government customers only.
Chemical Name or Material: TAPS CAS: 29915-38-6 Purity Grade: For biochemistry Assay: ≥99% Assay Percent Range: 99+% Molecular Formula: C7H17NO6S Linear Formula: (HOCH2)3CNH(CH2)3SO3H Formula Weight: 243.27 MDL Number: MFCD00007538 Synonym: 3-[Tris(hydroxymethyl)methylamino]-1-propanesulfonic acid
Tris(hydroxymethyl)aminomethane Hydrochloride (Crystalline/Certified) Reagent Grade, Fisher Chemical
Chemical Name or Material: Tris(hydroxymethyl)aminomethane Hydrochloride CAS: 1185-53-1 Purity Grade: Reagent Grade Physical Form: Solid Assay: ≥95% Assay Percent Range: ≥95 % Molecular Formula: C4H11NO3 · HCl Linear Formula: NH2C(CH2OH)3·HCl Formula Weight: 157.6 MDL Number: MFCD00012590 Synonym: 2-Amino-2-(hydroxymethyl)-1, 3-propanediol Hydrochloride
CAPS, 99%, for biochemistry, ACROS Organics™
Chemical Name or Material: CAPS Name Note: 99% CAS: 1135-40-6 Purity Grade: For biochemistry Assay: ≥98.5% (on dry substance) Assay Percent Range: 99% Molecular Formula: C9H19NO3S Linear Formula: C6H11NH(CH2)3SO3H Formula Weight: 221.31 MDL Number: MFCD00003837 Synonym: 3-Cyclohexylamino-1-propanesulfonic acid
Tris(hydroxymethyl)aminomethane (Crystalline/Laboratory), Fisher Chemical
Chemical Name or Material: Tris(hydroxymethyl)aminomethane CAS: 77-86-1 Purity Grade: Laboratory Grade Physical Form: Powder/Solid Molecular Formula: C4H11NO3 Linear Formula: H2NC(CH2OH)3 Formula Weight: 121.14 MDL Number: MFCD00004679 Synonym: 2-Amino-2-(hydroxymethyl)-1, 3-Propanediol Trimethylol Aminomethane
TAPSO, 99+%, for biochemistry, ACROS Organics™
Chemical Name or Material: TAPSO CAS: 68399-81-5 Purity Grade: For biochemistry Assay: ≥99% (Acid) Assay Percent Range: 99+% Molecular Formula: C7H17NO7S Linear Formula: (HOCH2)3CNHCH2CH(OH)CH2SO3H Formula Weight: 259.3 MDL Number: MFCD00038352 Synonym: 3-(N-Tris-(hydroxymethyl)methylamino)-2-hydroxypropanesulfonic acid
Tris(hydroxymethyl) Aminomethane Products for Science Education
Chemical Name or Material: Tris(hydroxymethyl) Aminomethane CAS: 77-86-1 Purity Grade: Laboratory Grade Assay Percent Range: >99% Linear Formula: H2NC(CH2OH)3 Formula Weight: 121.14 Synonym: THAM
MOPS, 99%, for biochemistry, ACROS Organics™
Chemical Name or Material: MOPS Name Note: 99% CAS: 1132-61-2 CAS Min %: 98.5 CAS Max %: 100.0 Purity Grade: For biochemistry Assay: ≥98.5% Assay Percent Range: 99% Molecular Formula: C7H15NO4S Formula Weight: 209.26 MDL Number: MFCD00006183 Synonym: 3-(N-Morpholino)propanesulfonic acid
Tris EDTA buffer, for molecular biology, DNAse, RNAse, Protease free ready to use, pH 8.0, ACROS Organics™
Chemical Name or Material: Tris EDTA buffer Name Note: pH 8.0 CAS: 38641-82-6 Purity Grade: For molecular biology MDL Number: MFCD00236359 Synonym: 2-Amino-2-(hydroxymethyl)-1, 3-propanediol
Tris(hydroxymethyl)aminomethane, 99.8%, ACS reagent, ACROS Organics™
Chemical Name or Material: Tris(hydroxymethyl)aminomethane CAS: 77-86-1 Purity Grade: ACS Reagent Grade Assay: 99.8 to 100.1% (on dry basis) Assay Percent Range: 99.8% Molecular Formula: C4H11NO3 Linear Formula: (HOCH2)3CNH2 Formula Weight: 121.14 MDL Number: MFCD00004679 Synonym: 2-Amino-2-(hydroxymethyl)-1, 3-propanediol
Tris Buffer, 2M Solution, ACROS Organics™
Chemical Name or Material: Tris buffer Name Note: 2M solution CAS: 77-86-1 Molecular Formula: C4H11NO3 Formula Weight: 121.14 MDL Number: MFCD00004679 Synonym: Tris(hydroxymethyl)aminomethane
Phosphate Buffer, 0.05M, Fisher Chemical
Chemical Name or Material: Phosphate Buffer, 0.05M CAS: 7732-18-5 CAS Min %: 91.0 CAS Max %: 95.0 CAS: 64-19-7 CAS Min %: 5.76 CAS Max %: 5.99 CAS: 7778-77-0 CAS Min %: 0.31 CAS Max %: 0.32 CAS: 7664-38-2 CAS Min %: 0.26 CAS Max %: 0.27 CAS: 6153-56-6 CAS Min %: 0.18 CAS Max %: 0.19 CAS: 79-09-4 CAS Min %: 0.09 CAS Max %: 0.09
Tris (Tromethamine) (Granular/USP), Fisher Chemical Available on GSA/VA Contract for Federal Government customers only.
Chemical Name or Material: Tris (Tromethamine) CAS: 77-86-1 Purity Grade: USP Physical Form: Powder/Solid Assay: 99.0 to 101.0% (USP) Molecular Formula: C4H11NO3 Linear Formula: H2NC(CH2OH)3 Formula Weight: 121.14 MDL Number: MFCD00004679 Synonym: THAM™
Tris(hydroxymethyl)aminomethane (Cryst./Certified ACS), Fisher Chemical
Chemical Name or Material: Tris(hydroxymethyl)aminomethane CAS: 77-86-1 Purity Grade: Certified ACS Grade Physical Form: Powder/Solid Assay: 99.8 to 100.1% Assay Percent Range: 99.8 to 100.1% Molecular Formula: C4H11NO3 Linear Formula: H2NC(CH2OH)3 Formula Weight: 121.14 MDL Number: MFCD00004679 Synonym: 2-Amino-2-(hydroxymethyl)-1, 3-propanediol
CHES, 98+%, for biochemistry, ACROS Organics™
Chemical Name or Material: CHES Name Note: 98+% CAS: 103-47-9 Purity Grade: For biochemistry Assay: ≥98.0% Assay Percent Range: 98+% Molecular Formula: C8H17NO3S Linear Formula: C6H11NHCH2CH2SO3H Formula Weight: 207.29 MDL Number: MFCD00003835 Synonym: 2-(Cyclohexylamino)ethanesulfonic acid
EPPS, 99%, for biochemistry, ACROS Organics™
Chemical Name or Material: EPPS CAS: 16052-06-5 Purity Grade: For biochemistry Assay: ≥98.5% Assay Percent Range: 99% Molecular Formula: C9H20N2O4S Formula Weight: 252.33 MDL Number: MFCD00006160 Synonym: 3-[4-(2-Hydroxyethyl)-1-piperazine]propanesulfonic acid
Tris Borate EDTA Products for Science Education
Chemical Name or Material: Tris Borate EDTA CAS: 10043-35-3 CAS: 6381-92-6 CAS: 77-86-1 CAS: 7732-18-5 Purity Grade: Laboratory Grade Assay Percent Range: 2.75% w/v Synonym: TBE 5X Buffer
Tris Borate EDTA buffer, for molecular biology, 10x, DNAse, RNAse and Protease free,, ACROS Organics™
Chemical Name or Material: Tris Borate EDTA buffer Name Note: pH 8.3 CAS: 610769-35-2 Purity Grade: For molecular biology MDL Number: MFCD00236358 Synonym: 2-Amino-2-(hydroxymethyl)-1, 3-propanediol
MOPS, 99%
Chemical Name or Material: MOPS CAS: 1132-61-2 Molecular Formula: C7H15NO4S Formula Weight: 209.27 MDL Number: MFCD00006183 Beilstein: 1106776 Merck Index: 14,6265
MOPS, 99.5%, for molecular biology, Dnase, Rnase, Prot. free, for peptide sequencing, ACROS Organics™
Chemical Name or Material: MOPS Name Note: DNAse, RNAse and Protease Free CAS: 1132-61-2 CAS Min %: 99.4 CAS Max %: 100.0 Purity Grade: For molecular biology Assay: ≥99.5% Assay Percent Range: 99.50% Molecular Formula: C7H15NO4S Formula Weight: 209.26 MDL Number: MFCD00006183 Synonym: 3-(N-Morpholino)propanesulfonic acid
Wright's Stain Buffer Products for Science Education
Chemical Name or Material: Wright's Blood Stain Buffer CAS: 7778-77-0 CAS: 7558-79-4 CAS: 7732-18-5 Assay Percent Range: 0.7% w/v Synonym: Giordan pH 6.4
Buffer Solution B, for low level Sulfate Analysis (Turbidimetric Method), Ricca Chemical
Chemical Name or Material: Buffer Solution B CAS: 7732-18-5 CAS Min %: 93.0 CAS Max %: 96.0 CAS: 7791-18-6 CAS Min %: 2.78 CAS Max %: 2.9 CAS: 64-19-7 CAS Min %: 1.95 CAS Max %: 2.03 CAS: 6131-90-4 CAS Min %: 0.46 CAS Max %: 0.48 CAS: 7757-79-1 CAS Min %: 0.09 CAS Max %: 0.1 CAS: 7757-82-6 CAS Min %: 0.01 CAS Max %: 0.01 Physical Form: Liquid
STET Buffer, 1X Solution (pH 8.0), Fisher BioReagents
Chemical Name or Material: STET Buffer, 1X Solution Name Note: pH 8.0 CAS: 77-86-1 (Tris Base) CAS: 60-00-4 (EDTA) CAS: 7647-01-0 (HCl) CAS: 7647-14-5 (NaCl) CAS: 9002-93-1 (Triton™ X-100)
TAE Buffer, Tris-Acetate-EDTA, 1X Solution, Electrophoresis, Fisher BioReagents
Chemical Name or Material: Tris-Acetate-EDTA Name Note: 1X Solution CAS: 77-86-1 CAS: 6850-28-8 CAS: 139-33-3 CAS: 10378-23-1 CAS: 7732-18-5 Purity Grade: Electrophoresis Grade
TE Buffer, Tris-EDTA, 10X Solution, pH 7.4, Molecular Biology, Fisher BioReagents
Chemical Name or Material: Tris-EDTA (TE), 10X Solution Name Note: pH 7.4 CAS: 77-86-1 CAS: 6381-92-6 CAS: 7647-01-0 CAS: 7732-18-5 Purity Grade: Molecular Biology Grade Synonym: TE
TE Buffer, Tris-EDTA, 1X Solution, pH 7.4, Molecular Biology, Fisher BioReagents
Chemical Name or Material: Tris-EDTA Name Note: 1X Solution, pH 7.4 CAS: 77-86-1 CAS: 60-00-4 CAS: 7647-01-0 CAS: 7732-18-5 Purity Grade: Molecular Biology Grade Synonym: TE
Buffer, Tris(hydroxymethyl)aminomethane, 1 Molar, pH 7.50, Ricca Chemical
Chemical Name or Material: Buffer, Tris(hydroxymethyl)aminomethane Name Note: pH 7.50 CAS: 7732-18-5 CAS Min %: 82.0 CAS Max %: 86.0 CAS: 77-86-1 CAS Min %: 9.97 CAS Max %: 10.37 CAS: 7647-01-0 CAS Min %: 5.82 CAS Max %: 6.06 Physical Form: Liquid
STE Buffer, Sodium Chloride-Tris-EDTA, 10X Solution, pH 8.0, Fisher BioReagents
Chemical Name or Material: Sodium Chloride-Tris-EDTA (STE) Buffer Name Note: 10X, pH 8.0 CAS: 7647-14-5 CAS: 60-00-4 CAS: 7647-01-0 CAS: 7732-18-5 Synonym: STE
TBE Buffer, Tris-Borate-EDTA, 1X Solution, Electrophoresis, Fisher BioReagents
Chemical Name or Material: Tris-Borate-EDTA Name Note: 1X Solution CAS: 77-86-1 CAS: 6381-92-6 CAS: 10043-35-3 CAS: 7732-18-5 Purity Grade: Electrophoresis Grade
MOPS 10X Solution, Fisher BioReagents™
Chemical Name or Material: MOPS Name Note: 10X Solution, pH 7 CAS: 7732-18-5 CAS: 1132-61-2 CAS: 1310-73-2 Synonym: 3-(N-morpholino)propanesulfonic acid
  spinner