Life Science Water -Cell Culture Grade
Water, Microbial Cell Culture Grade, Fisher BioReagents Available on GSA/VA Contract for Federal Government customers only.
Chemical Name or Material: Water Name Note: 0.03μm filtered and autoclaved CAS: 7732-18-5 Purity Grade: Microbial Cell Culture Molecular Formula: H2O Formula Weight: 18.01
EMD Millipore™ EmbryoMax™ Ultra Pure Sterile Water Available on GSA/VA Contract for Federal Government customers only.
Used for routine mouse embryonic stem cell culture applications.
  spinner