Liquid Scintillation Counting Cocktails and Reagents
ScintiVerse™ E Cocktail (Scintanalyzed™), Fisher Chemical
Chemical Name or Material: ScintiVerse™ E Cocktail CAS: 1330-20-7 CAS: 9016-45-9 CAS: 92-71-7 CAS: 1806-34-4 Purity Grade: Scintanalyzed™ MDL Number: MFCD00212669
ScintiVerse™ LC Cocktail (Scintanalyzed™), Fisher Chemical
Chemical Name or Material: ScintiVerse™ LC Cocktail CAS: 95-63-6 CAS: 68131-39-5 CAS: 67-63-0 CAS: 92-71-7 CAS: 1806-34-4 Purity Grade: Scintanalyzed™ MDL Number: MFCD00212672
2-Butoxyethanol, Scintillation Grade, ACROS Organics™
Chemical Name or Material: 2-Butoxyethanol CAS: 111-76-2 Assay: ≥99% Molecular Formula: C6H14O2 Linear Formula: CH3(CH2)3OCH2CH2OH Formula Weight: 118.18 Synonym: Butyl Cellosolve™
Naphthalene, 99+%, scintillation grade, ACROS Organics™
Chemical Name or Material: Naphthalene Name Note: 99+% CAS: 91-20-3 CAS Min %: 99.0 CAS Max %: 100.0 Assay: ≥99.0% (GC) Assay Percent Range: 99+% Molecular Formula: C10H8 Linear Formula: C10H8 Formula Weight: 128.17 MDL Number: MFCD00001742
ScintiVerse™ BD Cocktail (Scintanalyzed™), Fisher Chemical Available on GSA/VA Contract for Federal Government customers only.
Chemical Name or Material: ScintiVerse™ BD Cocktail CAS: 67774-74-7 CAS: 577-11-7 CAS: 9016-45-9 CAS: 92-71-7 CAS: 13280-61-0 Purity Grade: Scintanalyzed™ MDL Number: MFCD00212667
ScintiGest™ Tissue Solubilizer, 0.6N (Scintanalyzed™), Fisher Chemical
Chemical Name or Material: ScintiGest™ Tissue Solubilizer Name Note: 0.6N CAS: 57-09-0 CAS: 67-56-1 CAS: 1310-58-3 Purity Grade: Scintanalyzed™ MDL Number: MFCD00212665
2,5-Diphenyloxazole, Scintillation Grade 99%, ACROS Organics™ Available on GSA/VA Contract for Federal Government customers only.
Chemical Name or Material: 2, 5-Diphenyloxazole Name Note: 99% CAS: 92-71-7 CAS Min %: 98.5 CAS Max %: 100.0 Assay: ≥98.5% Assay Percent Range: 99% Molecular Formula: C15H11NO Formula Weight: 221.26 MDL Number: MFCD00005306 Synonym: PPO
p-Xylene, 99%, for spectroscopy, scintillation grade, ACROS Organics™ Available on GSA/VA Contract for Federal Government customers only.
Chemical Name or Material: p-Xylene Name Note: scintillation grade, 99% CAS: 106-42-3 Purity Grade: For spectroscopy Assay: ≥98.5% Assay Percent Range: 99% Molecular Formula: C8H10 Linear Formula: C6H4(CH3)2 Formula Weight: 106.17 MDL Number: MFCD00008556 Synonym: 1, 4-Dimethylbenzene
Mineral oil
Chemical Name or Material: Mineral oil CAS: 8020-83-5 MDL Number: MFCD00131611
Toluene (Scintanalyzed™), Fisher Chemical Available on GSA/VA Contract for Federal Government customers only.
Chemical Name or Material: Toluene CAS: 108-88-3 Purity Grade: Scintanalyzed™ Purity Grade Notes: Also meets ACS specifications Assay: ≥99.5% Assay Percent Range: ≥99.5 % Molecular Formula: C7H8 Linear Formula: C6H5CH3 Formula Weight: 92.14 MDL Number: MFCD00008512 Synonym: Methylbenzene
CAB-O-SIL™ M-5, Scintillation Grade, ACROS Organics™
Chemical Name or Material: CAB-O-SIL™ M-5 CAS: 7631-86-9 Molecular Formula: O2Si Linear Formula: SiO2 Formula Weight: 60.08 MDL Number: MFCD00011232 Synonym: Silicon dioxide
Xylenes (Scintanalyzed™), Fisher Chemical
Chemical Name or Material: Xylenes CAS: 1330-20-7 CAS: 100-41-4 Purity Grade: Scintanalyzed™ Purity Grade Notes: Also meets ACS specifications Assay: ≥98.5% Assay Percent Range: ≥98.5 % Molecular Formula: C8H10 Linear Formula: C6H4(CH3)2 Formula Weight: 106.17 MDL Number: MFCD00077264 Synonym: Dimethylbenzene
ScintiVerse™ II Cocktail (Scintanalyzed™), Fisher Chemical Available on GSA/VA Contract for Federal Government customers only.
Chemical Name or Material: ScintiVerse™ II Cocktail CAS: 95-63-6, 13280-61-0, 92-71-7, 9003-11-6 CAS: 13280-61-0 CAS: 92-71-7 CAS: 9003-11-6 Purity Grade: Scintanalyzed™ MDL Number: MFCD00212671
ScintiSafe™ Gel Cocktail (Scintanalyzed™), Fisher Chemical
Chemical Name or Material: ScintiSafe™ Gel Cocktail Purity Grade: Scintanalyzed™ MDL Number: MFCD00242091 Synonym: PXE-based (Phenylxylylethane)
CytoScint ES, MP Biomedicals
Chemical Name or Material: Liquid Scintillation Cocktails
EcoLume Scintillation Cocktail, MP Biomedicals
Chemical Name or Material: EcoLume™ Liquid Scintillation Cocktail
Ecolite(+) LC Fluid, MP Biomedicals
Chemical Name or Material: Liquid Scintillation Cocktails
Universol- ES liquid scintillation cocktail, MP Biomedicals™
Chemical Name or Material: Universol- ES liquid scintillation cocktail
beta Max-ES liquid scintillation cocktail, MP Biomedicals™
Chemical Name or Material: Beta Max-ES liquid scintillation cocktail
EcoLume LC Fluid, MP Biomedicals
Chemical Name or Material: Liquid Scintillation Cocktails
EcoLite(+) Scintillation Cocktail, MP Biomedicals
Chemical Name or Material: EcoLite(+)™ Liquid Scintillation Cocktail
Cytoscint- ES liquid scintillation cocktail, MP Biomedicals™
Chemical Name or Material: Cytoscint- ES liquid scintillation cocktail
ScintiVerse™ Bio-HP Cocktail (Scintanalyzed™), Fisher Chemical
Chemical Name or Material: ScintiVerse™ Bio-HP Cocktail CAS: 95-63-6 CAS: 9036-19-5 CAS: 68131-39-5 CAS: 68412-54-4 CAS: 112-80-1 Purity Grade: Scintanalyzed™ MDL Number: MFCD00212668
Cytoscint, Scintillation Cocktail, ACROS Organics™
Chemical Name or Material: Cytoscint CAS: 95-63-6 MDL Number: MFCD00211846
POPOP (Powder/Scintanalyzed™), Fisher Chemical
Chemical Name or Material: POPOP CAS: 1806-34-4 Purity Grade: Scintanalyzed™ Physical Form: Solid Molecular Formula: C24H16N2O2 Formula Weight: 364.39 MDL Number: MFCD00005309 Synonym: 1, 4-Bis[2-(5-Phenyloxazolyl)]benzene
Bis(MSB), Scintillation Grade 99.5%, ACROS Organics™
Chemical Name or Material: Bis(MSB) Name Note: 99.5% CAS: 13280-61-0 CAS Min %: 99.4 CAS Max %: 100.0 Assay: ≥99.4% Assay Percent Range: 99.50% Molecular Formula: C24H22 Formula Weight: 310.44 MDL Number: MFCD00008529 Synonym: 1, 4-Bis(2-methylstyryl)benzene
ScintiLene™ Cocktail (Scintanalyzed), Fisher Chemical
Chemical Name or Material: ScintiLene™ Cocktail CAS: 1330-20-7 CAS: 92-71-7 CAS: 13280-61-0 Purity Grade: Scintanalyzed™ MDL Number: MFCD00212666
Scintillation Surfactant (Scintanalyzed™), Fisher Chemical
Chemical Name or Material: Scintillation Surfactant CAS: 9036-19-5 Purity Grade: Scintanalyzed™ Molecular Formula: (C2H4O)10C14H22O Linear Formula: RO(CH2CH2O)10H MDL Number: MFCD00128254
ScintiSafe™ Econo 1 Cocktail (Scintanalyzed™), Fisher Chemical Available on GSA/VA Contract for Federal Government customers only.
Chemical Name or Material: ScintiSafe™ Econo 1 Cocktail CAS: 92-71-7 Purity Grade: Scintanalyzed™ MDL Number: MFCD00242091 Synonym: LAB-based (Linear Alkylbenzene)
ScintiVerse™ I Cocktail (Scintanalyzed™), Fisher Chemical
Chemical Name or Material: ScintiVerse™ I Cocktail CAS: 1330-20-7 CAS: 13280-61-0 CAS: 92-71-7 CAS: 112-80-1 CAS: 9036-19-5 Purity Grade: Scintanalyzed™ MDL Number: MFCD00212670
  spinner