Lithium -Li
Lithium bromide hydrate, White crystalline aggregates, Puratronic™, 99.995% (Metals basis), Alfa Aesar™
Chemical Name or Material: Lithium bromide hydrate CAS: 85017-82-9 Physical Form: Crystalline Aggregates Assay Percent Range: 99.995% (Metals basis) Molecular Formula: LiBr·xH2O Formula Weight: 86.85 (Anhydrous) MDL Number: MFCD00149763
Lithium carbonate, 99.99% (metals basis excluding Ca), Alfa Aesar™, Puratronic™
Chemical Name or Material: Lithium carbonate CAS: 554-13-2 Physical Form: Powder Assay Percent Range: 99.998% (Metals basis excluding Ca) Molecular Formula: Li2CO3 Formula Weight: 73.9 MDL Number: MFCD00011084
Lithium iodide hydrate, 99%, extra pure, ACROS Organics™
Chemical Name or Material: Lithium iodide hydrate CAS: 85017-80-7 Purity Grade: Extra Pure Assay: ≥98.5% (on dry substance) Assay Percent Range: 99% Molecular Formula: ILi·xH2O Linear Formula: LiI·xH2O Formula Weight: 133.85 MDL Number: MFCD00149769
Lithium chloride hydrate, White granules, 99.996% (Metals basis), Alfa Aesar™
Chemical Name or Material: Lithium chloride hydrate CAS: 85144-11-2 Physical Form: Granules Assay Percent Range: 99.996% (Metals basis) Molecular Formula: LiCl·xH2O Formula Weight: 42.39 (Anhydrous) MDL Number: MFCD00149764
Lithium aluminum hydride, 95% min, Alfa Aesar™
Chemical Name or Material: Lithium aluminum hydride CAS: 16853-85-3 Physical Form: Powder Assay Percent Range: 95% min. Molecular Formula: LiAlH4 Formula Weight: 37.95 MDL Number: MFCD00011075 Synonym: LAH
Lithium phosphate, Puratronic™, 99.99% (metals basis)
Chemical Name or Material: Lithium phosphate CAS: 10377-52-3 Purity Grade: Puratronic™ Molecular Formula: Li3PO4 Formula Weight: 115.79 MDL Number: MFCD00016187
Lithium tetraborate, White powder, Puratronic™, 99.998% (Metals basis), Alfa Aesar™
Chemical Name or Material: Lithium tetraborate CAS: 12007-60-2 Physical Form: Powder Assay Percent Range: 99.998% (Metals basis) Molecular Formula: B4Li2O7 Formula Weight: 169.12 MDL Number: MFCD00011083
Lithium fluoride, 99.99% (metals basis), Alfa Aesar™, Puratronic™
Chemical Name or Material: Lithium fluoride CAS: 7789-24-4 Physical Form: Powder Assay Percent Range: 99.99% (Metal basis) Molecular Formula: FLi Formula Weight: 25.94 MDL Number: MFCD00011090
Lithium tris[(3-ethyl-3-pentyl)oxy]aluminohydride, 0.5M solution in THF, AcroSeal™, ACROS Organics™
Chemical Name or Material: Lithium tris[(3-ethyl-3-pentyl)oxy]aluminohydride CAS: 79172-99-9 CAS: 109-99-9 Molecular Formula: C21H46AlLiO3 Formula Weight: 380.51
Lithium deuteride, 98%(Isotopic), Alfa Aesar™
Chemical Name or Material: Lithium deuteride CAS: 13587-16-1 Physical Form: Powder Assay Percent Range: 98% (Isotopic) Molecular Formula: DLi Formula Weight: 8.95 MDL Number: MFCD00011091
Lithium carbonate, 99%, pure, ACROS Organics™ Available on GSA/VA Contract for Federal Government customers only.
Chemical Name or Material: Lithium carbonate Name Note: 99% CAS: 554-13-2 Purity Grade: Pure Assay: ≥98.5% Assay Percent Range: 99% Molecular Formula: CLi2O3 Linear Formula: Li2CO3 Formula Weight: 73.88 MDL Number: MFCD00011084
Lithium bromide, 4M solution in THF, ACROS Organics™ Available on GSA/VA Contract for Federal Government customers only.
Chemical Name or Material: Lithium bromide CAS: 7550-35-8 CAS: 109-99-9 Molecular Formula: BrLi Linear Formula: LiBr Formula Weight: 86.84 MDL Number: MFCD00011077
Lithium nitrate, 99.999%, (trace metal basis), ACROS Organics™ Available on GSA/VA Contract for Federal Government customers only.
Chemical Name or Material: Lithium nitrate CAS: 7790-69-4 Purity Grade: For trace metal basis analysis Assay: 9 to 11% (Li) Assay Percent Range: 100.00% Molecular Formula: LiNO3 Linear Formula: LiNO3 Formula Weight: 68.95
Lithium hydroxide, anhydrous, 98%, Alfa Aesar™
Chemical Name or Material: Lithium hydroxide, anhydrous CAS: 1310-65-2 Physical Form: 4+14 Mesh Assay Percent Range: 98% Molecular Formula: LiOH Formula Weight: 23.95 MDL Number: MFCD00011095
Lithium fluoride, 97%, for analysis, ACROS Organics™
Chemical Name or Material: Lithium fluoride CAS: 7789-24-4 Purity Grade: For analysis Assay: ≥96.0% Assay Percent Range: 97% Molecular Formula: FLi Linear Formula: LiF Formula Weight: 25.93
Lithium aluminium hydride, 95%, in resealable tin cans, ACROS Organics™ Available on GSA/VA Contract for Federal Government customers only.
Chemical Name or Material: Lithium aluminium hydride Name Note: In Resealable Tin Cans CAS: 16853-85-3 Assay: ≥94% (LiAlH4) Assay Percent Range: 95% Molecular Formula: AlH4Li Linear Formula: LiAlH4 Formula Weight: 37.95 MDL Number: MFCD00011075 Synonym: LAH
Lithium hexafluorophosphate, 98%, pure, ACROS Organics™ Available on GSA/VA Contract for Federal Government customers only.
Chemical Name or Material: Lithium hexafluorophosphate CAS: 21324-40-3 Purity Grade: Extra Pure Assay: ≥93.5% (PF6) Assay Percent Range: 98% Molecular Formula: F6LiP Linear Formula: LiPF6 Formula Weight: 151.91 MDL Number: MFCD00011096
Lithium sulfate, anhydrous, 99.7% (metals basis), Alfa Aesar™
Chemical Name or Material: Lithium sulfate, anhydrous CAS: 10377-48-7 Physical Form: Powder Assay Percent Range: 99.7% (Metal basis) Molecular Formula: Li2SO4 Formula Weight: 109.94 MDL Number: MFCD00011086
Lithium bromide, 99+%, for analysis, anhydrous, ACROS Organics™
Chemical Name or Material: Lithium bromide Name Note: 99+% CAS: 7550-35-8 Purity Grade: For analysis Assay: ≥99.0% (Argentometry) Assay Percent Range: 99+% Molecular Formula: BrLi Linear Formula: LiBr Formula Weight: 86.84 MDL Number: MFCD00011077
Lithium Hydroxide, Anhydrous, ACROS Organics™ Available on GSA/VA Contract for Federal Government customers only.
Chemical Name or Material: Lithium hydroxide CAS: 1310-65-2 Molecular Formula: Li O H Linear Formula: LiOH Formula Weight: 23.95 MDL Number: MFCD00011095
Lithium hydroxide monohydrate, 56.5% min, Alfa Aesar™
Chemical Name or Material: Lithium hydroxide monohydrate CAS: 1310-66-3 Physical Form: Powder Assay Percent Range: 56.5% min Molecular Formula: LiOH·H2O Formula Weight: 41.96; 23.95 (anhydrous) MDL Number: MFCD00149772
Lithium Chloride Solution, 1M Products for Science Education
Chemical Name or Material: Lithium Chloride Solution, 1M CAS: 7447-41-8 Linear Formula: LiCl Formula Weight: 42.39 Synonym: Lithium Monochloride
Lithium nitrate, anhydrous, 99%, Alfa Aesar™
Chemical Name or Material: Lithium nitrate, anhydrous CAS: 7790-69-4 Physical Form: Crystals Assay Percent Range: 99% Molecular Formula: LiNO3 Formula Weight: 68.95 MDL Number: MFCD00011094
Lithium nitrate, 99+%, extra pure, ACROS Organics™
Chemical Name or Material: Lithium nitrate CAS: 7790-69-4 Purity Grade: Extra Pure Assay: ≥99.0% (LINO3) Assay Percent Range: 99+% Molecular Formula: LiNO3 Linear Formula: LiNO3 Formula Weight: 68.95 MDL Number: MFCD00011094
Lithium aluminium hydride bis(tetrahydrofuran), 1M solution in toluene, AcroSeal™, ACROS Organics™ Available on GSA/VA Contract for Federal Government customers only.
Chemical Name or Material: Lithium aluminium hydride bis(tetrahydrofuran) CAS: 123439-81-6 CAS: 108-88-3 Molecular Formula: C8H20AlLiO2 Formula Weight: 182.17
Lithium chloride, ≥99%, for analysis, anhydrous, ACROS Organics™
Chemical Name or Material: Lithium chloride Name Note: 99% CAS: 7447-41-8 Purity Grade: For analysis Assay: ≥98.5% Assay Percent Range: 99% Molecular Formula: ClLi Linear Formula: LiCl Formula Weight: 42.38 MDL Number: MFCD00011078
Lithium bromide, 99%, Acros Organics
Chemical Name or Material: Lithium bromide CAS: 7550-35-8 Purity Grade: Pure Assay Percent Range: 99% Molecular Formula: BrLi Formula Weight: 86.84
Lithium sulfate, 99%, extra pure, anhydrous, ACROS Organics™ Available on GSA/VA Contract for Federal Government customers only.
Chemical Name or Material: Lithium sulfate Name Note: 99% CAS: 10377-48-7 Purity Grade: Extra Pure Assay: ≥98.5% (LI2SO4) Assay Percent Range: 99% Molecular Formula: Li2O4S Linear Formula: Li2SO4 Formula Weight: 109.94 MDL Number: MFCD00011086
Lithium hydroxide monohydrate, ACS, 98% min, Alfa Aesar™
Chemical Name or Material: Lithium hydroxide monohydrate CAS: 1310-66-3 Purity Grade: ACS Grade Physical Form: Powder Assay Percent Range: 98% min Molecular Formula: LiOH·H2O Formula Weight: 41.96 MDL Number: MFCD00149772
Lithium carbonate, 99+%, ACS reagent, ACROS Organics™
Chemical Name or Material: Lithium carbonate CAS: 554-13-2 Purity Grade: ACS Reagent Grade Assay: ≥99.0% (Li2CO3) Assay Percent Range: 99+% Molecular Formula: CLi2O3 Linear Formula: Li2CO3 Formula Weight: 73.88 MDL Number: MFCD00011084
  spinner