Lysine -Lys and Derivatives
L-Lysine, 98%, ACROS Organics™
Chemical Name or Material: L-Lysine Assay: 98% Assay Percent Range: 98% Molecular Formula: C6H14N2O2 Formula Weight: 146.19 Synonym: (S)-2, 6-Diaminocaproic acid
L-Lysine acetate, 97%, Acros Organics
Chemical Name or Material: L-Lysine acetate Assay Percent Range: 97% Molecular Formula: C6H14N2O2·C2H4O2 Formula Weight: 206.24
DL-Lysine monohydrate, 97%, ACROS Organics™
Chemical Name or Material: DL-Lysine monohydrate Name Note: 98% Assay: ≥96.0% Assay Percent Range: 97% Molecular Formula: C6H14N2O2·H2O Linear Formula: HN(CH)4CH(NH)COH Formula Weight: 164.21 MDL Number: MFCD00151036
L-Lysine Dihydrochloride 99%, ACROS Organics™ Available on GSA/VA Contract for Federal Government customers only.
Chemical Name or Material: L-Lysine dihydrochloride Name Note: 99% CAS: 657-26-1 Assay: 98.5 to 101.5% (on dried substance) Assay Percent Range: 99% Molecular Formula: C6H14N2O2·2HCl Linear Formula: NH2(CH2)4CH(NH2)COOH·2HCl Formula Weight: 219.11 MDL Number: MFCD00066389
D-Lysine hydrochloride, 99+%, ACROS Organics™
Chemical Name or Material: D-Lysine hydrochloride Assay: 99.0 to 101.0% Assay Percent Range: 99+% Molecular Formula: C6H14N2O2·HCl Linear Formula: H2N(CH2)4CH(NH2)COOH·HCl Formula Weight: 182.65 MDL Number: MFCD00012920 Synonym: D-2, 6-Diaminocaproic acid, hydrochloride
DL-Lysine Monohydrochloride 99%, ACROS Organics™
Chemical Name or Material: DL-Lysine monohydrochloride Name Note: 99% CAS: 70-53-1 Assay: ≥98.5% (Total base) Assay Percent Range: 99% Molecular Formula: C6H14N2O2·HCl Linear Formula: NH2(CH2)4CH(NH2)COO·HCl Formula Weight: 182.65 MDL Number: MFCD00064563
L(+)-Lysine monohydrate, 99%, ACROS Organics™
Chemical Name or Material: L(+)-Lysine monohydrate Assay: 98.5 to 101.5% Assay Percent Range: 99% Molecular Formula: C6H14N2O2·H2O Linear Formula: H2N(CH2)4CH(NH2)CO2H·H2O Formula Weight: 164.21 MDL Number: MFCD00151035
L(+)-Lysine Monohydrochloride, ≥99%, ACROS Organics™
Chemical Name or Material: L(+)-Lysine monohydrochloride Assay: 99.0 to 101.0% (on dried substance) Assay Percent Range: 99+% Molecular Formula: C6H14N2O2·HCl Linear Formula: H2N(CH2)4CH(NH2)CO2H·HCl Formula Weight: 182.65 MDL Number: MFCD00064564 Synonym: L(+)-2, 6-Diaminocaproic acid, hydrochloride
N-ε-Carbobenzyloxy-L-lysine, 98%, ACROS Organics™
Chemical Name or Material: Nε-Carbobenzyloxy-L-lysine CAS: 1155-64-2 CAS Min %: 97.5 CAS Max %: 100.0 Assay: ≥97.5% Assay Percent Range: 98% Molecular Formula: C14H20N2O4 Linear Formula: C6H5CH2O2CNH(CH2)4CH(NH2)CO2H Formula Weight: 280.32 MDL Number: MFCD00002638 Synonym: Nε-CBZ-L-Lysine
L-NIL hydrochloride, Tocris
Chemical Name or Material: L-NIL hydrochloride Assay Percent Range: >98% Molecular Formula: C8H17N3O2·HCl Formula Weight: 223.7 Synonym: N6-(1-Iminoethyl)-L-lysine hydrochloride
L-Lysine Hydrochloride (White Powder), Fisher BioReagents Available on GSA/VA Contract for Federal Government customers only.
Chemical Name or Material: L-Lysine Hydrochloride CAS: 657-27-2 Physical Form: Solid Assay: 98.5 to 100.5% Assay Percent Range: 98.5 to 100.5 % Molecular Formula: C6H14N2O2 · HCl Formula Weight: 182.65
Biocytin, Tocris
Chemical Name or Material: Biocytin Assay Percent Range: >98% Molecular Formula: C16H28N4O4S Formula Weight: 372.48 Synonym: N6-[5-[(3aS,4S,6aR)-Hexahydro-2-oxo-1H-thieno[3,4-d]imidazol-4-yl]-1-oxopentyl]-L-lysine
L-Lysine Hydrochloride, 99+%, MP Biomedicals
Chemical Name or Material: L-Lysine, 99+% Name Note: Hydrochloride CAS: 657-27-2 Purity Grade: Cell Culture Reagent Grade Physical Form: Crystalline Molecular Formula: C6H14N2O2•HCl Formula Weight: 182.6
L-Lysine [Free Base], MP Biomedicals
Chemical Name or Material: L-Lysine [Free Base] CAS: 39665-12-8 Purity Grade: Cell Culture Reagent Grade Physical Form: Crystalline Molecular Formula: C6H14N2O2 Formula Weight: 146.2 Synonym: L-2, 6-Diaminohexanoic acid
L-lysine methyl ester dihydrochloride, MP Biomedicals™
Chemical Name or Material: L-Lysine methyl ester dihydrochloride CAS: 26348-70-9 Physical Form: Crystalline Powder Molecular Formula: C7H16N2O2·2HCI Formula Weight: 233.13 Synonym: H-Lys-OMe·2HCI
L-Lysine, 99.6%, MP Biomedicals™
Chemical Name or Material: L-lysine, Free Base, Monohydrate CAS: 39665-12-8 Physical Form: Crystalline Powder Molecular Formula: C6H14N2O2 Formula Weight: 164.2 Synonym: L-2, 6-Diaminohexanoic acid
N-epsilon-Acetyl-L-lysine, 99%, MP Biomedicals™
Chemical Name or Material: N-epsilon-acetyl-L-lysine CAS: 692-04-6 Physical Form: Powder Assay Percent Range: 99% Molecular Formula: C8H16N2O3 Formula Weight: 188.3 MDL Number: MFCD00002639
DL-Lysinamide Dihydrochloride, MP Biomedicals™
Chemical Name or Material: DL-Lysinamide Dihydrochloride CAS: 51127-08-3 Molecular Formula: C6H15N3O.2HCl Formula Weight: 218.1
L-lysine, free base, MP Biomedicals™
Chemical Name or Material: L-lysine Name Note: Free base CAS: 39665-12-8 Physical Form: Crystalline Powder Molecular Formula: C6H14N2O2 Formula Weight: 164.2 MDL Number: MFCD00064433 Synonym: (S)-2, 6-Diaminocaproic acid
L-Lysine-L-Glutamate, 99.4%, MP Biomedicals™
Chemical Name or Material: L-Lysine-L-Glutamate CAS: 5408-52-6 Physical Form: Powder Molecular Formula: C6H14N2O2 · C5H9NO4 Formula Weight: 293.3 MDL Number: MFCD00038974
  spinner