Malic Acid
DL-Malic acid, +99%, ACROS Organics™
Chemical Name or Material: DL-Malic acid Assay Percent Range: 99+% Molecular Formula: C4H6O5 Linear Formula: HO2CCH2CH(OH)CO2H Formula Weight: 134.09 MDL Number: MFCD00064212 Synonym: DL-Hydroxysuccinic acid
D(+)-Malic acid, 98%, ACROS Organics™ Available on GSA/VA Contract for Federal Government customers only.
Chemical Name or Material: D(+)-Malic acid Assay: ≥97.5% Assay Percent Range: 98% Molecular Formula: C4H6O5 Linear Formula: HO2CCH2CH(OH)CO2H Formula Weight: 134.09 MDL Number: MFCD00004245
L-(-)-Malic acid, 97%
Chemical Name or Material: L-(-)-Malic Acid CAS: 97-67-6 Molecular Formula: C4H6O5 Formula Weight: 134.09 MDL Number: MFCD00064213 Beilstein: 1723541 Merck Index: 14,5707
L(-)-Malic acid, 99%, ACROS Organics™
Chemical Name or Material: L(-)-Malic acid Assay: ≥98.5% Assay Percent Range: 99% Molecular Formula: C4H6O5 Linear Formula: HO2CCH2CH(OH)CO2H Formula Weight: 134.09 MDL Number: MFCD00064213 Synonym: Apple acid
DL-Malic acid, 98%, Alfa Aesar
Chemical Name or Material: DL-Malic acid Assay Percent Range: 0.98 Molecular Formula: C4H6O5 Formula Weight: 134.09
L-malic acid, 99%, MP Biomedicals™
Chemical Name or Material: L-Malic acid CAS: 97-67-6 Physical Form: Powder Molecular Formula: C4H6O5 Formula Weight: 134.1 Synonym: L-Hydroxybutanedioic acid
L-Malic acid, 99%, Cell culture reagent grade, MP Biomedicals™
Chemical Name or Material: L-Malic acid CAS: 97-67-6 Purity Grade: Cell Culture Reagent Grade Physical Form: Powder Assay Percent Range: 99% Molecular Formula: C4H6O5 Formula Weight: 134.1 Synonym: L -Hydroxysuccinic acid
  spinner