Manganese -Mn
Manganese(IV) oxide, Brown powder, Puratronic™, 99.997% (Metals basis), Alfa Aesar™
Chemical Name or Material: Manganese(IV) oxide CAS: 1313-13-9 Physical Form: Powder Assay Percent Range: 99.997% (Metals basis) Molecular Formula: MnO2 Formula Weight: 86.94 MDL Number: MFCD00003463
Manganese(II) nitrate hydrate, 99.999% (metals basis), Alfa Aesar™, Puratronic™
Chemical Name or Material: Manganese(II) nitrate hydrate CAS: 15710-66-4 Physical Form: Crystalline Aggregates Assay Percent Range: 99.999% (Metal basis) Molecular Formula: Mn(NO3)2·xH2O Formula Weight: 178.95 (Anhydrous) MDL Number: MFCD00149788
Manganese Sulfate Monohydrate (Granular Powder/USP/FCC), Fisher Chemical
Chemical Name or Material: Manganese Sulfate Monohydrate CAS: 10034-96-5 Purity Grade: FCC/USP Physical Form: Solid Assay: 98.0 to 102.0% (USP) Molecular Formula: MnSO4 · H2O Formula Weight: 169.02 MDL Number: MFCD00149159 Synonym: Manganese (IV) Sulfate Monohydrate
Manganese Powder, -325 Mesh, 99.95% (Metal Basis), Alfa Aesar™
Chemical Name or Material: Manganese CAS: 7439-96-5 Physical Form: Powder Assay Percent Range: 99.95%, metal basis MDL Number: MFCD00011111
Manganese Powder, <10 micron, 99.6% (Metals basis), Alfa Aesar™
Chemical Name or Material: Manganese CAS: 7439-96-5 Physical Form: Powder Assay Percent Range: 99.6% (Metals basis) MDL Number: MFCD00011111
Manganese(II) carbonate, 99.985% (Metals basis), Alfa Aesar™, Puratronic™
Chemical Name or Material: Manganese(II) carbonate CAS: 598-62-9 Physical Form: Powder Assay Percent Range: 99.99% (metals basis) Molecular Formula: CMnO3 Formula Weight: 114.95 MDL Number: MFCD00011116
Manganese(II) nitrate, Alfa Aesar™
Chemical Name or Material: Manganese(II) nitrate Name Note: 50% w/w aq. Soln CAS: 10377-66-9 Physical Form: Liquid Molecular Formula: MnN2O6 Formula Weight: 178.95 MDL Number: MFCD00011112
Manganese Nitrate Solution, 50% Products for Science Education
Chemical Name or Material: Manganese Nitrate Solution, 50% CAS: 7697-37-2 CAS: 1377-66-9 CAS: 7732-18-5 Synonym: Maganous Nitrate Solution
Manganese(II) chloride, 97%, ACROS Organics™
Chemical Name or Material: Manganese(II) chloride Assay: ≥96.0% (on dry substance) Assay Percent Range: 97% Molecular Formula: Cl2Mn Linear Formula: MnCl2 Formula Weight: 125.84 MDL Number: MFCD00011114
Manganese(II) oxide, 99.99%, (trace metal basis), ACROS Organics™ Available on GSA/VA Contract for Federal Government customers only.
Chemical Name or Material: Manganese(II) oxide CAS: 1344-43-0 Purity Grade: Trace Metal Basis Assay: 76.7 to 78.2% (Mn) Assay Percent Range: 99.99% Molecular Formula: MnO Formula Weight: 70.94
Manganese Peroxide Products for Science Education
Chemical Name or Material: Manganese Dioxide CAS: 1313-13-9 Assay Percent Range: >80% Linear Formula: MnO2 Formula Weight: 86.94 Synonym: Manganese (IV) Oxide
Manganese(II) Sulfate Products for Science Education
Chemical Name or Material: Manganous Sulfate CAS: 10034-96-5 Purity Grade: Laboratory Grade Assay Percent Range: >98% Linear Formula: MnSO4·H2O Formula Weight: 169.01 Synonym: Manganese (II) Sulfate Monohydrate
Manganese(II) nitrate hydrate, 99.98% (metals basis), Alfa Aesar™
Chemical Name or Material: Manganese(II) nitrate hydrate CAS: 15710-66-4 Physical Form: Lump Assay Percent Range: 99.98% (metals basis) Molecular Formula: Mn(NO3)2·xH2O Formula Weight: 178.95 MDL Number: MFCD00149788
Manganese(IV) oxide, 99%, -10 mesh, ACROS Organics™
Chemical Name or Material: Manganese(IV) oxide Name Note: 99+%, -100 mesh CAS: 1313-13-9 Assay Percent Range: 99% Molecular Formula: MnO2 Linear Formula: MnO2 Formula Weight: 86.94 Synonym: Manganese dioxide
Manganese(II) fluoride, Red Crystalline Powder, 99%, Alfa Aesar™
Chemical Name or Material: Manganese(II) fluoride CAS: 7782-64-1 Physical Form: Crystalline Powder Assay Percent Range: 99% Molecular Formula: F2Mn Formula Weight: 92.93 MDL Number: MFCD00016222
Manganese(III) acetate dihydrate, 98%, ACROS Organics™
Chemical Name or Material: Manganese(III) acetate dihydrate Name Note: 98% CAS: 19513-05-4 Assay: ≥97.5% Assay Percent Range: 98% Molecular Formula: C6H9MnO6·2H2O Linear Formula: (CH3CO2)3Mn·2H2O Formula Weight: 268.09 MDL Number: MFCD00150022
anganese(II) sulfate hydrate, 99.999% (metals basis), Alfa Aesar™, Puratronic™
Chemical Name or Material: Manganese(II) sulfate hydrate Name Note: Loses all water at 400-450° CAS: 15244-36-7 Physical Form: Crystalline Powder Assay Percent Range: 99.999% (Metal basis) Molecular Formula: MnSO4·xH2O Formula Weight: 151.00 (Anhydrous) MDL Number: MFCD00149159
Manganese(II) sulfate monohydrate, ACS, 98.0-101.0%, Alfa Aesar™
Chemical Name or Material: Manganese(II) sulfate monohydrate CAS: 10034-96-5 Purity Grade: ACS Grade Physical Form: Powder Assay Percent Range: 98.0-101.0% Molecular Formula: MnSO4·H2O Formula Weight: 169.02 (151.00 Anhydrous) MDL Number: MFCD00149159
Manganese(II) oxide, Mn 76.0-78.0%, typically 99%, Alfa Aesar
Chemical Name or Material: Manganese(II) oxide Name Note: Mn 76.0-78.0% CAS: 1344-43-0 Physical Form: Powder Assay Percent Range: Mn 76.0-78.0%, typically 99% Molecular Formula: MnO Formula Weight: 70.94 MDL Number: MFCD00016226
Manganese(IV) oxide, 80-85%, powder, ACROS Organics™ Available on GSA/VA Contract for Federal Government customers only.
Chemical Name or Material: Manganese(IV) oxide CAS: 1313-13-9 Physical Form: Powder Assay: ≥80% Assay Percent Range: 80-85% Molecular Formula: MnO2 Linear Formula: MnO2 Formula Weight: 86.94 MDL Number: MFCD00003463 Synonym: Manganese dioxide
Manganese(II) carbonate, Mn 44% min., Alfa Aesar™
Chemical Name or Material: Manganese(II) carbonate CAS: 598-62-9 Physical Form: Powder Assay Percent Range: Mn 44% min. Molecular Formula: CMnO3 Formula Weight: 114.95 MDL Number: MFCD00011116
Manganese Nanopowder, 30-50nm, 99.9% (Metals basis), Alfa Aesar™
Chemical Name or Material: Manganese CAS: 7439-96-5 Physical Form: Nanopowder Assay Percent Range: 99.9% (Metals basis) Molecular Formula: Mn(core)/Mn3O4(shell) MDL Number: MFCD00011111
Manganese(III) oxide, 98%
Chemical Name or Material: Manganese(III) oxide CAS: 1317-34-6 Molecular Formula: Mn2O3 Formula Weight: 157.86 MDL Number: MFCD00016217 Merck Index: 14,5737 Synonym: Manganese sesquioxide
Manganese(II) bromide tetrahydrate, 98%, ACROS Organics™
Chemical Name or Material: Manganese(II) bromide tetrahydrate Name Note: 98% CAS: 10031-20-6 Assay: ≥97.5% Assay Percent Range: 98% Molecular Formula: Br2Mn·4H2O Linear Formula: MnBr2·4H2O Formula Weight: 286.82 MDL Number: MFCD00149791
Manganese(II) chloride tetrahydrate, ACS, 98.0-101.0%, Alfa Aesar™
Chemical Name or Material: Manganese(II) chloride tetrahydrate CAS: 13446-34-9 Purity Grade: ACS Grade Physical Form: Crystalline Assay Percent Range: 98-101% Molecular Formula: Cl2Mn·4H2O Formula Weight: 197.90 (125.84anhydrous) MDL Number: MFCD00149792
Manganese(II) chloride tetrahydrate, Alfa Aesar™
Chemical Name or Material: Manganese(II) chloride tetrahydrate CAS: 13446-34-9 Physical Form: Crystalline Powder Assay Percent Range: 99% (Metal basis) Molecular Formula: Cl2Mn·4H2O Formula Weight: 197.90 (125.84 Anhydrous) MDL Number: MFCD00149792
Manganese pieces, irregular, 99.9% (metals basis), Alfa Aesar
Chemical Name or Material: Manganese CAS: 7439-96-5 Physical Form: Irregular Shaped Pieces Assay Percent Range: 99.9% (Metals basis) Molecular Formula: Mn Formula Weight: 54.94 MDL Number: MFCD00011111
Manganese(II) sulfate monohydrate, 99+%, extra pure, ACROS Organics™ Available on GSA/VA Contract for Federal Government customers only.
Chemical Name or Material: Manganese(II) sulfate monohydrate CAS: 10034-96-5 Purity Grade: Extra Pure Assay: ≥99.0% Assay Percent Range: 99+% Molecular Formula: MnO4S·H2O Linear Formula: MnSO4·H2O Formula Weight: 169.02 MDL Number: MFCD00149159
Manganese(II) acetate, +98%, anhydrous, ACROS Organics™ Available on GSA/VA Contract for Federal Government customers only.
Chemical Name or Material: Manganese(II) acetate Assay: ≥98.0% Assay Percent Range: 98+% Molecular Formula: C4H6MnO4 Linear Formula: Mn(OC(O)CH3)2 Formula Weight: 173.03 MDL Number: MFCD00013039
Manganese Irregular shaped pieces, 99.95% (Metals basis), Alfa Aesar™
Chemical Name or Material: Manganese CAS: 7439-96-5 Physical Form: Irregular Shaped Pieces Assay Percent Range: 99.95% (Metals basis) MDL Number: MFCD00011111
  spinner