Matrix Modifiers
Matrix Modifier, 0.2% Pd (2000ppm Pd), Ricca Chemical
Chemical Name or Material: Matrix Modifier CAS: 7732-18-5 CAS Min %: 92.0 CAS Max %: 95.0 CAS: 7697-37-2 CAS Min %: 6.16 CAS Max %: 6.42 CAS: 7440-05-3 CAS Min %: 0.15 CAS Max %: 0.15 CAS: 7647-01-0 CAS Min %: 0.03 CAS Max %: 0.03 Physical Form: Liquid
Matrix Modifier, 0.5% Pd (5,000ppm Pd), Ricca Chemical
Chemical Name or Material: Matrix Modifier CAS: 7732-18-5 CAS Min %: 88.0 CAS Max %: 92.0 CAS: 7697-37-2 CAS Min %: 9.29 CAS Max %: 9.67 CAS: 7440-05-3 CAS Min %: 0.35 CAS Max %: 0.36 CAS: 7647-01-0 CAS Min %: 0.06 CAS Max %: 0.06 Physical Form: Liquid
Matrix Modifier, 2.0%mg (20,000ppm Mg), Ricca Chemical
Chemical Name or Material: Matrix Modifier CAS: 7732-18-5 CAS Min %: 76.0 CAS Max %: 79.0 CAS: 13446-18-9 CAS Min %: 16.09 CAS Max %: 16.74 CAS: 7697-37-2 CAS Min %: 5.72 CAS Max %: 5.96 Physical Form: Liquid
Matrix Modifier, 1.0% Pd (10,000ppm Pd), Ricca Chemical
Chemical Name or Material: Matrix Modifier CAS: 7732-18-5 CAS Min %: 88.0 CAS Max %: 91.0 CAS: 7697-37-2 CAS Min %: 9.23 CAS Max %: 9.61 CAS: 7440-05-3 CAS Min %: 0.88 CAS Max %: 0.91 CAS: 7647-01-0 CAS Min %: 0.16 CAS Max %: 0.16 Physical Form: Liquid
Matrix Modifier, 5% Ni (50,000ppm Ni), Ricca Chemical
Chemical Name or Material: Matrix Modifier CAS: 7732-18-5 CAS Min %: 69.0 CAS Max %: 72.0 CAS: 7697-37-2 CAS Min %: 25.82 CAS Max %: 26.88 CAS: 7440-02-0 CAS Min %: 3.44 CAS Max %: 3.58 Physical Form: Liquid
Matrix Modifier, 40% (w/v) Ammonium Phosphate, Ricca Chemical
Chemical Name or Material: Matrix Modifier Name Note: 40% (w/v) Ammonium Phosphate CAS: 7732-18-5 CAS Min %: 59.0 CAS Max %: 61.0 CAS: 7783-28-0 CAS Min %: 39.0 CAS Max %: 41.0 Physical Form: Liquid
Palladium Matrix Modifier, 2,000ppm in 5% HNO, Ricca Chemical
Chemical Name or Material: Palladium Matrix Modifier Name Note: 2, 000 ppm in 5% HNO CAS: 7732-18-5 CAS Min %: 92.0 CAS Max %: 95.0 CAS: 7697-37-2 CAS Min %: 6.16 CAS Max %: 6.42 CAS: 7440-05-3 CAS Min %: 0.15 CAS Max %: 0.15 CAS: 7647-01-0 CAS Min %: 0.03 CAS Max %: 0.03 Physical Form: Liquid
Matrix Modifier, Ammonium Nitrate, 5% in 2% Nitric Acid, Ricca Chemical
Chemical Name or Material: Matrix Modifier Name Note: Ammonium Nitrate CAS: 7732-18-5 CAS Min %: 91.0 CAS Max %: 94.0 CAS: 6484-52-2 CAS Min %: 4.54 CAS Max %: 4.72 CAS: 7697-37-2 CAS Min %: 2.73 CAS Max %: 2.84 Physical Form: Liquid
Matrix Modifier, 2% Ammonium Nitrate in 2% HNO3, Ricca Chemical
Chemical Name or Material: Matrix Modifier CAS: 7732-18-5 CAS Min %: 93.0 CAS Max %: 97.0 CAS: 7697-37-2 CAS Min %: 2.89 CAS Max %: 3.01 CAS: 6484-52-2 CAS Min %: 1.93 CAS Max %: 2.0
Lithium Metaborate (100%), Fusion Flux Pure Crystal For Sample Preparation, SPEX CertiPrep™ Available on GSA/VA Contract for Federal Government customers only.
Chemical Name or Material: Lithium Metaborate CAS: Various Physical Form: Crystalline
Lithium Tetraborate (100%) Fusion Flux Pure Crystal For Sample Preparation, SPEX CertiPrep™ Available on GSA/VA Contract for Federal Government customers only.
Chemical Name or Material: Lithium Tetraborate CAS: Various Physical Form: Crystalline
Lithium Metaborate (100%), Fusion Flux Ultra-Pure Crystal For Sample Preparation, SPEX CertiPrep™
Chemical Name or Material: Lithium Metaborate CAS: Various Physical Form: Crystalline
Lithium Tetraborate (100%) Fusion Flux Ultra-Pure Crystal For Sample Preparation, SPEX CertiPrep™
Chemical Name or Material: Lithium Tetraborate CAS: Various Physical Form: Crystalline
Ammonium Nitrate Matrix Modifier, 10,000μg/mL in Water w/Dilute Nitric Acid, Ultra Scientific
Chemical Name or Material: Ammonium Nitrate Matrix Modifier Name Note: 10, 000μg/mL CAS: 007732-18-5 CAS: 007697-37-2 CAS: 006484-52-2 Physical Form: Liquid
Palladium Matrix Modifier, at 2000μg/mL in Water with Dilute Nitric Acid, Ultra Scientific
Chemical Name or Material: Palladium Matrix Modifier Name Note: 2000μg/mL CAS: 007732-18-5 CAS: 007697-37-2 CAS: 007440-05-3 Physical Form: Liquid
Nickel Matrix Modifier, 4000μg/mL in water with dilute nitric acid, Ultra Scientific
Chemical Name or Material: Nickel Matrix Modifier Name Note: 4000μg/mL CAS: 007732-18-5 CAS: 007697-37-2 CAS: 007440-02-0 Physical Form: Liquid
Sodium Nitrate, Fusion Flux Additive (Crystalline/For Sample Prepn.)
Chemical Name or Material: Sodium Nitrate, Fusion Flux Additive Physical Form: Crystalline
Lithium Bromide, Fusion Flux Additive (Crystalline or Liq.; for Sample Prepn.)
Chemical Name or Material: Lithium Bromide, Fusion Flux Additive Name Note: Contents at 100μg/mL
Potassium Iodide, Fusion Flux Additive (Crystalline or Liq.; for Sample Prepn.)
Chemical Name or Material: Potassium Iodide, Fusion Flux Additive Physical Form: Crystalline or Liquid
  spinner