Metal Alloys
Magnesium Aluminum Zinc foil, Alfa Aesar™
Chemical Name or Material: Magnesium Aluminum Zinc foil Physical Form: ∽25g/30X30cm MDL Number: MFCD01074901
Nitinol Wire, 0.25mm (0.01 in.) dia., Alfa Aesar™
Chemical Name or Material: Nitinol Name Note: Alloy N, Straight Annealed Physical Form: Wire, 0.25mm (0.01 in.) dia. MDL Number: MFCD02091734
Bronze Gauze, 20 mesh woven from 0.41mm (0.016 in.) in dia. wire, Alfa Aesar™
Chemical Name or Material: Bronze Physical Form: Gauze MDL Number: MFCD00198187
Aluminum Foil, alloy 6061, 1.27mm (0.05 in.) thick, , Alfa Aesar™
Chemical Name or Material: Aluminum Physical Form: Foil, Alloy 6061, 1.27mm (0.05 in.) thick MDL Number: MFCD00134029
Stainless Steel Foil, 0.2mm (0.008 in.) thick, Alfa Aesar™
Chemical Name or Material: Stainless Steel Name Note: Type 304 Physical Form: Foil, 0.2mm (0.008 in.) thick Molecular Formula: Fe:Cr:Ni; 70:19:11 wt% MDL Number: MFCD00148499
Aluminum nitride Rod, Round, 22mm diameter, Alfa Aesar™
Chemical Name or Material: Aluminum nitride CAS: 24304-00-5 MDL Number: MFCD00003429
Nitinol wire, 1mm, Alfa Aesar™
Chemical Name or Material: Nitinol Physical Form: Wire, 1.0mm (0.039 in.) dia. MDL Number: MFCD02091734
Devarda's Alloy, granular, Alfa Aesar™
Chemical Name or Material: Devarda's Alloy CAS: 8049-11-4 Physical Form: Granular Molecular Formula: Cu:Al:Zn;50:45:5 MDL Number: MFCD00198176
Copper Nickle Wire, 0.13mm (0.005 in.) dia, Alfa Aesar™
Chemical Name or Material: Copper Nickle Physical Form: Wire, 0.13mm (0.005 in.) dia. Molecular Formula: Cu:Ni 55:45 wt% MDL Number: MFCD00801098
Nitinol tension coil spring, Alloy M, Alfa Aesar™
Chemical Name or Material: Nitinol tension coil spring Name Note: Wire diameter 0.75mm, mean coil diameter 5.55mm CAS: 7440-02-0 CAS: 7440-32-6 Physical Form: Solid MDL Number: MFCD02091734
Alfa Aesar™ Nickel Chromium wire, 0.25mm dia., Alfa Aesar™
Chemical Name or Material: Nickel Chromium CAS: 11106-97-1 Molecular Formula: Ni:Cr; 80:20 wt% MDL Number: MFCD00673110
Platinum Ruthenium black, nominally Pt 50%, Ru 50% (Atomic wt%), Alfa Aesar™
Chemical Name or Material: Platinum Ruthenium black Name Note: HiSPEC 6000 Physical Form: Solid Molecular Formula: Pt:Ru; 50:50 MDL Number: MFCD00798552
Lithium aluminium hydride, 95%, powder, ACROS Organics™
Chemical Name or Material: Lithium aluminium hydride CAS: 16853-85-3 Physical Form: Powder Assay Percent Range: 95% Molecular Formula: AlH4Li Linear Formula: LiAlH4 Formula Weight: 37.95 MDL Number: MFCD00011075 Synonym: LAH
Stainless Steel Spheres, 4mm (0.16 in.) dia., Alfa Aesar™
Chemical Name or Material: Stainless Steel Physical Form: Spheres, 4mm (0.16 in.) dia. Molecular Formula: Fe:Cr:Ni:Mo; 67.5:17:13:12.5 wt% MDL Number: MFCD00148499
Nitinol wire, , 0.3mmAlfa Aesar™
Chemical Name or Material: Nitinol wire Physical Form: ∽5.52g/10m; Af approx. 10°C MDL Number: MFCD02091734
Platinum Iridium Wire, 0.25mm (0.01 in.) dia., Alfa Aesar™
Chemical Name or Material: Platinum Iridium Physical Form: Wire, 0.25mm (0.01 in.) dia. Molecular Formula: Pt:Ir; 85:15 wt% MDL Number: MFCD02091683
Aluminum Copper Square bar, 1.27 x 1.27cm (0.5 x 0.5 in.), Alfa Aesar™
Chemical Name or Material: Aluminum Copper Physical Form: Square Bar, 1.27 X 1.27cm (0.5 X 0.5 in.) Molecular Formula: Al:Cu:Mg:Mn; 93.5:4.4:1.5:0.6 wt% MDL Number: MFCD00213972
Nickel Copper Foil, 0.51mm (0.02 in.) thick, Alfa Aesar™
Chemical Name or Material: Nickel Copper Name Note: Alloy 400 Physical Form: Foil, 0.5mm (0.02 in.) thick Molecular Formula: Ni:Cu; 67:33 wt.% MDL Number: MFCD00801098
Zinc copper couple, ACROS Organics™
Chemical Name or Material: Zinc copper couple CAS: 53801-63-1 Linear Formula: ZnCu MDL Number: MFCD00084851
Nitinol wire, , 0.58mmAlfa Aesar™
Chemical Name or Material: Nitinol Name Note: Alloy N, straight annealed Physical Form: Wire, 0.58mm (0.023 in.) dia. MDL Number: MFCD02091734
Nitinol Foil, 0.38mm (0.015 in.) thick, Alfa Aesar™
Chemical Name or Material: Nitinol Name Note: Flat annealed Physical Form: Foil, 0.38mm (0.015 in.) thick, MDL Number: MFCD02091734
Onion's Fusible Alloy Products for Science Education
Chemical Name or Material: Onion's Fusible Alloy CAS: 7440-31-5 CAS: 744-69-9 CAS: 7439-92-1 Assay Percent Range: 20% w/v
Nickel aluminum, 99.0% min (metals basis), Alfa Aesar™
Chemical Name or Material: Nickel aluminum CAS: 12003-78-0 Physical Form: Intermetallic Powder Assay Percent Range: 99.0% min. (metals basis) Molecular Formula: NiAl MDL Number: MFCD00167392
Aluminum-nickel, Raney-type Alloy, Powder Al-Ni 50/50, ACROS Organics™
Chemical Name or Material: Aluminium-nickel CAS: 12635-29-9 Molecular Formula: Al Ni Formula Weight: 85.68
Nitinol Wire, 0.5mm (0.02 in.) dia., Alfa Aesar™
Chemical Name or Material: Nitinol Name Note: Alloy N, Straight Annealed Physical Form: Wire, 0.5mm (0.02 in.) dia. MDL Number: MFCD02091734
Platinum Ruthenium foil, 0.05mm (0.002 in.) thick, Alfa Aesar™
Chemical Name or Material: Platinum Ruthenium Physical Form: Foil Molecular Formula: Pt:Ru; 95.2:4.8 MDL Number: MFCD00798552
Steel Products for Science Education
Chemical Name or Material: Steel
Gallium Indium alloy, 99.99% (metals basis), Alfa Aesar™
Chemical Name or Material: Gallium Indium alloy Physical Form: Solid Assay Percent Range: 99.99% (metals basis) Molecular Formula: Ga:In; 40:60 wt.% MDL Number: MFCD00144387
Nitinol Wire, 0.41mm (0.016 in.) dia., Alfa Aesar™
Chemical Name or Material: Nitinol Name Note: Alloy N, Straight Annealed Physical Form: Wire, 0.41mm (0.016 in.) dia. MDL Number: MFCD02091734
Stainless Steel Wire, 1.0mm (0.04 in.) dia., Alfa Aesar™
Chemical Name or Material: Stainless Steel Physical Form: Wire, 1.0mm (0.04 in.) dia. Molecular Formula: Fe:Cr:Ni; 70:19:11 wt% MDL Number: MFCD00148499
  spinner