Methionine -Met and Derivatives
DL-Methionine, 99+%, ACROS Organics™
Chemical Name or Material: DL-Methionine Name Note: 99+% CAS: 59-51-8 Assay: 99.0 to 101.0% (on dried substance) Assay Percent Range: 99+% Molecular Formula: C5H11NO2S Linear Formula: CH3SCH2CH2CH(NH2)CO2H Formula Weight: 149.21 MDL Number: MFCD00063096 Synonym: DL-2-Amino-4-(methylthio)butyric acid
BOC-L-Methionine, +99%, ACROS Organics™
Chemical Name or Material: BOC-L-Methionine Name Note: 99+% CAS: 2488-15-5 Assay: 99.0 to 101.0% Assay Percent Range: 99+% Molecular Formula: C10H19NO4S Linear Formula: CH3SCH2CH2CH(COOH)NHCOOC(CH3)3 Formula Weight: 249.33 MDL Number: MFCD00065586 Synonym: N-(tert-Butoxycarbonyl)-L-methionine
L-Methionine, 98+%, ACROS Organics™
Chemical Name or Material: L-Methionine Name Note: 98+% CAS: 63-68-3 Assay: 98.0 to 102.0% Total Base (on dried basis) Assay Percent Range: 98+% Molecular Formula: C5H11NO2S Linear Formula: CH3SCH2CH2CH(NH2)CO2H Formula Weight: 149.21 MDL Number: MFCD00063097 Synonym: L-2-Amino-4-(methylthio)butyric acid
FMOC-L-Methionine, 98%, ACROS Organics™
Chemical Name or Material: FMOC-L-Methionine Name Note: 98% CAS: 71989-28-1 Assay: ≥97.5% Assay Percent Range: 98% Molecular Formula: C20H21NO4S Linear Formula: FMOC-NHCH(COOH)CH2CH2SCH3 Formula Weight: 371.45 MDL Number: MFCD00037134 Synonym: N-(9-Fluorenylmethoxycarbonyl)-L-methionine
D-Methionine, 99+%, ACROS Organics™
Chemical Name or Material: D-Methionine Name Note: 99+% CAS: 348-67-4 Assay: 99.0 to 101.0% (on dried substance) Assay Percent Range: 99+% Molecular Formula: C5H11NO2S Linear Formula: CH3SCH2CH2CH(NH2)CO2H Formula Weight: 149.21 MDL Number: MFCD00002622
L-Methionine (White Crystals or Crystalline Powder), Fisher BioReagents Available on GSA/VA Contract for Federal Government customers only.
Chemical Name or Material: L-Methionine CAS: 63-68-3 Physical Form: Solid Assay: 98.5 to 100.5% Assay Percent Range: 98.5 to 100.5 % Molecular Formula: C5H11NO2S Formula Weight: 149.21
L-Methionine, 99+%, Powder, MP Biomedicals™
Chemical Name or Material: L-Methionine CAS: 63-68-3 Physical Form: Crystalline Powder Assay Percent Range: 99+% Molecular Formula: C5H11NO2S Formula Weight: 149.21 MDL Number: MFCD00063097 Synonym: 1-2-Amino-4-methyl thiobutanoir acid
L-Methionine, 99+%, MP Biomedicals
Chemical Name or Material: L-Methionine, 99+% CAS: 63-68-3 Purity Grade: Cell Culture Reagent Grade Physical Form: Crystalline Molecular Formula: C5H11NO2S Formula Weight: 149.2 Synonym: L-2-Amino-4-methyl-thiobutanoic acid
L-Methionine Methyl Ester Hydrochloride, 99+%, MP biomedicals™
Chemical Name or Material: L-Methionine Methyl Ester Hydrochloride Name Note: For TLC analysis CAS: 2491-18-1 Physical Form: Powder Assay Percent Range: 99+% Molecular Formula: C6H13NO2S · HCl Formula Weight: 199.7 MDL Number: MFCD00012491
  spinner