Methyl-2-pyrrolidinone -1-
1-Methyl-2-pyrrolidinone, 99+%, for spectroscopy, ACROS Organics™ Available on GSA/VA Contract for Federal Government customers only.
Chemical Name or Material: 1-Methyl-2-pyrrolidinone Name Note: 99+% CAS: 872-50-4 Purity Grade: For spectroscopy Assay: ≥99.0% Assay Percent Range: 99+% Molecular Formula: C5H9NO Formula Weight: 99.13 MDL Number: MFCD00003193 Synonym: N-Methylpyrrolidone
1-Methyl-2-pyrrolidinone, 99.5%, Extra Dry, AcroSeal™, ACROS Organics™ Available on GSA/VA Contract for Federal Government customers only.
Chemical Name or Material: 1-Methyl-2-pyrrolidinone CAS: 872-50-4 CAS Min %: 99.4 CAS Max %: 100.0 Purity Grade: Extra Dry Assay: ≥99.4% (GC) Assay Percent Range: 99.5% Molecular Formula: C5H9NO Formula Weight: 99.13 MDL Number: MFCD00003193 Synonym: N-Methylpyrrolidone
1-Methyl-2-pyrrolidinone, 99%, extra pure, ACROS Organics™ Available on GSA/VA Contract for Federal Government customers only.
Chemical Name or Material: 1-Methyl-2-pyrrolidinone CAS: 872-50-4 CAS Min %: 98.5 CAS Max %: 100.0 Purity Grade: Extra Pure Assay: ≥98.5% Assay Percent Range: 99% Molecular Formula: C5H9NO Formula Weight: 99.13 MDL Number: MFCD00003193 Synonym: N-Methylpyrrolidone
[2-(1-Pyrrolidinylmethyl)phenyl]magnesium bromide, 0.25M solution in THF, AcroSeal™, ACROS Organics™
Chemical Name or Material: [2-(1-Pyrrolidinylmethyl)phenyl]magnesium bromide CAS: 480424-80-4 CAS Min %: 4.32 CAS Max %: 10.07 CAS: 109-99-9 CAS Min %: 89.93 CAS Max %: 95.68 Molecular Formula: C11H14BrMgN Formula Weight: 264.44
[4-(1-Pyrrolidinylmethyl)phenyl]magnesium bromide, 0.25M solution in THF, AcroSeal™, ACROS Organics™
Chemical Name or Material: [4-(1-Pyrrolidinylmethyl)phenyl]magnesium bromide CAS: 480424-78-0 CAS Min %: 4.33 CAS Max %: 10.1 CAS: 109-99-9 CAS Min %: 89.9 CAS Max %: 95.67 Molecular Formula: C11H14BrMgN Formula Weight: 264.44
1-Methyl-2-pyrrolidinone, 99.5%, Extra Dry over Molecular Sieve, AcroSeal™, ACROS Organics™
Chemical Name or Material: 1-Methyl-2-pyrrolidinone Name Note: Over Molecular Sieves CAS: 872-50-4 CAS Min %: 99.4 CAS Max %: 100.0 Purity Grade: Extra Dry over Molecular Sieve Assay: ≥99.5% (GC) Assay Percent Range: 99.5% Molecular Formula: C5H9NO Formula Weight: 99.13 MDL Number: MFCD00003193 Synonym: N-Methylpyrrolidone
[3-(1-Pyrrolidinylmethyl)phenyl]magnesium bromide, 0.25M solution in THF, AcroSeal™, ACROS Organics™
Chemical Name or Material: [3-(1-Pyrrolidinylmethyl)phenyl]magnesium bromide CAS: 480424-79-1 CAS Min %: 4.3 CAS Max %: 10.04 CAS: 109-99-9 CAS Min %: 89.96 CAS Max %: 95.7 Molecular Formula: C11H14BrMgN Formula Weight: 264.44
1-Methyl-2-pyrrolidinone, 99.5%, for HPLC, ACROS Organics™ Available on GSA/VA Contract for Federal Government customers only.
Chemical Name or Material: 1-Methyl-2-pyrrolidinone CAS: 872-50-4 CAS Min %: 99.4 CAS Max %: 100.0 Purity Grade: For HPLC Assay: ≥99.5% Assay Percent Range: 99.5% Molecular Formula: C5H9NO Formula Weight: 99.13 MDL Number: MFCD00003193 Synonym: N-Methylpyrrolidone
1-Methyl-2-pyrrolidinone, ACS reagent, ACROS Organics™ Available on GSA/VA Contract for Federal Government customers only.
Chemical Name or Material: 1-Methyl-2-pyrrolidinone CAS: 872-50-4 Purity Grade: ACS Reagent Grade Assay: ≥99.5% Molecular Formula: C5H9NO Formula Weight: 99.13 Synonym: N-Methylpyrrolidone
1-Methyl-2-pyrrolidinone, 99.5%, Extra Dry, AcroSeal™, ACROS Organics™ Available on GSA/VA Contract for Federal Government customers only.
Chemical Name or Material: 1-Methyl-2-pyrrolidinone CAS: 872-50-4 Purity Grade: Extra Dry Assay: ≥99.5% Assay Percent Range: 99.5% Molecular Formula: C5H9NO Formula Weight: 99.13 MDL Number: MFCD00003193 Synonym: N-Methylpyrrolidone
N-Methyl-2-pyrrolidone, 99.0% ChromAR Grade ACS), Macron Fine Chemicals™
Chemical Name or Material: N-Methyl-2-Pyrrolidone, 99.0% CAS: 872-50-4 Formula Weight: 99.13
1-Methyl-2-Pyrrolidone, EMD Millipore Available on GSA/VA Contract for Federal Government customers only.
Chemical Name or Material: 1-Methyl-2-Pyrrolidone Name Note: OmniPure™ CAS: 872-50-4 Physical Form: Liquid Linear Formula: (CH3)-NC(:O)CH2CH2CH2- Formula Weight: 99.13 Synonym: N-Methyl-2-Pyrrolidinone
  spinner