Methyl Isobutyl Ketone
Methyl iso-Butyl Ketone (Certified ACS), Fisher Chemical Available on GSA/VA Contract for Federal Government customers only.
Chemical Name or Material: Methyl iso-Butyl Ketone CAS: 108-10-1 Purity Grade: Certified ACS Reagent Grade Assay: ≥98.5% Assay Percent Range: ≥98.5 % Molecular Formula: C6H12O Formula Weight: 100.16 MDL Number: MFCD00008938 Synonym: 4-Methyl-2-Pentanone
Methyl Isobutyl Ketone ACS AR, Macron Fine Chemicals™
Chemical Name or Material: Methyl Isobutyl Ketone CAS: 108-10-1 Purity Grade: ACS Reagent Grade Molecular Formula: CH3COCH2CH(CH3)2 Formula Weight: 100.16
Methyl Isobutyl Ketone ChromAR, Macron Fine Chemicals™
Chemical Name or Material: Methyl Isobutyl Ketone CAS: 108-10-1 Molecular Formula: CH3COCH2CH(CH3)2 Formula Weight: 100.16
MIBK, GMP/ACS/USP/NF Grade, PHARMCO-AAPER
Chemical Name or Material: Methyl Isobutyl Ketone CAS: 108-10-1 Physical Form: Liquid Molecular Formula: C6H12O Formula Weight: 100.16 Synonym: 4-Methyl-2-Pentanone;
  spinner