MS Grade Chromatography Water
Water, LC/MS grade, Alfa Aesar™
Chemical Name or Material: Water CAS: 7732-18-5 Purity Grade: LC/MS grade Molecular Formula: H20 Formula Weight: 18.02 Solubility Information: Soluble in water. Insoluble in polar liquids. Recommended Storage: Suitable for any general chemical storage area. Protect from freezing Refractive Index: 1.34 Odor: Odorless Melting Point: 0°C Density: 1gm/cm3 Color: Colorless pH: 7 at 20°C
Water with 0.1% Trifluoroacetic Acid (v/v), Optima™ LC/MS Grade
Aluminum (Al): 20 ppb max. Assay Percent Range: 99.0 to 103.0 % Calcium (Ca): 50 ppb max. CAS: 76-05-1 CAS: 76-05-1 Chemical Name or Material: 0.1% Trifluoroacetic Acid in Water (v/v) Copper (Cu): 10 ppb max. Assay: 0.095 to 0.105% (v/v) Trifluoroacetic Acid Color: Clear Physical Form: Liquid Purity Grade: Optima™ LC/MS Health Hazard 2: CAUTION! Health Hazard 3: Emergency Overview
May cause eye, skin, and respiratory tract irritation. Use personal protective equipment. Ensure adequate ventilation. Wash off immediately with plenty of water for at least 15 minutes. Get medical attention immediately if symptoms occur. Rinse immediately with plenty of water, also under the eyelids, for at least 15 minutes. Obtain medical attention. Move to fresh air. If breathing is difficult, give oxygen. Get medical attention immediately if symptoms occur. Do not induce vomiting. Obtain medical attention. Obtain medical attention. .
NFPA
Health:1
Flammability:0
Instability:0 Identification: Pass Test Impurities: ≤20ppb (Ionic, Potassium) Iron (Fe): 10 ppb max. LCMS Gradient Suitability: Pass Test Lead (Pb): 10 ppb max. Magnesium (Mg): 10 ppb max. Manganese (Mn): 10 ppb max. Merck Index: 15, 9847 MDL Number: 4169 Molecular Formula: C2HF3O2 Name Note: Solvent Blends Nickel (Ni): 10 ppb max. Packaging: Glass Bottle Potassium (K): 20 ppb max. Protease: Protease free Residue after Evaporation: ≤1ppm Silver (Ag): 10 ppb max. Sodium (Na): 50 ppb max. Trifluoroacetic Acid: Inclusive between 0.095% Zinc (Zn): 20 ppb max.
Water, Optima™ LC/MS Grade Available on GSA/VA Contract for Federal Government customers only.
Aluminum (Al): 10 ppb max. Barium (Ba): 10 ppb max. Cadmium (Cd): 10 ppb max. Calcium (Ca): 20 ppb max. CAS: 7732-18-5 Certificate of Analysis: Actual Lot Analysis reported on label; Certificate of Actual Lot Analysis available on request Chemical Name or Material: Water Chromium (Cr): 10 ppb max. Cobalt (Co): 10 ppb max. Copper (Cu): 10 ppb max. Density: 1.000g/cm³ Boiling Point: 100°C Color: Colorless Formula Weight: 18.02 Melting Point: 0°C pH: 7 Physical Form: Liquid Purity Grade: Optima™ LC/MS Filtered Through: 0.03 micron filtered. Halogens: Not Detected Health Hazard 2: Low hazard for usual industrial or commercial handling. Health Hazard 3: Emergency Overview
Handle in accordance with good industrial hygiene and safety practice. Flush eyes with water as a precaution. Move to fresh air. If breathing is difficult, give oxygen. Do not induce vomiting. Obtain medical attention.
NFPA
Health: 0
Flammability: 0
Instability: 0 Impurities: ≤10ppb (Ionic, Potassium) Iron (Fe): 10 ppb max. LCMS at Negative Mode as Aldicarb: 50 ppb max. LCMS at Positive Mode as Reserpine: 50 ppb max. LCMS Gradient Suitability: ≤2 mAU - PDA (200-400nm) LCMS Negative Mode as Mecoprop: ≤300ppb LCMS Positive Mode as Propazine: ≤50ppb Lead (Pb): 10 ppb max. Magnesium (Mg): 10 ppb max. Manganese (Mn): 10 ppb max. Merck Index: 15, 10237 MDL Number: 11332 Molecular Formula: H2O Nickel (Ni): 10 ppb max. Packaging: Glass Bottle Physical State: Liquid Potassium (K): 10 ppb max. Protease: Protease free Purity Grade Notes: Formulated for UHPLC-UV Residue after Evaporation: ≤1ppm Silver (Ag): 10 ppb max. Sodium (Na): 20 ppb max. Tin (Sn): 10 ppb max. Vapor Pressure: 17.5mmHg at 20°C Zinc (Zn): 10 ppb max.
Water with 0.1% Formic Acid (v/v), Optima™ LC/MS Grade Available on GSA/VA Contract for Federal Government customers only.
Aluminum (Al): 20 ppb max. Assay Percent Range: ≥99.0 % Calcium (Ca): 50 ppb max. CAS: 64-18-6 CAS: 7732-18-5 Chemical Name or Material: 0.1% Formic Acid in Water (v/v) Copper (Cu): 10 ppb max. Density: 1g/cm³ Boiling Point: 100°C Color: Clear pH: 2 Physical Form: Liquid Purity Grade: Optima™ LC/MS Formic Acid: 0.095 to 0.105% Health Hazard 2: CAUTION! Health Hazard 3: Emergency Overview
May cause eye, skin, and respiratory tract irritation. Use personal protective equipment. Ensure adequate ventilation. Wash off immediately with plenty of water for at least 15 minutes. Get medical attention immediately if symptoms occur. Rinse immediately with plenty of water, also under the eyelids, for at least 15 minutes. Obtain medical attention. Move to fresh air. If breathing is difficult, give oxygen. Get medical attention immediately if symptoms occur. Do not induce vomiting. Obtain medical attention. Obtain medical attention. .
NFPA
Health:1
Flammability:0
Instability:0 Identification: Pass Test Iron (Fe): 10 ppb max. LCMS Gradient Suitability: Pass Test Lead (Pb): 10 ppb max. Magnesium (Mg): 10 ppb max. Manganese (Mn): 10 ppb max. Merck Index: 15, 4269 MDL Number: 3297 Molecular Formula: CH2O2 Name Note: Solvent Blends Nickel (Ni): 10 ppb max. Packaging: Glass Bottle Potassium (K): 20 ppb max. Protease: Protease free Residue after Evaporation: 1ppm max. Silver (Ag): 10 ppb max. Sodium (Na): 50 ppb max. Zinc (Zn): 20 ppb max.
Water, LC/MS grade, Acros Organics™ Available on GSA/VA Contract for Federal Government customers only.
Chemical Name or Material: Water CAS: 7732-18-5 Molecular Formula: H2O Linear Formula: H2O Formula Weight: 18.02 MDL Number: MFCD00011332
  spinner