Multicompendial Acetone
Acetone (NF/FCC), Fisher Chemical Available on GSA/VA Contract for Federal Government customers only.
Chemical Name or Material: Acetone CAS: 67-64-1 Purity Grade: NF/FCC Assay: NLT 99.5%, NMT 100.5%% (FCC) Molecular Formula: C3H6O Linear Formula: CH3COCH3 Formula Weight: 58.08 MDL Number: MFCD00008765 Synonym: 2-Propanone
Acetone (NF/FCC/EP), Fisher Chemical Available on GSA/VA Contract for Federal Government customers only.
Chemical Name or Material: Acetone CAS: 67-64-1 Purity Grade: BP/EP/FCC/JP/NF/USP Assay: NLT 99.5%, NMT 100.5%% (FCC) Molecular Formula: C3H6O Linear Formula: CH3COCH3 Formula Weight: 58.08 MDL Number: MFCD00008765 Synonym: 2-Propanone
Acetone (NF), Macron Fine Chemicals™
Chemical Name or Material: Acetone CAS: 67-64-1 Purity Grade: BP/EP/FCC/JP/NF/USP Molecular Formula: CH3COCH3 Formula Weight: 58.08
Acetone, puriss., ≥99% (GC), Honeywell™ Riedel-de Haën™
Vapor Pressure: 184mmHg (20°C) Vapor Density: 2 (vs air) Density: 0.791g/mL at 25°C, 0.790 to 0.792g/mL at 20°C Explosion Limit: 0.132 Flash Point: −17°C (1.4°F) Linear Formula: CH3COCH3 Name Note: Meets analytical specification of Ph. Eur.BP, NF MDL Number: MFCD00008765 UN Number: UN1090 Assay: ≥99% (GC) Formula Weight: 58.08g/mol Purity Grade: puriss. Chemical Name or Material: Acetone CAS: 67-64-1 Beilstein: 635680 Melting Point: −95°.C Physical State: Liquid Color: Colorless
Acetone, Tested according to Ph.Eur., Honeywell™ Riedel-de Haen™
Vapor Pressure: 184mmHg (20°C) Vapor Density: 2 (vs air) Density: 0.791g/mL at 25°C Color: Colorless Boiling Point: 56°C Melting Point: −94°C Explosion Limit: 0.132 Flash Point: −17°C (1.4°F) Linear Formula: CH3COCH3 Name Note: Tested According to Ph.Eur. MDL Number: MFCD00008765 UN Number: UN1090 Formula Weight: 58.08g/mol Chemical Name or Material: Acetone CAS: 67-64-1 Beilstein: 635680 Physical State: Liquid Refractive Index: n20/D 1.359(lit.)
Acetone, World/GMP, ACS/USP/NF/BP/EP Grade, PHARMCO-AAPER
Chemical Name or Material: Acetone CAS: 67-64-1 Physical Form: Liquid Molecular Formula: C3H6O Formula Weight: 58.08 Synonym: 2-Propanone
  spinner