Multicompendial Ethyl Acetate
Ethyl Acetate (NF), Fisher Chemical Available on GSA/VA Contract for Federal Government customers only.
Chemical Name or Material: Ethyl Acetate CAS: 141-78-6 Purity Grade: USP/NF/FCC/EP/BP/JP Assay: 99.0 to 100.5% (NF) Assay Percent Range: 99.0 to 100.5 % Molecular Formula: C4H8O2 Linear Formula: CH3COOC2H5 Formula Weight: 88.11g/mol MDL Number: MFCD00009171 Synonym: Acetic Acid Ethyl Ester
Ethyl Acetate, World/GMP, ACS/USP/NF/BP/EP Grade, PHARMCO-AAPER
Chemical Name or Material: Ethyl Acetate CAS: 141-78-6 Physical Form: Liquid Molecular Formula: C4H8O2 Formula Weight: 88.1g/mol Synonym: Acetic Acid Ethyl Ester
  spinner