Multicompendial Methanol
Methanol (NF), Fisher Chemical Available on GSA/VA Contract for Federal Government customers only.
Chemical Name or Material: Methanol CAS: 67-56-1 Purity Grade: BP/EP/FCC/JP/NF/USP Assay: ≥99.8% Assay Percent Range: ≥99.8 % Molecular Formula: CH4O Linear Formula: CH3OH Formula Weight: 32.04 MDL Number: MFCD00004595 Synonym: Methyl Alcohol
Isopropanol/Methanol, 60:40, Ricca Chemical
Chemical Name or Material: Isopropanol/Methanol Name Note: 60:40
Methanol, 80% (v/v), Ricca Chemical
Chemical Name or Material: Methanol CAS: 67-56-1 CAS: 7732-18-5
Methanol, World/GMP, ACS/USP/NF/BP/EP Grade, PHARMCO-AAPER
Chemical Name or Material: Methanol CAS: 67-56-1 Physical Form: Liquid Molecular Formula: CH4O Formula Weight: 32.04 Synonym: Methyl Alcohol
Methanol, puriss., ≥99.7% (GC), Honeywell™ Riedel-de Haën™
Vapor Pressure: 97.68mmHg (20°C), 410mmHg (50°C) Vapor Density: 1.11 (vs air) Density: 0.791 to 0.793g/mL at 20°C, 0.791g/mL at 25°C Explosion Limit: 0.36 Flash Point: 9°C (48.2°F) Ignition Point: 725°F Linear Formula: CH3OH Name Note: Meets analytical specification of Ph Eur MDL Number: MFCD00004595 UN Number: UN1230 Assay: ≥99.7% (GC) Formula Weight: 32.04g/mol Purity Grade: puriss. Chemical Name or Material: Methanol CAS: 67-56-1 Beilstein: 1098229 Melting Point: −98°C Boiling Point: 64.7°C Color: Colorless Physical State: Liquid
Methyl Alcohol NF, Macron™
Chemical Name or Material: Methyl Alcohol CAS: 67-56-1 Purity Grade: BP/EP/FCC/JP/NF/USP Molecular Formula: CH3OH Formula Weight: 32.04
  spinner