Nitrosylsulfuric Acid
Nitrosylsulfuric acid, 40% in sulfuric acid, ACROS Organics™
Chemical Name or Material: Nitrosylsulfuric acid Name Note: 40% CAS: 7782-78-7 CAS: 7664-93-9 Assay: 38 to 41% Molecular Formula: HNO5S Linear Formula: HOSO2NO2 Formula Weight: 127.08 MDL Number: MFCD00011495 Synonym: Nitrosyl sulfate
  spinner