ORP Standards
ORP Standard 200mV, Ricca Chemical
Chemical Name or Material: Oxidation-Reduction Potential Standard Name Note: +200mV vs. Ag/AgCl CAS: 7732-18-5 CAS Min %: 97.0 CAS Max %: 100.0 CAS: 7447-40-7 CAS Min %: 0.73 CAS Max %: 0.76 CAS: 14459-95-1 CAS Min %: 0.29 CAS Max %: 0.3 CAS: 13746-66-2 CAS Min %: 0.03 CAS Max %: 0.03
Light's Solution, Modified, Ricca Chemical
Chemical Name or Material: Light's Solution Name Note: Modified, Oxidation-Reduction Potential (ORP) Standard CAS: 7732-18-5 CAS Min %: 82.0 CAS Max %: 86.0 CAS: 7664-93-9 CAS Min %: 8.51 CAS Max %: 8.86 CAS: 7783-83-7 CAS Min %: 3.97 CAS Max %: 4.13 CAS: 7783-85-9 CAS Min %: 3.23 CAS Max %: 3.36 Physical Form: Liquid
ORP Standard, +600 mV vs. Ag/AgCl (4M KCl sat. with AgCl filling solution), Ricca Chemical
Chemical Name or Material: Oxidation-Reduction Potential Standard Name Note: +600mV vs. Ag/AgCl CAS: 7732-18-5 CAS Min %: 92.0 CAS Max %: 96.0 CAS: 7783-83-7 CAS Min %: 4.43 CAS Max %: 4.61 CAS: 7664-93-9 CAS Min %: 1.69 CAS Max %: 1.76 CAS: 7783-85-9 CAS Min %: 0.02 CAS Max %: 0.02 Physical Form: Liquid
Light's Solution, Ricca Chemical
Chemical Name or Material: Light's Solution Name Note: Oxidation-Reduction Potential (ORP) Standard CAS: 7732-18-5 CAS Min %: 82.0 CAS Max %: 86.0 CAS: 7664-93-9 CAS Min %: 8.51 CAS Max %: 8.86 CAS: 7783-83-7 CAS Min %: 3.97 CAS Max %: 4.13 CAS: 7783-85-9 CAS Min %: 3.23 CAS Max %: 3.36 Physical Form: Liquid
ZoBell's Solution, Modified, Oxidation-Reduction Potential (ORP) Standard, Ricca Chemical
Chemical Name or Material: ZoBell's Solution Name Note: Oxidation-Reduction Potential (ORP) Standard CAS: 7732-18-5 CAS Min %: 97.0 CAS Max %: 100.0 CAS: 14459-95-1 CAS Min %: 0.73 CAS Max %: 0.76 CAS: 7447-40-7 CAS Min %: 0.72 CAS Max %: 0.75 CAS: 13746-66-2 CAS Min %: 7.0E-4 CAS Max %: 7.0E-4 Physical Form: Liquid
ZoBell's Solution, Oxidation-Reduction Potential (ORP) Standard, Ricca Chemical
Chemical Name or Material: ZoBell's Solution Name Note: Oxidation-Reduction Potential (ORP) Standard CAS: 7732-18-5 CAS Min %: 97.0 CAS Max %: 100.0 CAS: 7447-40-7 CAS Min %: 0.73 CAS Max %: 0.76 CAS: 14459-95-1 CAS Min %: 0.23 CAS Max %: 0.24 CAS: 13746-66-2 CAS Min %: 0.11 CAS Max %: 0.11 Physical Form: Liquid
  spinner