Oxalic Acid
Oxalic acid, ACROS Organics™
Chemical Name or Material: Oxalic acid CAS: 144-62-7 Molecular Formula: C2H2O4 Linear Formula: HO2CCO2H Formula Weight: 90.04 MDL Number: MFCD00002573 Synonym: Ethanedionic acid
Oxalic Acid Dihydrate (Crystalline/Technical), Fisher Chemical
Chemical Name or Material: Oxalic Acid Dihydrate CAS: 6153-56-6 Purity Grade: Technical Grade Physical Form: Powder/Solid Molecular Formula: C2H2O4 · 2 H2O Linear Formula: HO2CCO2H·2H2O Formula Weight: 126.07 MDL Number: MFCD00149102
Oxalic acid dihydrate, 99.5+%, ACS reagent, ACROS Organics™ Available on GSA/VA Contract for Federal Government customers only.
Chemical Name or Material: Oxalic acid dihydrate Name Note: For analysis ACS CAS: 6153-56-6 Purity Grade: ACS Reagent Grade Assay: 99.5 to 102.5% Assay Percent Range: 99.5+% Molecular Formula: C2H2O4·2H2O Linear Formula: HO2CCO2H·2H2O Formula Weight: 126.04 MDL Number: MFCD00149102 Synonym: Ethanedionic acid
Oxalic Acid Dihydrate (Crystalline/Certified ACS), Fisher Chemical
Chemical Name or Material: Oxalic Acid Dihydrate CAS: 6153-56-6 Purity Grade: Certified ACS Grade Physical Form: Powder/Solid Assay: 99.5 to 102.5% Assay Percent Range: 99.5 to 102.5 % Molecular Formula: C2H2O4 · 2 H2O Linear Formula: HO2CCO2H·2H2O Formula Weight: 126.07 MDL Number: MFCD00149102 Synonym: Ethanedioic Acid
Oxalic acid, 98%, anhydrous, ACROS Organics™ Available on GSA/VA Contract for Federal Government customers only.
Chemical Name or Material: Oxalic acid CAS: 144-62-7 Assay: ≥97.5% Assay Percent Range: 98% Molecular Formula: C2H2O4 Formula Weight: 90.04 MDL Number: MFCD00002573 Synonym: Ethanedionic acid
Oxalic acid dihydrate, 99+%, extra pure, ACROS Organics™
Chemical Name or Material: Oxalic acid dihydrate CAS: 6153-56-6 Purity Grade: Extra Pure Assay: ≥99.0% (KMnO4 titration) (Titrimetry other) Assay Percent Range: 99+% Molecular Formula: C2H2O4·2H2O Linear Formula: HO2CCO2H·2H2O Formula Weight: 126.04 MDL Number: MFCD00149102 Synonym: Ethanedionic acid
Oxalic Acid, Dihydrate Products for Science Education
Chemical Name or Material: Oxalic Acid, Dihydrate CAS: 6153-56-6 Purity Grade: Laboratory Grade Assay Percent Range: >98% Linear Formula: H2C2O4· 2H2O Formula Weight: 126.07 Synonym: Ethanedioic Acid Dehydrate
Oxalic acid dihydrate, ACS, 99.5-102.5%
Chemical Name or Material: Oxalic acid dihydrate CAS: 6153-56-6 Purity Grade: ACS Grade Molecular Formula: C2H2O4·2H2O Linear Formula: HO2CCO2H·2H2O Formula Weight: 126.07 MDL Number: MFCD00149102 UN Number: UN3261 Beilstein: 3679436 Merck Index: 14,6911
Oxalic Acid, 10% (w/v) Aqueous Solution, Ricca Chemical
Chemical Name or Material: Oxalic Acid CAS: 7732-18-5 CAS Min %: 89.0 CAS Max %: 93.0 CAS: 6153-56-6 CAS Min %: 8.91 CAS Max %: 9.28 Physical Form: Liquid
Oxalic Acid, 10%, APHA, Macron Fine Chemicals™
Chemical Name or Material: Oxalic Acid
Oxalic Acid TS, 6.3% (w/v) Aqueous Solution, Ricca Chemical
Chemical Name or Material: Oxalic Acid Name Note: Test Solution CAS: 7732-18-5 CAS Min %: 92.0 CAS Max %: 96.0 CAS: 6153-56-6 CAS Min %: 5.81 CAS Max %: 6.05 Physical Form: Liquid
Oxalic Acid Stock Solution, Ricca Chemical
Chemical Name or Material: OXALIC ACID STOCK SOLUTION CAS: 7732-18-5 CAS Min %: 97.0 CAS Max %: 100.0 CAS: 6153-56-6 CAS Min %: 0.63 CAS Max %: 0.65
Oxalic Acid, 0.100 Normal (N/10), 0.0500 Molar (M/20) for Intel, Ricca Chemical
Chemical Name or Material: Oxalic Acid Name Note: 0.100 Normal (N/10), 0.0500 Molar (M/20) for Intel CAS: 7732-18-5 CAS: 6153-56-6 Physical Form: Liquid Assay Percent Range: 0.1000 ± 0.0001 N
Oxalic Acid, 10% (w/v), Aqueous Solution, Ricca Chemical
Chemical Name or Material: Oxalic Acid Name Note: 10% (w/v) CAS: 6153-56-6 CAS: 7732-18-5 Physical Form: Liquid
Oxalic Acid, 7.5% (w/v), Aqueous Solution, Ricca Chemical
Chemical Name or Material: Oxalic Acid Name Note: 7.5% (w/v), 0.4 Molar (0.8 N) CAS: 6153-56-6 CAS: 7732-18-5 Physical Form: Liquid
Oxalic Acid, 5% (w/v) Aqueous Solution, Ricca Chemical
Chemical Name or Material: Oxalic Acid Name Note: 5% (w/v) Aqueous Solution CAS: 7732-18-5 CAS: 6153-56-6 Physical Form: Liquid
Oxalic acid 2-hydrate, For ACS analysis, 99.5 to 102.5%, MP Biomedicals™
Chemical Name or Material: Oxalic acid 2-hydrate Name Note: For ACS analysis CAS: 6153-56-6 Physical Form: Crystals Molecular Formula: C2H2O4·2H2O Formula Weight: 126.07 Synonym: Ethanedioic acid dihydrate
Oxalic acid, dihydrate, 99.5-102.5%, ACS Grade, EMD Millipore
Chemical Name or Material: Oxalic acid, dihydrate CAS: 6153-56-6 Assay Percent Range: 99.5-102.5% Molecular Formula: HOOCCOOH·2H2O Formula Weight: 126 Synonym: Ethanedioic acid, dihydrate
Oxalic acid, dihydrate, 99.5 to 102.5%, ACS, EMD Millipore
Chemical Name or Material: Oxalic acid, dihydrate Name Note: For ACS analysis CAS: 6153-56-6 Physical Form: Crystals Assay Percent Range: 99.5 to 102.5% Molecular Formula: HOOCCOOH·2H2O Formula Weight: 126.07 Synonym: Ethanedioic Acid, Dihydrate
Oxalic Acid Dihydrate Products for Science Education
Chemical Name or Material: Oxalic Acid CAS: 6153-56-6 Linear Formula: C2H2O4·2H2O Formula Weight: 126.07
Oxalic Acid ACS AR Crystal, Macron Fine Chemicals™
Chemical Name or Material: Oxalic Acid Name Note: Dihydrate CAS: 6153-56-6 Purity Grade: ACS Grade Physical Form: Crystalline Molecular Formula: H2C2O4·2H2O Formula Weight: 126.07
  spinner