Pentane -n-
Pentane Products for Science Education
Chemical Name or Material: Pentane CAS: 109-66-0 Assay Percent Range: 100% Molecular Formula: C5H12 Formula Weight: 72.15 Synonym: Amyl Hydride
Pentane (Certified), Fisher Chemical
Chemical Name or Material: Pentane CAS: 109-66-0 Purity Grade: Certified Grade Assay: ≥98% Assay Percent Range: ≥98% Molecular Formula: C5H12 Linear Formula: CH3(CH2)3CH3 Formula Weight: 72.15 MDL Number: MFCD00009498
Pentane Products for Science Education
Chemical Name or Material: Pentane CAS: 109-66-0 Purity Grade: Reagent Grade Molecular Formula: C5H12 Linear Formula: CH3(CH2)3CH3
n-Pentane, +99%, AcroSeal™, ACROS Organics™ Available on GSA/VA Contract for Federal Government customers only.
Chemical Name or Material: n-Pentane Name Note: Extra Dry CAS: 109-66-0 CAS Min %: 99.0 CAS Max %: 100.0 Purity Grade: Extra Dry Assay: ≥99.0% Assay Percent Range: 99+% Molecular Formula: C5H12 Linear Formula: CH3(CH2)3CH3 Formula Weight: 72.15 MDL Number: MFCD00009498
n-Pentane, +99%, extra pure, anhydrous, ACROS Organics™
Chemical Name or Material: n-Pentane Name Note: p.a. CAS: 109-66-0 CAS Min %: 99.0 CAS Max %: 100.0 Purity Grade: Extra Pure Assay: ≥99% Assay Percent Range: 99+% Molecular Formula: C5H12 Linear Formula: CH3(CH2)3CH3 Formula Weight: 72.15 MDL Number: MFCD00009498
n-Pentane, +99%, extra pure, ACROS Organics™ Available on GSA/VA Contract for Federal Government customers only.
Chemical Name or Material: n-Pentane CAS: 109-66-0 CAS Min %: 99.0 CAS Max %: 100.0 Purity Grade: Extra Pure Assay: ≥99.0% Assay Percent Range: 99+% Assay Percent Notes: Determine assay (GC) instead of accepting value from supplier's Certificate of Analysi Molecular Formula: C5H12 Linear Formula: CH3(CH2)3CH3 Formula Weight: 72.15 MDL Number: MFCD00009498
n-Pentane, for analysis, ACROS Organics™
Chemical Name or Material: n-Pentane CAS: 109-66-0 Purity Grade: For analysis Assay: ≥99.0% Molecular Formula: C5H12 Linear Formula: CH3(CH2)3CH3 Formula Weight: 72.15 MDL Number: MFCD00009498
n-Pentane, 98%
Chemical Name or Material: n-Pentane CAS: 109-66-0 Molecular Formula: C5H12 Linear Formula: CH3(CH2)3CH3 Formula Weight: 72.15 MDL Number: MFCD00009498 UN Number: UN1265 Merck Index: 14,7116
n-Pentane, 95%, technical grade, ACROS Organics™
Chemical Name or Material: n-Pentane CAS: 109-66-0 CAS Min %: 94.0 CAS Max %: 100.0 Assay: ≥94% Assay Percent Range: 95% Molecular Formula: C5H12 Linear Formula: CH3(CH2)3CH3 Formula Weight: 72.15
Pentane UltimAR, Macron Fine Chemicals™
Chemical Name or Material: Pentane CAS: 109-66-0 Molecular Formula: CH3(CH2)3CH3 Formula Weight: 72.15
Pentane, 98% min., EMD Millipore Available on GSA/VA Contract for Federal Government customers only.
Chemical Name or Material: Pentane CAS: 109-66-0 Physical Form: Liquid Assay Percent Range: 98% Min. Molecular Formula: CH3(CH2)3CH3 Linear Formula: CH3(CH2)3CH3 Formula Weight: 72.15
  spinner