Phenylalanine -Phe and Derivatives
β-Methyl-DL-phenylalanine hydrochloride, 99%, ACROS Organics™
Chemical Name or Material: β-Methyl-DL-phenylalanine hydrochloride CAS: 80997-87-1 CAS Min %: 98.5 CAS Max %: 100.0 Assay: ≥98.5% Assay Percent Range: 99% Molecular Formula: C10H13NO2·HCl Linear Formula: C6H5CH(CH3)CH(NH2)CO2H·HCl Formula Weight: 215.68 MDL Number: MFCD00012623 Synonym: 2-Amino-3-phenylbutanoic acid hydrochloride
BOC-D-Phenylalanine, 99+%, ACROS Organics™ Available on GSA/VA Contract for Federal Government customers only.
Chemical Name or Material: BOC-D-Phenylalanine Assay: ≥99.0% Assay Percent Range: 99+% Molecular Formula: C14H19NO4 Linear Formula: C6H5CH2CH(COOH)nHCOOC(CH3)3 Formula Weight: 265.3 MDL Number: MFCD00063149 Synonym: N-(tert-Butoxycarbonyl)-D-phenylalanine
BOC-L-Phenylalanine, 99+%, ACROS Organics™ Available on GSA/VA Contract for Federal Government customers only.
Chemical Name or Material: BOC-L-Phenylalanine Assay: ≥99.0% Assay Percent Range: 99+% Molecular Formula: C14H19NO4 Linear Formula: C6H5CH2CH(COOH)nHCOOC(CH3)3 Formula Weight: 265.3 MDL Number: MFCD00002663 Synonym: N-(tert-Butoxycarbonyl)-L-phenylalanine
L-Phenylalanine, 98.5+%, ACROS Organics™
Chemical Name or Material: L-Phenylalanine Assay: 98.50 to 102.00% (on dried basis) Assay Percent Range: 98.5+% Molecular Formula: C9H11NO2 Linear Formula: C6H5CH2CH(NH2)CO2H Formula Weight: 165.19 MDL Number: MFCD00064227
L-Phenylalanine, 99%
Chemical Name or Material: L-Phenylalanine CAS: 63-91-2 Molecular Formula: C9H11NO2 Formula Weight: 165.19 MDL Number: MFCD00064227 Beilstein: 1910408 Merck Index: 14,7271
D-Phenylalanine, 99+%, ACROS Organics™
Chemical Name or Material: D-Phenylalanine Assay: ≥99.0% Assay Percent Range: 99+% Molecular Formula: C9H11NO2 Linear Formula: C6H5CH2CH(NH2)CO2H Formula Weight: 165.19 MDL Number: MFCD00004270
(R)-N-BOC-3-Fluorophenylalanine, 95%, 98% e.e., ACROS Organics™
Chemical Name or Material: (R)-N-BOC-3-Fluorophenylalanine Name Note: 98% ee CAS: 114873-11-9 CAS Min %: 94.0 CAS Max %: 100.0 Assay: ≥94% Assay Percent Range: 95% Molecular Formula: C14 H18 F N O4 Formula Weight: 283.3 MDL Number: MFCD00672523
p-Chlorophenylalanine, Tocris
Chemical Name or Material: p-Chlorophenylalanine CAS: 7424-00-2 Assay Percent Range: >99% Molecular Formula: C9H10ClNO2 Formula Weight: 199.64
Ochratoxin A, Tocris
Chemical Name or Material: Ochratoxin A CAS: 303-47-9 Assay Percent Range: >99% Molecular Formula: C20H18ClNO6 Formula Weight: 403.82 Synonym: N-[[(3R)-5-Chloro-3, 4-dihydro-8-hydroxy-3-methyl-1-oxo-1H-2-benzopyran-7-yl]carbonyl]-L-phenylalanine
L-phenylalanine, 99.85%, MP Biomedicals™
Chemical Name or Material: L-Phenylalanine CAS: 63-91-2 Physical Form: Powder Molecular Formula: C9H11NO2 Formula Weight: 165.2 Synonym: L-2-amino-3-phenylpropanoic acid
L-Phenylalanine, >99%, MP Biomedicals
Chemical Name or Material: L-Phenylalanine, >99% CAS: 63-91-2 Purity Grade: Cell Culture Reagent Grade Molecular Formula: C9H11NO2 Formula Weight: 165.2 Synonym: L-2-Amino-3-phenylpropanoic acid
N-t-Boc-Met-Lec-Phe, MP Biomedicals™
Chemical Name or Material: N-t-Boc-Met-Lec-Phe CAS: 67247-12-5 Molecular Formula: C25H39N3O6S Formula Weight: 509.7
D-phenylalanine, 99.9%, MP Biomedicals™
Chemical Name or Material: D-Phenylalanine CAS: 673-06-3 Physical Form: Powder Molecular Formula: C9H11NO2 Formula Weight: 165.2 Synonym: D-α-amino-β-phenyl-propionic acid
DL-Phenylalanine, ≥99%, MP Biomedicals™
Chemical Name or Material: DL-Phenylalanine Name Note: FTIR CAS: 150-30-1 Physical Form: Powder Assay Percent Range: ≥99% Molecular Formula: C9H11NO2 Formula Weight: 165.2 MDL Number: MFCD00064225 Synonym: α-Amino-β-Phenylpropionic Acid
4-nitro-L-phenylalanine, Maybridge
Chemical Name or Material: 2-Amino-3-(4-nitrophenyl)propanoic acid Assay: 97% Assay Percent Range: 97% Molecular Formula: C9H10N2O4 Formula Weight: 210.19
L-Phenylalanine (White Crystals or Crystalline Powder), Fisher BioReagents
Chemical Name or Material: L-Phenylalanine CAS: 63-91-2 Physical Form: Powder/Solid Assay: 98.5 to 100.5% Assay Percent Range: 98.5 to 100.5 % Molecular Formula: C9H11NO2 Formula Weight: 165.19
DL-β-3,4-Dihydroxyphenylalanine, 99%, MP Biomedicals™
Chemical Name or Material: DL-β-3, 4-Dihydroxyphenylalanine CAS: 63-84-3 Physical Form: Powder Molecular Formula: C9H11NO4 Formula Weight: 197.2 MDL Number: MFCD00063060 Synonym: 3-(3, 4-Dihydroxyphenyl)-DL-alanine
N-Glutaryl-L-Phenylalanine-p-Nitroanilide, MP Biomedicals™
Chemical Name or Material: N-Glutaryl-L-Phenylalanine-p-Nitroanilide CAS: 5800-34-0 Physical Form: Powder Assay Percent Range: 98.2% (HPLC), >99% (TLC) Molecular Formula: C20H21N3O6 Formula Weight: 399.4 Synonym: Glutaryl-L-phenylalanine 4-nitroanilide
  spinner