Phosphomolybdic Acid
Phosphomolybdic acid hydrate, ACS reagent, ACROS Organics™
Chemical Name or Material: Phosphomolybdic Acid hydrate CAS: 51429-74-4, 12026-57-2 CAS: 12026-57-2 Purity Grade: ACS Reagent Grade Physical Form: Crystals Molecular Formula: H3Mo12O40P·xH2O Linear Formula: H3[P(Mo3O10)4]·xH2O Formula Weight: 1825.25 MDL Number: MFCD00149913 Synonym: Molybdophosphoric Acid
Phosphomolybdic acid, ammonium salt hydrate, ACROS Organics™ Available on GSA/VA Contract for Federal Government customers only.
Chemical Name or Material: Phosphomolybdic acid, ammonium salt hydrate CAS: 54723-94-3 Molecular Formula: H12Mo12N3O40P·xH2O Linear Formula: (NH4)3PMo12O40·xH2O Formula Weight: 1876.35 MDL Number: MFCD00150423 Synonym: Ammonium phosphomolybdate hydrate
Phosphomolybdic Acid Hydrate (Crystalline/Certified ACS), Fisher Chemical Available on GSA/VA Contract for Federal Government customers only.
Chemical Name or Material: Phosphomolybdic Acid Hydrate CAS: 51429-74-4 Purity Grade: Certified ACS Grade Physical Form: Solid Molecular Formula: H3MO12O40P Formula Weight: 1825.25 MDL Number: MFCD00149913 Synonym: Molybdophosphoric Acid Hydrate
Phosphomolybdic Acid ACS AR Crystal, Macron Fine Chemicals™
Chemical Name or Material: Phosphomolybdic Acid CAS: 11104-88-4 Purity Grade: ACS Grade Physical Form: Crystalline Molecular Formula: H3Mo12O40P·xH2O
Phosphomolybdic Acid ACS MP Biomedicals
Chemical Name or Material: Phosphomolybdic Acid CAS: 51429-74-4 Purity Grade: ACS Grade Molecular Formula: 12(MoO3) - H3PO4- xH2O Formula Weight: 3939.45 Synonym: Hydrogen Tetrachlorolaurate Trihydrate
Phosphomolybdic Acid (ACS Grade), MP Biomedicals
Chemical Name or Material: Phosphomolybdic Acid CAS: 51429-74-4 Purity Grade: ACS Grade Formula Weight: 3939.5
  spinner