Picric Acid
Picric Acid, Saturated (AOAC), Fisher Chemical
Chemical Name or Material: Picric Acid, Saturated CAS: 7732-18-5 CAS Min %: 97.0 CAS Max %: 100.0 CAS: 88-89-1 CAS Min %: 1.26 CAS Max %: 1.31
Picric Acid, Reagent Grade, Ricca Chemical
Chemical Name or Material: Picric Acid CAS: 88-89-1 CAS Min %: 98.0 CAS Max %: 100.0 CAS: 7732-18-5 CAS Min %: 29.0 CAS Max %: 31.0 Purity Grade: Reagent Grade Physical Form: Crystalline Solid Packed In 30% Water
Picric Acid, Saturated Aqueous Solution, approximately 1.2% (w/v), Ricca Chemical
Chemical Name or Material: Picric Acid CAS: 7732-18-5 CAS Min %: 97.0 CAS Max %: 100.0 CAS: 88-89-1 CAS Min %: 1.26 CAS Max %: 1.31 Physical Form: Liquid
Trinitrophenol TS (Picric Acid TS), 1% (w/v) Aqueous Solution, Ricca Chemical
Chemical Name or Material: Trinitrophenol TS (Picric Acid TS) CAS: 7732-18-5 CAS Min %: 97.0 CAS Max %: 100.0 CAS: 88-89-1 CAS Min %: 1.26 CAS Max %: 1.31 Physical Form: Liquid
Picric Acid, 4% (w/v) in Denatured Ethanol (Reagent Alcohol), Ricca Chemical
Chemical Name or Material: Picric Acid CAS: 64-17-5 CAS Min %: 84.0 CAS Max %: 88.0 CAS: 88-89-1 CAS Min %: 4.67 CAS Max %: 4.86 CAS: 67-63-0 CAS Min %: 4.46 CAS Max %: 4.64 CAS: 88-89-1 (Picric Acid (2, 4, 6-Trinitrophenol)) Physical Form: Liquid
  spinner