Platinum -Pt
Alfa Aesar™ Platinum gauze, 52 mesh woven from 0.1mm dia. wire, 99.9% (metals basis), Alfa Aesar™
Chemical Name or Material: Platinum CAS: 7440-06-4 MDL Number: MFCD00011179
Platinum Wire, 0.5mm (0.02 in.) dia., hard, 99.95% (Metals basis), Alfa Aesar™
Chemical Name or Material: Platinum Name Note: Hard CAS: 7440-06-4 Physical Form: Wire, 0.5mm (0.02 in.) dia. Assay Percent Range: 99.95% (Metals basis) MDL Number: MFCD00011179
Dihydrogen dinitrosulfatoplatinate(II) solution, Pt 4-6% (cont. Pt), Alfa Aesar™
Chemical Name or Material: Dihydrogen dinitrosulfatoplatinate(II) solution Name Note: Pt 4-6% (cont. Pt) CAS: 12033-81-7 Physical Form: Liquid Molecular Formula: H2Pt(NO2)2SO4 Formula Weight: 384.17 (anhydrous) MDL Number: MFCD00064668 Synonym: Dinitrosulfatoplatinum(II)
Platinum Potassium Chloride (Laboratory), Fisher Chemical Available on GSA/VA Contract for Federal Government customers only.
Chemical Name or Material: Platinum Potassium Chloride CAS: 16921-30-5 Purity Grade: Laboratory Grade Molecular Formula: Cl6K2Pt Formula Weight: 486.01 MDL Number: MFCD00011389 Synonym: Potassium Chloroplatinate
Platinum(IV) oxide hydrate, typical Pt-content 79-84%, ACROS Organics™
Chemical Name or Material: Platinum(IV) oxide hydrate Assay: 79 to 84% (Pt) Molecular Formula: O2Pt·xH2O Linear Formula: PtO2 Formula Weight: 227.09 MDL Number: MFCD00066964 Synonym: Adams' catalyst
Platinum Wire, 0.25mm (0.010 in.) dia., 99.9% (Metals basis), Alfa Aesar™
Chemical Name or Material: Platinum CAS: 7440-06-4 Physical Form: Wire, 0.25mm (0.01 in.) dia. Assay Percent Range: 99.9% (Metals basis) MDL Number: MFCD00011179
Platinum wire, Alfa Aesar™
Chemical Name or Material: Platinum CAS: 7440-06-4 Purity Grade: For ISA Type R or S Standard Grade Physical Form: Wire, 0.25mm (0.01 in.) dia. MDL Number: MFCD00011179
Hydrogen hexachloroplatinate(IV) hydrate, 99.9%, (trace metal basis), 38 to 40% Pt, ACROS Organics™
Chemical Name or Material: Hydrogen hexachloroplatinate(IV) hydrate Name Note: 38 to 40% Pt CAS: 26023-84-7 Purity Grade: Trace Metal Basis Assay: 38 to 40% (Pt) Assay Percent Range: 99.9% Molecular Formula: H2Cl6Pt·6H2O Formula Weight: 517.92 MDL Number: MFCD00149910 Synonym: Hexachloroplatinic acid hexahydrate
Dihydrogen hexachloroplatinate(IV) hexahydrate, ACS, 99.95%, Alfa Aesar™, Premion™
Chemical Name or Material: Dihydrogen hexachloroplatinate(IV) hexahydrate CAS: 18497-13-7 Purity Grade: ACS Grade Physical Form: Lumps Assay Percent Range: 99.95% (metals basis) Molecular Formula: H2Cl6Pt·6H2O Formula Weight: 517.91 (409.82 Anhydrous) MDL Number: MFCD00149910
Platinum Granules, 6.35mm (0.25 in.) & down, 99.95% (Metals basis), Alfa Aesar™
Chemical Name or Material: Platinum CAS: 7440-06-4 Physical Form: Granular Assay Percent Range: 99.95% (Metals basis) MDL Number: MFCD00011179
Platinum wire, 99.95% (metals basis), Alfa Aesar™
Chemical Name or Material: Platinum Name Note: Hard CAS: 7440-06-4 Physical Form: Wire, 1.0mm (0.04 in.) dia. Assay Percent Range: 99.95% (Metals basis) MDL Number: MFCD00011179
Potassium tetrachloroplatinate(II), 46-47% Pt, ACROS Organics™ Available on GSA/VA Contract for Federal Government customers only.
Chemical Name or Material: Potassium tetrachloroplatinate(II) CAS: 10025-99-7 Molecular Formula: Cl4K2Pt Linear Formula: K2PtCl4 Formula Weight: 415.09 MDL Number: MFCD00011378
Platinum Foil, 0.25mm (0.01 in.) thick, Premion™, 99.99% (Metals basis), Alfa Aesar™
Chemical Name or Material: Platinum CAS: 7440-06-4 Physical Form: Foil, 0.25mm (0.01 in.) thick Assay Percent Range: 99.99% (Metals basis) MDL Number: MFCD00011179
Platinum on activated carbon, 5% Pt, not reduced, ACROS Organics™
Chemical Name or Material: Platinum on activated carbon Name Note: 5% Pt CAS: 2023600 Assay: 5% (Pt) Molecular Formula: Pt Formula Weight: 195.08 MDL Number: MFCD00011179
Platinum Wire, 0.025mm (0.001 in.) dia., 99.95% (Metals basis), Alfa Aesar™
Chemical Name or Material: Platinum CAS: 7440-06-4 Physical Form: Wire, 0.025mm (0.001 in.) dia. Assay Percent Range: 99.95% (Metals basis) MDL Number: MFCD00011179
Platinum wire, 0.25mm dia., Alfa Aesar™
Chemical Name or Material: Platinum CAS: 7440-06-4 Purity Grade: For ISA Type R or S Standard Grade MDL Number: MFCD00011179
Platinum, nominally 40% on carbon black, Alfa Aesar™
Chemical Name or Material: Platinum Assay Percent Range: nominally 40% on carbon black MDL Number: MFCD00011179
Platinum wire, For ISA Type R or S Standard Grade Thermocouple, Alfa Aesar™
Chemical Name or Material: Platinum CAS: 7440-06-4 Purity Grade: For ISA Type R or S Standard Grade Thermocouple Physical Form: Wire, 0.508mm (0.02 in.) dia. MDL Number: MFCD00011179
Platinum Paste, Alfa Aesar™
Chemical Name or Material: Platinum Name Note: 3% on activated carbon powder Physical Form: Paste MDL Number: MFCD00011179
Platinum foil, 99.99% (metals basis), Alfa Aesar™, Premion™
Chemical Name or Material: Platinum CAS: 7440-06-4 Physical Form: Foil, 0.025mm (0.001 in.) thick Assay Percent Range: 99.99% (Metals basis) MDL Number: MFCD00011179
Platinum wire, 99.9% (metals basis), Alfa Aesar™
Chemical Name or Material: Platinum CAS: 7440-06-4 Physical Form: Wire, 0.127mm dia. Assay Percent Range: 99.9% (Metals basis) MDL Number: MFCD00011179
hydrogen hexachloroplatinate(IV) hydrate, ACS reagent, ACROS Organics™ Available on GSA/VA Contract for Federal Government customers only.
Chemical Name or Material: Hydrogen hexachloroplatinate(IV) hydrate CAS: 26023-84-7 Purity Grade: ACS Reagent Grade Assay: ≥37.50% (Pt) Molecular Formula: H2Cl6Pt·xH2O Linear Formula: H2PtCl6·xH2O Formula Weight: 409.82 Synonym: Hexachloroplatinic acid hydrate
Tetraammineplatinum(II) nitrate, Alfa Aesar™
Chemical Name or Material: Tetraammineplatinum(II) nitrate CAS: 20634-12-2 Physical Form: Powder/Chunks Molecular Formula: H12N6O6Pt Formula Weight: 387.21 MDL Number: MFCD00011624
Platinum Foil, 0.127mm (0.005 in.) thick, 99.9% (Metals basis), Alfa Aesar™
Chemical Name or Material: Platinum CAS: 7440-06-4 Physical Form: Foil, 0.127mm (0.005 in.) thick Assay Percent Range: 99.9% (Metals basis) MDL Number: MFCD00011179
Alfa Aesar™ Platinum wire, 0.404mm dia., annealed, 99.9% (metals basis), Alfa Aesar™
Chemical Name or Material: Platinum CAS: 7440-06-4 MDL Number: MFCD00011179
Platinum Wire, 0.0508mm (0.002 in.) dia., 99.99% (Metals basis), Alfa Aesar™
Chemical Name or Material: Platinum CAS: 7440-06-4 Physical Form: Wire, 0.0508mm (0.002 in.) dia. Assay Percent Range: 99.99% (Metals basis) MDL Number: MFCD00011179
Platinum gauze, 99.9% (metals basis), Alfa Aesar™
Chemical Name or Material: Platinum CAS: 7440-06-4 Physical Form: Gauze Assay Percent Range: 99.9% MDL Number: MFCD00011179
Platinum Wire, 1.0mm (0.04 in.) dia., 99.95% (Metals basis), Alfa Aesar™
Chemical Name or Material: Platinum CAS: 7440-06-4 Physical Form: Wire, 1.0mm (0.04 in.) dia. Assay Percent Range: 99.95% (Metals basis) MDL Number: MFCD00011179
Alfa Aesar™ Platinum black, HiSPEC 1000, Alfa Aesar™
Chemical Name or Material: Platinum black CAS: 7440-06-4 MDL Number: MFCD00011179
  spinner