Rhenium -Re
Rhenium(VII) oxide, 99.99% (metals basis) , Alfa Aesar™
Chemical Name or Material: Rhenium(VII) oxide CAS: 1314-68-7 Physical Form: Crystalline Assay Percent Range: 99.99% (metals basis) Molecular Formula: O7Re2 Formula Weight: 484.4 MDL Number: MFCD00011199
Rhenium Wire, 0.5mm (0.02 in.) dia., 99.97% (Metals basis), Alfa Aesar™
Chemical Name or Material: Rhenium CAS: 7440-15-5 Physical Form: Wire, 0.5mm (0.02 in.) dia. Assay Percent Range: 99.97% (Metals basis) MDL Number: MFCD00011195
Rhenium Sponge/powder, 99.95% (Metals basis), Alfa Aesar™
Chemical Name or Material: Rhenium CAS: 7440-16-6 Physical Form: Sponge/Powder Assay Percent Range: 99.95% (Metals basis) MDL Number: MFCD00011201
Rhenium Foil, 0.25mm (0.01 in.) thick, 99.97% (Metals basis), Alfa Aesar™
Chemical Name or Material: Rhenium CAS: 7440-15-5 Physical Form: Foil, 0.25mm (0.01 in.) thick Assay Percent Range: 99.97% (Metals basis) MDL Number: MFCD00011195
Rhenium(VII) oxide, 99.995% (metals basis), Alfa Aesar™
Chemical Name or Material: Rhenium(VII) oxide CAS: 1314-68-7 Physical Form: Crystals Molecular Formula: O7Re2 Formula Weight: 484.4 MDL Number: MFCD00011199
Rhenium Foil, 0.025mm (0.001 in.) thick, 99.97% (Metals basis), Alfa Aesar™
Chemical Name or Material: Rhenium CAS: 7440-15-5 Physical Form: Foil, 0.025mm (0.001 in.) thick Assay Percent Range: 99.97% (Metals basis) MDL Number: MFCD00011195
Rhenium powder, 99.99%, Alfa Aesar™
Chemical Name or Material: Rhenium CAS: 7440-15-5 Physical Form: Powder Assay Percent Range: 99.99% (Metals basis) MDL Number: MFCD00011195
Rhenium(VI) oxide, 99.9% (metals basis), Alfa Aesar™
Chemical Name or Material: Rhenium(VI) oxide CAS: 1314-28-9 Physical Form: Powder Assay Percent Range: 99.9% (metals basis) Molecular Formula: ReO3 Formula Weight: 236.2 MDL Number: MFCD00016306
Rhenium(III) chloride, Alfa Aesar™
Chemical Name or Material: Rhenium(III) chloride CAS: 13569-63-6 Physical Form: Powder Molecular Formula: ReCl3 Formula Weight: 292.56 MDL Number: MFCD00011196
Oxotrichlorobis(triphenylphosphine)rhenium(V), ACROS Organics™
Chemical Name or Material: Oxotrichlorobis(triphenylphosphine)rhenium(V) CAS: 17442-18-1 Molecular Formula: C36H30Cl3OP2Re Formula Weight: 833.14 MDL Number: MFCD00075554
  spinner