Salicylic Acid
Salicylic Acid, 99+%, ACROS Organics™
Chemical Name or Material: Salicylic acid CAS: 69-72-7 Assay: 99.0 to 101.0% Assay Percent Range: 99+% Molecular Formula: C7H6O3 Formula Weight: 138.12 MDL Number: MFCD00002439 Synonym: 2-Hydroxybenzoic acid
Salicylic Acid (Crystalline/USP), Fisher Chemical
Chemical Name or Material: Salicylic Acid CAS: 69-72-7 Purity Grade: USP Physical Form: Solid Assay: 99.5 to 101.0% (USP) Molecular Formula: C7H6O3 Linear Formula: 2-HOC6H4COOH Formula Weight: 138.12 MDL Number: MFCD00002439 Synonym: 2-Hydroxybenzoic Acid
Salicylic acid, sodium salt, 99+%, ACROS Organics™ Available on GSA/VA Contract for Federal Government customers only.
Chemical Name or Material: Salicylic acid, sodium salt Assay: 99.0 to 101.0% (on dried substance) (Total base) Assay Percent Range: 99+% Molecular Formula: C7H5NaO3 Linear Formula: 2-(HO)C6H4CO2Na Formula Weight: 160.1 MDL Number: MFCD00002440 Synonym: 2-Hydroxybenzoic acid, sodium salt
Salicylic Acid, For analysis ACS, +99%, ACROS Organics™
Chemical Name or Material: Salicylic acid CAS: 69-72-7 Purity Grade: ACS Reagent Grade Assay: ≥99.0% Assay Percent Range: 99+% Molecular Formula: C7H6O3 Formula Weight: 138.12 MDL Number: MFCD00002439 Synonym: 2-Hydroxybenzoic acid
Salicylic Acid (Crystalline Powder/Certified ACS), Fisher Chemical
Chemical Name or Material: Salicylic Acid CAS: 69-72-7 Purity Grade: Certified ACS Grade Physical Form: Solid Assay: ≥99.0% Molecular Formula: C7H6O3 Linear Formula: 2-HOC6H4COOH Formula Weight: 138.12 MDL Number: MFCD00002439 Synonym: 2-Hydroxybenzoic Acid
Salicylic Acid Products for Science Education
Chemical Name or Material: Salicylic Acid CAS: 69-72-7 Purity Grade: Laboratory Grade Assay Percent Range: >99% Linear Formula: 2·HOC6H4COOH Formula Weight: 138.12 Synonym: 2-Hydroxybenzoic Acid
5-Aminosalicylic acid, MP Biomedicals™
Chemical Name or Material: 5-Aminosalicylic acid CAS: 89-57-6 Physical Form: Powder Assay Percent Range: ≥99% Molecular Formula: C7H7NO3 Formula Weight: 153.1 Synonym: 5-amino-2-hydroxybenzoic acid
3-Aminosalicylic acid, MP Biomedicals™
Chemical Name or Material: 3-Aminosalicylic acid CAS: 570-23-0 Physical Form: Crystalline Powder Assay Percent Range: 99.1% (Neutralization) Molecular Formula: C7H7NO3 Formula Weight: 153.1
Salicylic acid, For ACS analysis, 99.68%, MP Biomedicals™
Chemical Name or Material: Salicylic acid Name Note: For ACS analysis CAS: 69-72-7 Physical Form: Powder Assay Percent Range: >99% Molecular Formula: C7H6O3 Formula Weight: 138.12 MDL Number: MFCD00002439 Synonym: 2-Hydroxybenzoic acid
Salicylic Acid, 99.0% min., EMD Millipore Available on GSA/VA Contract for Federal Government customers only.
Chemical Name or Material: Salicylic Acid Name Note: For Synthesis CAS: 69-72-7 Physical Form: Crystals Assay Percent Range: 99.0% min. Molecular Formula: C6H4(OH)COOH Formula Weight: 138.12 Synonym: 2-Hydroxybenzoic acid
  spinner