Scandium -Sc
Scandium(III) oxide, 99.9% (REO), Alfa Aesar™, Reacton™
Chemical Name or Material: Scandium(III) oxide CAS: 12060-08-1 Physical Form: Powder Assay Percent Range: 99.9% Molecular Formula: Sc2O3 Formula Weight: 137.91 MDL Number: MFCD00011223
Scandium(III) oxide, 99.99% (REO), Alfa Aesar™, Reacton™
Chemical Name or Material: Scandium(III) oxide CAS: 12060-08-1 Physical Form: Powder Assay Percent Range: 99.99% Molecular Formula: Sc2O3 Formula Weight: 137.91 MDL Number: MFCD00011223
Scandium Foil, 0.127mm (0.005 in.) thick, 99.9% (REO), Alfa Aesar™
Chemical Name or Material: Scandium CAS: 7440-20-2 Physical Form: Foil, 0.127mm (0.005 in.) thick Assay Percent Range: 99.9% (REO) MDL Number: MFCD00016323
Scandium(III) oxide, 99.998% (metals basis), REM 50ppm, Alfa Aesar™, REacton™
Chemical Name or Material: Scandium(III) oxide Name Note: REM 50ppm max. CAS: 12060-08-1 Physical Form: Powder Assay Percent Range: 99.998% (metals basis) Molecular Formula: Sc2O3 Formula Weight: 137.91 MDL Number: MFCD00011223
Scandium Solid in various forms, 99.9% (REO), Alfa Aesar™
Chemical Name or Material: Scandium Name Note: pieces CAS: 7440-20-2 Physical Form: Solid In Various Forms Assay Percent Range: 99.9% (REO) MDL Number: MFCD00016323
Scandium(III) 2,4-pentanedionate hydrate, Alfa Aesar™
Chemical Name or Material: Scandium(III) 2, 4-pentanedionate hydrate CAS: 699012-88-9 Physical Form: Powder Molecular Formula: C15H21O6Sc·xH2O Formula Weight: 342.26 MDL Number: MFCD00064715 Synonym: Scandium(III) acetylacetonate hydrate
Scandium Pellet, 15.9mm (0.63 in.) dia., , Alfa Aesar™
Chemical Name or Material: Scandium Name Note: arc-cast pellet CAS: 7440-20-2 Physical Form: Pellet, 15.9mm (0.63 in.) dia. MDL Number: MFCD00016323
Scandium(III) oxide, 99.999%, (trace metal basis), ACROS Organics™ Available on GSA/VA Contract for Federal Government customers only.
Chemical Name or Material: Scandium(III) oxide CAS: 12060-08-1 Purity Grade: For trace metal basis analysis Assay Percent Range: 100.00% Molecular Formula: O3Sc2 Linear Formula: SC2O3 Formula Weight: 137.91
Scandium(III) oxide, 99.9%, Acros Organics
Chemical Name or Material: Scandium(III) oxide Name Note: 99.9% CAS: 12060-08-1 Assay Percent Range: 99.90% Molecular Formula: O3Sc2 Linear Formula: ScO3 Formula Weight: 137.91 MDL Number: MFCD00011223
  spinner