Selenous Acid
Selenious acid, 99.99%, (trace metal basis), ACROS Organics™
Chemical Name or Material: Selenious acid CAS: 7783-00-8 Purity Grade: Trace Metal Basis Assay: ≥61% (Se) Assay Percent Range: 99.999% Molecular Formula: H2O3Se Formula Weight: 128.97
Selenous acid, 97%, Alfa Aesar™
Chemical Name or Material: Selenous acid CAS: 7783-00-8 Physical Form: Crystalline Assay Percent Range: 97% Molecular Formula: H2SeO3 Formula Weight: 128.97 MDL Number: MFCD00011331 Synonym: Selenious acid
  spinner