Serine -Ser and Derivatives
N-BOC-D-Serine methyleester, 97%, ACROS Organics™
Chemical Name or Material: N-BOC-D-Serine methyleester CAS: 95715-85-8 CAS Min %: 96.0 CAS Max %: 100.0 Assay Percent Range: 97% Molecular Formula: C9H17NO5 Formula Weight: 219.24 MDL Number: MFCD00270516
L-Serine methyleester hydrochloride, 98%, ACROS Organics™
Chemical Name or Material: L-Serine methyl ester hydrochloride Assay: ≥97.5% Assay Percent Range: 98% Molecular Formula: C4H9NO3·HCl Linear Formula: HOCH2CH(NH2)CO2CH3·HCl Formula Weight: 155.58 MDL Number: MFCD00063680
BOC-L-Serine hydrate, 98%, ACROS Organics™
Chemical Name or Material: BOC-L-Serine hydrate CAS: 3262-72-4 Purity Grade: Technical Grade Assay: ≥97.5% Assay Percent Range: 98% Molecular Formula: C8H15NO5 Linear Formula: HOCH2CH(COOH)NHCOOC(CH3)3· Formula Weight: 205.21 MDL Number: MFCD00150501 Synonym: N-(tert-Butoxycarbonyl)-L-serine hydrate
D-Serine, 98%, ACROS Organics™
Chemical Name or Material: D-Serine Assay Percent Range: 98% Molecular Formula: C3H7NO3 Linear Formula: HOCH2CH(NH2)CO2H Formula Weight: 105.09 MDL Number: MFCD00004269
L-Serine, 99%, ACROS Organics™
Chemical Name or Material: L-Serine Assay: 98.5 to 101.5% (total base) Assay Percent Range: 99% Molecular Formula: C3H7NO3 Linear Formula: HOCH2CH(NH2)CO2H Formula Weight: 105.09 Synonym: Ser
D-Serine methyleester hydrochloride, 98%, ACROS Organics™ Available on GSA/VA Contract for Federal Government customers only.
Chemical Name or Material: D-Serine methyleester hydrochloride Assay: ≥97.5% (Argentometry) Assay Percent Range: 98% Molecular Formula: C4H9NO3·ClH Linear Formula: HOCH2CH(NH2)CO2CH3·HCl Formula Weight: 155.58 MDL Number: MFCD00066121
L-Serinamide hydrochloride, 98%, ACROS Organics™
Chemical Name or Material: L-Serinamide hydrochloride Assay: ≥97.5% (Argentometry) Assay Percent Range: 98% Molecular Formula: C3H8N2O2·HCl Linear Formula: HOCHCH(NH)CONH·HCl Formula Weight: 140.57 MDL Number: MFCD00067560
DL-Serine methyleester hydrochloride, 99%, ACROS Organics™
Chemical Name or Material: DL-Serine methyleester hydrochloride Assay: ≥98.5% Assay Percent Range: 99% Molecular Formula: C4H9NO3·HCl Linear Formula: HOCH2CH(NH2)CO2CH3·HCl Formula Weight: 155.58 MDL Number: MFCD00012593
MSOP, Tocris
Chemical Name or Material: MSOP Assay Percent Range: >98% Molecular Formula: C4H10NO6P Formula Weight: 199.1 Synonym: (RS)-α-Methylserine-O-phosphate
D-Serine, Tocris
Chemical Name or Material: D-Serine CAS: 312-84-5 Assay Percent Range: >98% Molecular Formula: C3H7NO3 Formula Weight: 105.09
L-Serine, Tocris
Chemical Name or Material: L-Serine CAS: 56-45-1 Assay Percent Range: >99% Molecular Formula: C3H7NO3 Formula Weight: 105.09 Synonym: β-Hydroxyalanine
O-Phospho-L-serine, Tocris
Chemical Name or Material: O-Phospho-L-serine CAS: 407-41-0 Assay Percent Range: >98% Molecular Formula: C3H8NO6P Formula Weight: 185.07 Synonym: L-Serine-O-phosphate
L-Serine (White Crystals or Crystalline Powder), Fisher BioReagents
Chemical Name or Material: L-Serine CAS: 56-45-1 Physical Form: Solid Assay: 98.5 to 101.0% Assay Percent Range: 98.5 to 101.0 % Molecular Formula: C3H7NO3 Formula Weight: 105.05
L-Serine, ≥99%, MP Biomedicals™
Chemical Name or Material: L-Serine CAS: 56-45-1 Physical Form: Free-Flowing Powder Assay Percent Range: ≥99% Molecular Formula: C3H7NO3 Formula Weight: 105.09 MDL Number: MFCD00064224 Synonym: (S)-2-amino-3-hydroxypropionic acid
DL-Serine, ≥98%, MP Biomedicals™
Chemical Name or Material: DL-Serine CAS: 302-84-1 Physical Form: Crystalline Powder Assay Percent Range: ≥98% Molecular Formula: C3H7NO3 Formula Weight: 105.1 Synonym: DL-2-Amino-3-hydroxypropionic acid
D-Serine, 100.7%, MP Biomedicals™
Chemical Name or Material: D-Serine CAS: 312-84-5 Physical Form: Crystalline Powder Assay Percent Range: 1.007 Molecular Formula: C3H7NO3 Formula Weight: 105.1 Synonym: βHydroxyalanine
DNP-L-Serine, (Crystalline),MP Biomedicals
Chemical Name or Material: DNP-L-Serine CAS: 1655-64-7 Physical Form: Crystalline Molecular Formula: C9H9N3O7 Formula Weight: 271.2
Phosphatidyl-L-serine, ∽98%, MP Biomedicals™
Chemical Name or Material: Phosphatidyl-L-serine CAS: 51446-62-9 Physical Form: Liquid Assay Percent Range: ∽ 98% Formula Weight: 788 Synonym: 3-sn-Phosphatidyl-L-serine
o-Phospho-L-serine, 98%, MP Biomedicals™
Chemical Name or Material: o-Phospho-L-serine CAS: 407-41-0 Physical Form: Crystalline Powder Assay Percent Range: 0.98 Molecular Formula: C3H8NO6P Formula Weight: 185.1 Synonym: L-o-Phosphoserine
  spinner